Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16990036-16990516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607272 c.2363C>T p.P788L Chr.1: 16990161-16990161 on Build GRCh38
5480937 c.2309A>G p.Q770R Chr.1: 16990230-16990230 on Build GRCh38
1245700 c.2379G>T p.P793P Chr.1: 16990160-16990160 on Build GRCh38
3802889 c.2368G>A p.E790K Chr.1: 16990171-16990171 on Build GRCh38
6607280 c.2392G>A p.V798I Chr.1: 16990132-16990132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139909551 Chr.1: 16990279-16990279 on Build GRCh38
rs754208001 Chr.1: 16990171-16990171 on Build GRCh38
rs140048110 Chr.1: 16990172-16990172 on Build GRCh38
rs199655251 Chr.1: 16990147-16990147 on Build GRCh38
rs147758029 Chr.1: 16990326-16990326 on Build GRCh38
rs530498867 Chr.1: 16990384-16990384 on Build GRCh38
rs376236999 Chr.1: 16990192-16990192 on Build GRCh38
rs56170027 Chr.1: 16990213-16990213 on Build GRCh38
rs941921003 Chr.1: 16990066-16990066 on Build GRCh38
rs759587904 Chr.1: 16990157-16990157 on Build GRCh38
rs755936102 Chr.1: 16990098-16990100 on Build GRCh38
rs137955309 Chr.1: 16990191-16990191 on Build GRCh38
rs144708504 Chr.1: 16990208-16990208 on Build GRCh38
rs758764563 Chr.1: 16990253-16990253 on Build GRCh38
rs1017806715 Chr.1: 16990211-16990211 on Build GRCh38
rs76340633 Chr.1: 16990472-16990472 on Build GRCh38
rs764444063 Chr.1: 16990163-16990163 on Build GRCh38
rs926438143 Chr.1: 16990360-16990360 on Build GRCh38
rs77880760 Chr.1: 16990082-16990082 on Build GRCh38
rs530655624 Chr.1: 16990483-16990483 on Build GRCh38
rs978493820 Chr.1: 16990127-16990127 on Build GRCh38
rs375324126 Chr.1: 16990290-16990290 on Build GRCh38
rs769392537 Chr.1: 16990199-16990199 on Build GRCh38
rs772920644 Chr.1: 16990142-16990142 on Build GRCh38
rs763394181 Chr.1: 16990316-16990316 on Build GRCh38
rs919203517 Chr.1: 16990368-16990368 on Build GRCh38
rs760861030 Chr.1: 16990219-16990219 on Build GRCh38
rs199921859 Chr.1: 16990327-16990327 on Build GRCh38
rs756467036 Chr.1: 16990085-16990085 on Build GRCh38
rs757799277 Chr.1: 16990109-16990109 on Build GRCh38
rs752965372 Chr.1: 16990084-16990084 on Build GRCh38
rs752487771 Chr.1: 16990135-16990135 on Build GRCh38
rs768017868 Chr.1: 16990121-16990121 on Build GRCh38
rs759020707 Chr.1: 16990295-16990295 on Build GRCh38
rs754083353 Chr.1: 16990236-16990236 on Build GRCh38
rs764319654 Chr.1: 16990229-16990229 on Build GRCh38
rs778229092 Chr.1: 16990087-16990087 on Build GRCh38
rs528731161 Chr.1: 16990379-16990379 on Build GRCh38
rs941876051 Chr.1: 16990362-16990362 on Build GRCh38
rs755595496 Chr.1: 16990261-16990261 on Build GRCh38
rs750889358 Chr.1: 16990182-16990182 on Build GRCh38
rs886045580 Chr.1: 16990133-16990133 on Build GRCh38
rs151222153 Chr.1: 16990158-16990158 on Build GRCh38
rs746438512 Chr.1: 16990115-16990115 on Build GRCh38
rs1017231900 Chr.1: 16990083-16990083 on Build GRCh38
rs200924194 Chr.1: 16990276-16990276 on Build GRCh38
rs182748624 Chr.1: 16990160-16990160 on Build GRCh38
rs774130460 Chr.1: 16990154-16990154 on Build GRCh38
rs754704785 Chr.1: 16990290-16990291 on Build GRCh38
rs761882954 Chr.1: 16990303-16990303 on Build GRCh38
rs369917224 Chr.1: 16990313-16990313 on Build GRCh38
rs766227206 Chr.1: 16990077-16990077 on Build GRCh38
rs776907514 Chr.1: 16990301-16990301 on Build GRCh38
rs56164593 Chr.1: 16990312-16990312 on Build GRCh38
rs779373993 Chr.1: 16990180-16990180 on Build GRCh38
rs148374514 Chr.1: 16990214-16990214 on Build GRCh38
rs867014635 Chr.1: 16990200-16990200 on Build GRCh38
rs777422289 Chr.1: 16990206-16990206 on Build GRCh38
rs775080939 Chr.1: 16990461-16990461 on Build GRCh38
rs941242789 Chr.1: 16990470-16990470 on Build GRCh38
rs777302964 Chr.1: 16990268-16990268 on Build GRCh38
rs747707072 Chr.1: 16990297-16990298 on Build GRCh38
rs1051805466 Chr.1: 16990467-16990467 on Build GRCh38
rs55635527 Chr.1: 16990255-16990255 on Build GRCh38
rs765500602 Chr.1: 16990311-16990311 on Build GRCh38
rs748986504 Chr.1: 16990259-16990260 on Build GRCh38
rs750813148 Chr.1: 16990251-16990251 on Build GRCh38
rs980936056 Chr.1: 16990356-16990356 on Build GRCh38
rs766899425 Chr.1: 16990254-16990254 on Build GRCh38
rs980026377 Chr.1: 16990465-16990465 on Build GRCh38
rs767492622 Chr.1: 16990079-16990086 on Build GRCh38
rs7535478 Chr.1: 16990367-16990367 on Build GRCh38
rs775547036 Chr.1: 16990298-16990298 on Build GRCh38
rs760914179 Chr.1: 16990161-16990161 on Build GRCh38
rs779911360 Chr.1: 16990102-16990113 on Build GRCh38
rs778672725 Chr.1: 16990292-16990296 on Build GRCh38
rs374203880 Chr.1: 16990097-16990097 on Build GRCh38
rs770849259 Chr.1: 16990148-16990148 on Build GRCh38
rs1024560145 Chr.1: 16990405-16990405 on Build GRCh38
rs779312019 Chr.1: 16990110-16990110 on Build GRCh38
rs780819882 Chr.1: 16990124-16990124 on Build GRCh38
rs771691843 Chr.1: 16990310-16990311 on Build GRCh38
rs762160087 Chr.1: 16990237-16990237 on Build GRCh38
rs56268651 Chr.1: 16990299-16990299 on Build GRCh38
rs77630788 Chr.1: 16990230-16990230 on Build GRCh38
rs956016608 Chr.1: 16990383-16990383 on Build GRCh38
rs748986186 Chr.1: 16990207-16990207 on Build GRCh38
rs751615609 Chr.1: 16990081-16990081 on Build GRCh38
rs563379285 Chr.1: 16990321-16990321 on Build GRCh38
rs969853772 Chr.1: 16990421-16990421 on Build GRCh38
rs574617285 Chr.1: 16990294-16990294 on Build GRCh38
rs765505077 Chr.1: 16990238-16990238 on Build GRCh38
rs750369195 Chr.1: 16990090-16990094 on Build GRCh38
rs747785443 Chr.1: 16990132-16990132 on Build GRCh38

View Full Product Details