Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17607686-17608181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF10L
Entrez Gene ID
55160
Gene Name
Rho guanine nucleotide exchange factor 10 like
Gene Aliases
GrinchGEF
UniGene
Hs.443460
Gene Chromosome Location
Chr.1: 17520197-17697875 on Build GRCh38
Transcripts
NM_001011722.2 NM_001319837.1 NM_018125.3 NR_137287.1 NR_137288.1 XM_005245923.2 XM_005245925.2 XM_006710728.1 XM_006710729.1 XM_006710731.1 XM_011541691.2 XM_011541692.2 XM_011541693.2 XM_011541695.2 XM_011541696.2 XM_017001617.1 XM_017001618.1 XM_017001619.1 XM_017001620.1 XM_017001621.1 XM_017001622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718079 c.522C>T p.F174F Chr.1: 17607890-17607890 on Build GRCh38
5844643 c.596C>G p.S199C Chr.1: 17607964-17607964 on Build GRCh38
900250 c.590G>A p.R197Q Chr.1: 17607958-17607958 on Build GRCh38
301699 c.523G>T p.E175* Chr.1: 17607891-17607891 on Build GRCh38
3479218 c.573C>T p.V191V Chr.1: 17607941-17607941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985176075 Chr.1: 17607770-17607770 on Build GRCh38
rs760227532 Chr.1: 17607782-17607782 on Build GRCh38
rs116140466 Chr.1: 17607994-17607994 on Build GRCh38
rs977092765 Chr.1: 17607708-17607708 on Build GRCh38
rs551539611 Chr.1: 17608029-17608030 on Build GRCh38
rs961504946 Chr.1: 17607917-17607917 on Build GRCh38
rs939523503 Chr.1: 17608023-17608023 on Build GRCh38
rs763761197 Chr.1: 17607788-17607788 on Build GRCh38
rs147225879 Chr.1: 17607865-17607869 on Build GRCh38
rs536657524 Chr.1: 17607765-17607765 on Build GRCh38
rs866363094 Chr.1: 17607781-17607781 on Build GRCh38
rs773556683 Chr.1: 17607760-17607760 on Build GRCh38
rs190421714 Chr.1: 17607709-17607709 on Build GRCh38
rs527903933 Chr.1: 17607960-17607960 on Build GRCh38
rs1004883525 Chr.1: 17608125-17608125 on Build GRCh38
rs890912725 Chr.1: 17608104-17608104 on Build GRCh38
rs953850987 Chr.1: 17607720-17607720 on Build GRCh38
rs756906500 Chr.1: 17607798-17607798 on Build GRCh38
rs781192667 Chr.1: 17607830-17607830 on Build GRCh38
rs545859145 Chr.1: 17607946-17607946 on Build GRCh38
rs767420688 Chr.1: 17607958-17607958 on Build GRCh38
rs749535452 Chr.1: 17607911-17607911 on Build GRCh38
rs200250179 Chr.1: 17608017-17608017 on Build GRCh38
rs373936331 Chr.1: 17608147-17608147 on Build GRCh38
rs777769203 Chr.1: 17607997-17607997 on Build GRCh38
rs770015721 Chr.1: 17607756-17607756 on Build GRCh38
rs916817946 Chr.1: 17608047-17608047 on Build GRCh38
rs115082470 Chr.1: 17607935-17607935 on Build GRCh38
rs750087513 Chr.1: 17607853-17607853 on Build GRCh38
rs878966067 Chr.1: 17607726-17607726 on Build GRCh38
rs534818421 Chr.1: 17608145-17608145 on Build GRCh38
rs754842481 Chr.1: 17607860-17607860 on Build GRCh38
rs998424279 Chr.1: 17608146-17608146 on Build GRCh38
rs112018664 Chr.1: 17608137-17608137 on Build GRCh38
rs371906812 Chr.1: 17607772-17607772 on Build GRCh38
rs750187607 Chr.1: 17607815-17607815 on Build GRCh38
rs774913499 Chr.1: 17607840-17607840 on Build GRCh38
rs76504639 Chr.1: 17607989-17607989 on Build GRCh38
rs867858137 Chr.1: 17608106-17608106 on Build GRCh38
rs570211122 Chr.1: 17607803-17607803 on Build GRCh38
rs758180018 Chr.1: 17607816-17607816 on Build GRCh38
rs760530219 Chr.1: 17608030-17608033 on Build GRCh38
rs757891123 Chr.1: 17607793-17607801 on Build GRCh38
rs373564688 Chr.1: 17607859-17607859 on Build GRCh38
rs145133878 Chr.1: 17607852-17607852 on Build GRCh38
rs549115688 Chr.1: 17608084-17608084 on Build GRCh38
rs552654872 Chr.1: 17608123-17608123 on Build GRCh38
rs1056671158 Chr.1: 17608042-17608042 on Build GRCh38
rs561573014 Chr.1: 17607993-17607993 on Build GRCh38
rs543152492 Chr.1: 17607928-17607928 on Build GRCh38
rs974966673 Chr.1: 17607902-17607902 on Build GRCh38
rs760176324 Chr.1: 17607844-17607844 on Build GRCh38
rs1038308690 Chr.1: 17608010-17608010 on Build GRCh38
rs753361301 Chr.1: 17607847-17607847 on Build GRCh38
rs769813234 Chr.1: 17607905-17607905 on Build GRCh38
rs879470657 Chr.1: 17607948-17607948 on Build GRCh38
rs754787647 Chr.1: 17607957-17607957 on Build GRCh38
rs142000555 Chr.1: 17607837-17607837 on Build GRCh38
rs761379049 Chr.1: 17607939-17607939 on Build GRCh38
rs201989104 Chr.1: 17607832-17607832 on Build GRCh38
rs79939526 Chr.1: 17608018-17608018 on Build GRCh38
rs748176689 Chr.1: 17607867-17607867 on Build GRCh38
rs550153017 Chr.1: 17607789-17607789 on Build GRCh38
rs140580322 Chr.1: 17607942-17607942 on Build GRCh38
rs752052065 Chr.1: 17607776-17607776 on Build GRCh38
rs770546363 Chr.1: 17608127-17608127 on Build GRCh38
rs771311133 Chr.1: 17607913-17607913 on Build GRCh38
rs777219953 Chr.1: 17607996-17607996 on Build GRCh38
rs768711519 Chr.1: 17607825-17607825 on Build GRCh38
rs532526927 Chr.1: 17608064-17608064 on Build GRCh38
rs781137099 Chr.1: 17607862-17607862 on Build GRCh38
rs948357189 Chr.1: 17608057-17608057 on Build GRCh38
rs751314206 Chr.1: 17607949-17607949 on Build GRCh38
rs565999344 Chr.1: 17608135-17608135 on Build GRCh38
rs114551094 Chr.1: 17607824-17607824 on Build GRCh38
rs201209534 Chr.1: 17607947-17607947 on Build GRCh38
rs964897580 Chr.1: 17607893-17607893 on Build GRCh38
rs758209099 Chr.1: 17607856-17607856 on Build GRCh38
rs576561341 Chr.1: 17607941-17607941 on Build GRCh38
rs887795264 Chr.1: 17608103-17608103 on Build GRCh38
rs768149341 Chr.1: 17607845-17607845 on Build GRCh38
rs556527872 Chr.1: 17607936-17607936 on Build GRCh38
rs770015443 Chr.1: 17607833-17607833 on Build GRCh38
rs913025563 Chr.1: 17607797-17607797 on Build GRCh38
rs774796100 Chr.1: 17607914-17607914 on Build GRCh38
rs369174741 Chr.1: 17607838-17607838 on Build GRCh38
rs796425911 Chr.1: 17608082-17608082 on Build GRCh38
rs778021915 Chr.1: 17607910-17607910 on Build GRCh38
rs113588510 Chr.1: 17607877-17607877 on Build GRCh38
rs1038004808 Chr.1: 17607831-17607831 on Build GRCh38
rs539114174 Chr.1: 17607823-17607823 on Build GRCh38
rs895115951 Chr.1: 17607835-17607835 on Build GRCh38

View Full Product Details