Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17621779-17622287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF10L
Entrez Gene ID
55160
Gene Name
Rho guanine nucleotide exchange factor 10 like
Gene Aliases
GrinchGEF
UniGene
Hs.443460
Gene Chromosome Location
Chr.1: 17520197-17697875 on Build GRCh38
Transcripts
NM_001011722.2 NM_001319837.1 NM_001319838.1 NM_001328124.1 NM_018125.3 NR_137287.1 NR_137288.1 XM_005245923.2 XM_005245925.2 XM_005245927.2 XM_005245929.2 XM_006710728.1 XM_006710729.1 XM_006710731.1 XM_011541691.2 XM_011541692.2 XM_011541693.2 XM_011541695.2 XM_011541696.2 XM_017001617.1 XM_017001618.1 XM_017001619.1 XM_017001620.1 XM_017001621.1 XM_017001622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
130457 c.994G>T p.V332L Chr.1: 17621915-17621915 on Build GRCh38
5992181 c.328G>A p.V110M Chr.1: 17621915-17621915 on Build GRCh38
4412580 c.246C>T p.I82I Chr.1: 17621893-17621893 on Build GRCh38
177747 c.982G>A p.E328K Chr.1: 17621903-17621903 on Build GRCh38
3479239 c.950G>A p.R317Q Chr.1: 17621871-17621871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142465382 Chr.1: 17621981-17621981 on Build GRCh38
rs949331100 Chr.1: 17622199-17622199 on Build GRCh38
rs146595527 Chr.1: 17621914-17621914 on Build GRCh38
rs921648526 Chr.1: 17622017-17622017 on Build GRCh38
rs139730445 Chr.1: 17622148-17622148 on Build GRCh38
rs925397489 Chr.1: 17622127-17622128 on Build GRCh38
rs887613070 Chr.1: 17622179-17622179 on Build GRCh38
rs139501792 Chr.1: 17621902-17621902 on Build GRCh38
rs767517904 Chr.1: 17621859-17621859 on Build GRCh38
rs987952708 Chr.1: 17622133-17622133 on Build GRCh38
rs535441309 Chr.1: 17621854-17621854 on Build GRCh38
rs562643262 Chr.1: 17622072-17622072 on Build GRCh38
rs542253874 Chr.1: 17622021-17622021 on Build GRCh38
rs767330608 Chr.1: 17621978-17621978 on Build GRCh38
rs1044716147 Chr.1: 17622201-17622201 on Build GRCh38
rs576328375 Chr.1: 17621844-17621844 on Build GRCh38
rs553740362 Chr.1: 17622004-17622004 on Build GRCh38
rs904894658 Chr.1: 17622234-17622234 on Build GRCh38
rs774992404 Chr.1: 17621830-17621830 on Build GRCh38
rs142503205 Chr.1: 17621894-17621894 on Build GRCh38
rs183634180 Chr.1: 17622126-17622126 on Build GRCh38
rs964143619 Chr.1: 17622103-17622103 on Build GRCh38
rs1005160177 Chr.1: 17622185-17622185 on Build GRCh38
rs914733516 Chr.1: 17622088-17622088 on Build GRCh38
rs1025865533 Chr.1: 17622026-17622026 on Build GRCh38
rs112117398 Chr.1: 17621842-17621842 on Build GRCh38
rs1014812086 Chr.1: 17622247-17622247 on Build GRCh38
rs992866196 Chr.1: 17622230-17622230 on Build GRCh38
rs533422864 Chr.1: 17621819-17621819 on Build GRCh38
rs777826378 Chr.1: 17621868-17621868 on Build GRCh38
rs751367085 Chr.1: 17621857-17621857 on Build GRCh38
rs772961177 Chr.1: 17621843-17621843 on Build GRCh38
rs754179196 Chr.1: 17621871-17621871 on Build GRCh38
rs376652146 Chr.1: 17621915-17621915 on Build GRCh38
rs114377192 Chr.1: 17622011-17622011 on Build GRCh38
rs955628963 Chr.1: 17622034-17622034 on Build GRCh38
rs1055821554 Chr.1: 17622141-17622141 on Build GRCh38
rs746321587 Chr.1: 17621831-17621831 on Build GRCh38
rs1048979415 Chr.1: 17622163-17622163 on Build GRCh38
rs375077212 Chr.1: 17621834-17621834 on Build GRCh38
rs760665400 Chr.1: 17621984-17621984 on Build GRCh38
rs771460943 Chr.1: 17621825-17621825 on Build GRCh38
rs770572310 Chr.1: 17621931-17621931 on Build GRCh38
rs762536930 Chr.1: 17621930-17621930 on Build GRCh38
rs371515222 Chr.1: 17621903-17621903 on Build GRCh38
rs774089879 Chr.1: 17621945-17621945 on Build GRCh38
rs757680865 Chr.1: 17621878-17621878 on Build GRCh38
rs555835738 Chr.1: 17621863-17621863 on Build GRCh38
rs759393762 Chr.1: 17621952-17621952 on Build GRCh38
rs1018720223 Chr.1: 17622097-17622097 on Build GRCh38
rs764675707 Chr.1: 17621972-17621973 on Build GRCh38
rs112046373 Chr.1: 17621815-17621818 on Build GRCh38
rs994347979 Chr.1: 17622023-17622023 on Build GRCh38
rs546862166 Chr.1: 17621820-17621820 on Build GRCh38
rs755700856 Chr.1: 17622132-17622132 on Build GRCh38
rs995576651 Chr.1: 17622157-17622157 on Build GRCh38
rs747312138 Chr.1: 17621837-17621837 on Build GRCh38
rs761527814 Chr.1: 17621836-17621836 on Build GRCh38
rs946230263 Chr.1: 17622104-17622104 on Build GRCh38
rs764289679 Chr.1: 17621989-17621989 on Build GRCh38
rs139721173 Chr.1: 17621810-17621811 on Build GRCh38
rs759398693 Chr.1: 17621858-17621858 on Build GRCh38
rs928888314 Chr.1: 17622131-17622131 on Build GRCh38
rs531288438 Chr.1: 17622081-17622081 on Build GRCh38
rs1027500105 Chr.1: 17621906-17621906 on Build GRCh38
rs779355456 Chr.1: 17621893-17621893 on Build GRCh38
rs983366607 Chr.1: 17621939-17621939 on Build GRCh38

View Full Product Details