Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19312113-19312650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR7A2
Entrez Gene ID
8574
Gene Name
aldo-keto reductase family 7 member A2
Gene Aliases
AFAR, AFAR1, AFB1-AR1, AKR7
UniGene
Hs.571886
Gene Chromosome Location
Chr.1: 19302708-19312146 on Build GRCh38
Transcripts
NM_001320979.1 NM_003689.3 NM_001040125.1 NM_001040126.1 NM_001287531.1 NM_017765.2 NR_109848.1 XM_005245916.2 XM_006710710.2 XM_006710711.2 XM_011541634.2

Gene Symbol
PQLC2
Entrez Gene ID
54896
Gene Name
PQ loop repeat containing 2
Gene Aliases
-
UniGene
Hs.647620
Gene Chromosome Location
Chr.1: 19312246-19332611 on Build GRCh38
Transcripts
NM_001320979.1 NM_003689.3 NM_001040125.1 NM_001040126.1 NM_001287531.1 NM_017765.2 NR_109848.1 XM_005245916.2 XM_006710710.2 XM_006710711.2 XM_011541634.2

SNP Detail

SNP ID Chromosome Location
rs930677060 Chr.1: 19312608-19312608 on Build GRCh38
rs989562619 Chr.1: 19312318-19312318 on Build GRCh38
rs1042925681 Chr.1: 19312249-19312249 on Build GRCh38
rs775613690 Chr.1: 19312135-19312139 on Build GRCh38
rs535857895 Chr.1: 19312288-19312288 on Build GRCh38
rs1019833233 Chr.1: 19312281-19312281 on Build GRCh38
rs560724662 Chr.1: 19312131-19312131 on Build GRCh38
rs906581645 Chr.1: 19312153-19312154 on Build GRCh38
rs564380433 Chr.1: 19312450-19312450 on Build GRCh38
rs953773257 Chr.1: 19312465-19312465 on Build GRCh38
rs112996273 Chr.1: 19312219-19312219 on Build GRCh38
rs887263786 Chr.1: 19312274-19312274 on Build GRCh38
rs983484622 Chr.1: 19312570-19312570 on Build GRCh38
rs550294768 Chr.1: 19312514-19312514 on Build GRCh38
rs966393339 Chr.1: 19312529-19312529 on Build GRCh38
rs538206322 Chr.1: 19312207-19312207 on Build GRCh38
rs542476206 Chr.1: 19312376-19312376 on Build GRCh38
rs907670726 Chr.1: 19312576-19312576 on Build GRCh38
rs953820430 Chr.1: 19312343-19312343 on Build GRCh38
rs920260393 Chr.1: 19312370-19312370 on Build GRCh38
rs2231194 Chr.1: 19312594-19312594 on Build GRCh38
rs554582139 Chr.1: 19312244-19312244 on Build GRCh38
rs996842392 Chr.1: 19312289-19312289 on Build GRCh38
rs965563030 Chr.1: 19312287-19312287 on Build GRCh38
rs990733960 Chr.1: 19312496-19312496 on Build GRCh38
rs930338149 Chr.1: 19312553-19312553 on Build GRCh38
rs966545181 Chr.1: 19312356-19312356 on Build GRCh38
rs550151827 Chr.1: 19312212-19312212 on Build GRCh38
rs921351155 Chr.1: 19312196-19312196 on Build GRCh38
rs965363303 Chr.1: 19312160-19312160 on Build GRCh38
rs930411235 Chr.1: 19312372-19312372 on Build GRCh38
rs945300505 Chr.1: 19312230-19312230 on Build GRCh38
rs1011296895 Chr.1: 19312444-19312444 on Build GRCh38
rs895217451 Chr.1: 19312373-19312373 on Build GRCh38
rs939530819 Chr.1: 19312369-19312369 on Build GRCh38
rs2473808 Chr.1: 19312389-19312389 on Build GRCh38
rs16860346 Chr.1: 19312149-19312149 on Build GRCh38
rs994872325 Chr.1: 19312163-19312163 on Build GRCh38
rs1019596450 Chr.1: 19312158-19312158 on Build GRCh38
rs937906607 Chr.1: 19312367-19312367 on Build GRCh38
rs927866074 Chr.1: 19312365-19312365 on Build GRCh38
rs901124861 Chr.1: 19312428-19312428 on Build GRCh38
rs1049068545 Chr.1: 19312273-19312273 on Build GRCh38
rs572963992 Chr.1: 19312257-19312257 on Build GRCh38
rs977026304 Chr.1: 19312335-19312335 on Build GRCh38
rs192385011 Chr.1: 19312604-19312604 on Build GRCh38
rs1015503849 Chr.1: 19312423-19312423 on Build GRCh38
rs932866578 Chr.1: 19312149-19312155 on Build GRCh38
rs1034057000 Chr.1: 19312159-19312160 on Build GRCh38
rs533977541 Chr.1: 19312282-19312282 on Build GRCh38
rs146897332 Chr.1: 19312497-19312497 on Build GRCh38
rs1004504360 Chr.1: 19312401-19312401 on Build GRCh38
rs901467268 Chr.1: 19312397-19312397 on Build GRCh38
rs932790396 Chr.1: 19312620-19312620 on Build GRCh38
rs544760396 Chr.1: 19312325-19312325 on Build GRCh38
rs975352171 Chr.1: 19312179-19312179 on Build GRCh38
rs913952874 Chr.1: 19312336-19312336 on Build GRCh38
rs546261703 Chr.1: 19312155-19312155 on Build GRCh38
rs1041226525 Chr.1: 19312596-19312596 on Build GRCh38
rs1047491922 Chr.1: 19312375-19312375 on Build GRCh38
rs908387261 Chr.1: 19312227-19312227 on Build GRCh38
rs1028027177 Chr.1: 19312341-19312341 on Build GRCh38
rs966743276 Chr.1: 19312457-19312457 on Build GRCh38
rs1004001344 Chr.1: 19312256-19312256 on Build GRCh38
rs1004313845 Chr.1: 19312279-19312279 on Build GRCh38
rs958394516 Chr.1: 19312314-19312314 on Build GRCh38
rs981978410 Chr.1: 19312458-19312458 on Build GRCh38
rs945661845 Chr.1: 19312246-19312246 on Build GRCh38
rs3830823 Chr.1: 19312165-19312166 on Build GRCh38
rs927772660 Chr.1: 19312460-19312460 on Build GRCh38
rs1040648098 Chr.1: 19312402-19312402 on Build GRCh38
rs568787924 Chr.1: 19312233-19312233 on Build GRCh38
rs959304349 Chr.1: 19312468-19312468 on Build GRCh38
rs913724136 Chr.1: 19312223-19312223 on Build GRCh38
rs1004341699 Chr.1: 19312378-19312378 on Build GRCh38
rs552363949 Chr.1: 19312438-19312438 on Build GRCh38
rs1054757368 Chr.1: 19312236-19312236 on Build GRCh38
rs951751698 Chr.1: 19312142-19312142 on Build GRCh38
rs879843338 Chr.1: 19312396-19312396 on Build GRCh38
rs952628226 Chr.1: 19312195-19312195 on Build GRCh38
rs571014745 Chr.1: 19312175-19312175 on Build GRCh38
rs1028318949 Chr.1: 19312290-19312290 on Build GRCh38
rs536532392 Chr.1: 19312235-19312235 on Build GRCh38
rs6143148 Chr.1: 19312149-19312150 on Build GRCh38
rs1021881377 Chr.1: 19312517-19312517 on Build GRCh38
rs906703956 Chr.1: 19312251-19312251 on Build GRCh38
rs574777414 Chr.1: 19312368-19312368 on Build GRCh38
rs927835062 Chr.1: 19312234-19312234 on Build GRCh38
rs997044612 Chr.1: 19312415-19312415 on Build GRCh38
rs1028768096 Chr.1: 19312429-19312429 on Build GRCh38
rs527840079 Chr.1: 19312424-19312424 on Build GRCh38
rs559051701 Chr.1: 19312286-19312286 on Build GRCh38
rs928217360 Chr.1: 19312565-19312565 on Build GRCh38
rs140093801 Chr.1: 19312152-19312152 on Build GRCh38
rs562851474 Chr.1: 19312357-19312357 on Build GRCh38
rs994587542 Chr.1: 19312265-19312265 on Build GRCh38

View Full Product Details