Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21218472-21218981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE1
Entrez Gene ID
1889
Gene Name
endothelin converting enzyme 1
Gene Aliases
ECE
UniGene
Hs.195080
Gene Chromosome Location
Chr.1: 21217247-21345541 on Build GRCh38
Transcripts
NM_001113347.1 NM_001113348.1 NM_001113349.1 NM_001397.2 XM_006710398.2 XM_011540872.2 XM_011540873.2 XM_017000511.1

SNP Detail

SNP ID Chromosome Location
rs935240257 Chr.1: 21218735-21218735 on Build GRCh38
rs910935760 Chr.1: 21218945-21218945 on Build GRCh38
rs1054540682 Chr.1: 21218679-21218679 on Build GRCh38
rs869085513 Chr.1: 21218763-21218764 on Build GRCh38
rs1003390773 Chr.1: 21218761-21218761 on Build GRCh38
rs922501743 Chr.1: 21218729-21218729 on Build GRCh38
rs1052381923 Chr.1: 21218738-21218738 on Build GRCh38
rs781741958 Chr.1: 21218948-21218948 on Build GRCh38
rs891835052 Chr.1: 21218706-21218706 on Build GRCh38
rs1010206979 Chr.1: 21218740-21218740 on Build GRCh38
rs1041279291 Chr.1: 21218719-21218720 on Build GRCh38
rs201028418 Chr.1: 21218711-21218711 on Build GRCh38
rs530105981 Chr.1: 21218541-21218541 on Build GRCh38
rs530363592 Chr.1: 21218940-21218940 on Build GRCh38
rs952409358 Chr.1: 21218637-21218637 on Build GRCh38
rs1003673367 Chr.1: 21218762-21218763 on Build GRCh38
rs564544955 Chr.1: 21218921-21218921 on Build GRCh38
rs190154452 Chr.1: 21218803-21218803 on Build GRCh38
rs1004130649 Chr.1: 21218560-21218560 on Build GRCh38
rs754810032 Chr.1: 21218500-21218500 on Build GRCh38
rs76178796 Chr.1: 21218935-21218935 on Build GRCh38
rs893847871 Chr.1: 21218501-21218501 on Build GRCh38
rs548688203 Chr.1: 21218564-21218564 on Build GRCh38
rs912166605 Chr.1: 21218856-21218856 on Build GRCh38
rs950397302 Chr.1: 21218833-21218833 on Build GRCh38
rs556843320 Chr.1: 21218647-21218647 on Build GRCh38
rs78774900 Chr.1: 21218680-21218680 on Build GRCh38
rs975001848 Chr.1: 21218866-21218866 on Build GRCh38
rs577096683 Chr.1: 21218931-21218931 on Build GRCh38
rs184697574 Chr.1: 21218747-21218747 on Build GRCh38
rs952354379 Chr.1: 21218851-21218851 on Build GRCh38
rs972327707 Chr.1: 21218609-21218609 on Build GRCh38
rs936105983 Chr.1: 21218648-21218648 on Build GRCh38
rs541946628 Chr.1: 21218951-21218951 on Build GRCh38
rs1033959193 Chr.1: 21218792-21218792 on Build GRCh38
rs28368055 Chr.1: 21218571-21218571 on Build GRCh38
rs573481193 Chr.1: 21218638-21218638 on Build GRCh38
rs575430323 Chr.1: 21218741-21218741 on Build GRCh38
rs142315334 Chr.1: 21218834-21218834 on Build GRCh38
rs959217217 Chr.1: 21218793-21218793 on Build GRCh38
rs1010984628 Chr.1: 21218514-21218514 on Build GRCh38
rs1045143121 Chr.1: 21218759-21218759 on Build GRCh38
rs912114052 Chr.1: 21218601-21218601 on Build GRCh38
rs566746968 Chr.1: 21218566-21218566 on Build GRCh38
rs552611822 Chr.1: 21218604-21218604 on Build GRCh38
rs752372158 Chr.1: 21218587-21218587 on Build GRCh38
rs555962219 Chr.1: 21218611-21218611 on Build GRCh38
rs147642303 Chr.1: 21218798-21218798 on Build GRCh38
rs544522606 Chr.1: 21218932-21218932 on Build GRCh38
rs1023748134 Chr.1: 21218525-21218525 on Build GRCh38
rs966669189 Chr.1: 21218859-21218859 on Build GRCh38

View Full Product Details