Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21219349-21219847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE1
Entrez Gene ID
1889
Gene Name
endothelin converting enzyme 1
Gene Aliases
ECE
UniGene
Hs.195080
Gene Chromosome Location
Chr.1: 21217247-21345541 on Build GRCh38
Transcripts
NM_001113347.1 NM_001113348.1 NM_001113349.1 NM_001397.2 XM_006710398.2 XM_011540872.2 XM_011540873.2 XM_017000511.1

SNP Detail

SNP ID Chromosome Location
rs919299788 Chr.1: 21219671-21219671 on Build GRCh38
rs554478099 Chr.1: 21219398-21219398 on Build GRCh38
rs1020488165 Chr.1: 21219627-21219627 on Build GRCh38
rs146140052 Chr.1: 21219659-21219659 on Build GRCh38
rs116537934 Chr.1: 21219623-21219623 on Build GRCh38
rs987926005 Chr.1: 21219591-21219592 on Build GRCh38
rs544572182 Chr.1: 21219653-21219653 on Build GRCh38
rs762017322 Chr.1: 21219422-21219422 on Build GRCh38
rs563416561 Chr.1: 21219737-21219737 on Build GRCh38
rs539098899 Chr.1: 21219736-21219736 on Build GRCh38
rs1017880800 Chr.1: 21219469-21219469 on Build GRCh38
rs886304845 Chr.1: 21219759-21219759 on Build GRCh38
rs558097516 Chr.1: 21219607-21219607 on Build GRCh38
rs1029791236 Chr.1: 21219557-21219557 on Build GRCh38
rs939389960 Chr.1: 21219374-21219374 on Build GRCh38
rs995816049 Chr.1: 21219535-21219535 on Build GRCh38
rs572630539 Chr.1: 21219396-21219396 on Build GRCh38
rs529041783 Chr.1: 21219792-21219792 on Build GRCh38
rs777584772 Chr.1: 21219668-21219668 on Build GRCh38
rs114232246 Chr.1: 21219660-21219660 on Build GRCh38
rs181637959 Chr.1: 21219540-21219540 on Build GRCh38
rs531786776 Chr.1: 21219764-21219764 on Build GRCh38
rs867348639 Chr.1: 21219822-21219822 on Build GRCh38
rs532844615 Chr.1: 21219423-21219423 on Build GRCh38
rs113340102 Chr.1: 21219784-21219784 on Build GRCh38
rs546353782 Chr.1: 21219743-21219743 on Build GRCh38
rs957579491 Chr.1: 21219591-21219591 on Build GRCh38
rs996257020 Chr.1: 21219785-21219785 on Build GRCh38
rs899474547 Chr.1: 21219539-21219540 on Build GRCh38
rs1055460478 Chr.1: 21219417-21219417 on Build GRCh38
rs916499275 Chr.1: 21219713-21219713 on Build GRCh38
rs149292526 Chr.1: 21219787-21219787 on Build GRCh38
rs1007754795 Chr.1: 21219460-21219460 on Build GRCh38
rs372073084 Chr.1: 21219809-21219809 on Build GRCh38
rs901512412 Chr.1: 21219525-21219525 on Build GRCh38
rs892167883 Chr.1: 21219782-21219782 on Build GRCh38
rs1048854371 Chr.1: 21219450-21219450 on Build GRCh38
rs1040528092 Chr.1: 21219783-21219783 on Build GRCh38
rs1004626721 Chr.1: 21219504-21219504 on Build GRCh38
rs937350867 Chr.1: 21219410-21219410 on Build GRCh38
rs566185770 Chr.1: 21219419-21219419 on Build GRCh38
rs929385227 Chr.1: 21219699-21219699 on Build GRCh38
rs1018686186 Chr.1: 21219524-21219524 on Build GRCh38
rs542361350 Chr.1: 21219669-21219669 on Build GRCh38
rs564451188 Chr.1: 21219751-21219751 on Build GRCh38
rs956443573 Chr.1: 21219585-21219585 on Build GRCh38
rs765051785 Chr.1: 21219656-21219656 on Build GRCh38
rs940491947 Chr.1: 21219760-21219760 on Build GRCh38
rs3026908 Chr.1: 21219821-21219821 on Build GRCh38
rs138610036 Chr.1: 21219725-21219725 on Build GRCh38
rs981076908 Chr.1: 21219647-21219647 on Build GRCh38
rs909369085 Chr.1: 21219716-21219716 on Build GRCh38
rs767467285 Chr.1: 21219454-21219454 on Build GRCh38

View Full Product Details