Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40440965-40441448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
UniGene
Hs.22109
Gene Chromosome Location
Chr.15: 40439721-40468242 on Build GRCh38
Transcripts
NM_001301132.1 NM_014952.4 NR_125372.1 XM_011521369.2

SNP Detail

SNP ID Chromosome Location
rs1054871362 Chr.15: 40441352-40441352 on Build GRCh38
rs577473118 Chr.15: 40441237-40441237 on Build GRCh38
rs1009773879 Chr.15: 40441001-40441001 on Build GRCh38
rs932874401 Chr.15: 40441295-40441295 on Build GRCh38
rs926272818 Chr.15: 40441026-40441026 on Build GRCh38
rs971020560 Chr.15: 40441008-40441008 on Build GRCh38
rs544228274 Chr.15: 40441315-40441315 on Build GRCh38
rs983600026 Chr.15: 40441250-40441250 on Build GRCh38
rs983245525 Chr.15: 40441126-40441126 on Build GRCh38
rs899828913 Chr.15: 40441072-40441072 on Build GRCh38
rs1042789404 Chr.15: 40441334-40441334 on Build GRCh38
rs546443179 Chr.15: 40441403-40441403 on Build GRCh38
rs970609786 Chr.15: 40441081-40441081 on Build GRCh38
rs906350958 Chr.15: 40441161-40441161 on Build GRCh38
rs1004514866 Chr.15: 40441368-40441368 on Build GRCh38
rs996818426 Chr.15: 40441077-40441077 on Build GRCh38
rs959290208 Chr.15: 40441086-40441086 on Build GRCh38
rs1018664904 Chr.15: 40441258-40441258 on Build GRCh38
rs959070354 Chr.15: 40441031-40441031 on Build GRCh38
rs915224482 Chr.15: 40441320-40441323 on Build GRCh38
rs944255210 Chr.15: 40441131-40441131 on Build GRCh38
rs554412068 Chr.15: 40441148-40441148 on Build GRCh38
rs1047646831 Chr.15: 40441047-40441047 on Build GRCh38
rs1009666337 Chr.15: 40441079-40441079 on Build GRCh38
rs950508548 Chr.15: 40441117-40441117 on Build GRCh38
rs1028475193 Chr.15: 40441422-40441429 on Build GRCh38
rs945430491 Chr.15: 40441326-40441327 on Build GRCh38
rs575228223 Chr.15: 40441294-40441294 on Build GRCh38
rs1049083110 Chr.15: 40441054-40441072 on Build GRCh38
rs908703207 Chr.15: 40441129-40441129 on Build GRCh38
rs893869245 Chr.15: 40441078-40441079 on Build GRCh38
rs895038321 Chr.15: 40441188-40441188 on Build GRCh38
rs895007350 Chr.15: 40441356-40441356 on Build GRCh38
rs1033491689 Chr.15: 40441188-40441189 on Build GRCh38
rs565020899 Chr.15: 40441247-40441247 on Build GRCh38
rs973119283 Chr.15: 40441034-40441034 on Build GRCh38
rs1003867076 Chr.15: 40441084-40441084 on Build GRCh38
rs568911086 Chr.15: 40441233-40441234 on Build GRCh38
rs1021192114 Chr.15: 40441080-40441080 on Build GRCh38
rs976892773 Chr.15: 40441283-40441283 on Build GRCh38
rs908739668 Chr.15: 40441053-40441053 on Build GRCh38
rs939451341 Chr.15: 40441340-40441340 on Build GRCh38
rs966192264 Chr.15: 40441276-40441277 on Build GRCh38
rs1003343339 Chr.15: 40441167-40441167 on Build GRCh38
rs920274540 Chr.15: 40441116-40441116 on Build GRCh38
rs866230804 Chr.15: 40441079-40441079 on Build GRCh38
rs1014948494 Chr.15: 40441052-40441052 on Build GRCh38
rs546411036 Chr.15: 40441241-40441241 on Build GRCh38
rs1053787051 Chr.15: 40441078-40441078 on Build GRCh38
rs994742413 Chr.15: 40441194-40441194 on Build GRCh38
rs929089371 Chr.15: 40441044-40441044 on Build GRCh38
rs958119562 Chr.15: 40440994-40440995 on Build GRCh38
rs893834455 Chr.15: 40441146-40441146 on Build GRCh38
rs560686806 Chr.15: 40441339-40441339 on Build GRCh38
rs1041361138 Chr.15: 40441057-40441057 on Build GRCh38
rs1033591261 Chr.15: 40441085-40441085 on Build GRCh38
rs973020100 Chr.15: 40441110-40441110 on Build GRCh38
rs921479417 Chr.15: 40441132-40441133 on Build GRCh38
rs1004945953 Chr.15: 40441002-40441002 on Build GRCh38
rs974516352 Chr.15: 40441416-40441416 on Build GRCh38
rs530023605 Chr.15: 40441364-40441364 on Build GRCh38
rs1027259547 Chr.15: 40441199-40441199 on Build GRCh38
rs1032464240 Chr.15: 40440985-40440985 on Build GRCh38
rs982078546 Chr.15: 40441018-40441018 on Build GRCh38
rs965792300 Chr.15: 40441415-40441415 on Build GRCh38
rs1053755938 Chr.15: 40441138-40441138 on Build GRCh38
rs1019047619 Chr.15: 40441374-40441374 on Build GRCh38
rs932905417 Chr.15: 40441134-40441134 on Build GRCh38
rs994712071 Chr.15: 40441359-40441359 on Build GRCh38
rs920305779 Chr.15: 40441043-40441043 on Build GRCh38
rs1042515061 Chr.15: 40441155-40441155 on Build GRCh38

View Full Product Details