Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40472688-40473196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST14
Entrez Gene ID
113189
Gene Name
carbohydrate sulfotransferase 14
Gene Aliases
ATCS, D4ST1, EDSMC1, HNK1ST
UniGene
Hs.442449
Gene Chromosome Location
Chr.15: 40470961-40473158 on Build GRCh38
Transcripts
NM_130468.3

SNP Detail

SNP ID Chromosome Location
rs1047733069 Chr.15: 40472743-40472743 on Build GRCh38
rs1031401581 Chr.15: 40472848-40472848 on Build GRCh38
rs974201881 Chr.15: 40472925-40472925 on Build GRCh38
rs667623 Chr.15: 40473161-40473161 on Build GRCh38
rs886466232 Chr.15: 40472753-40472753 on Build GRCh38
rs373363464 Chr.15: 40472888-40472888 on Build GRCh38
rs1048319400 Chr.15: 40473020-40473020 on Build GRCh38
rs1003545738 Chr.15: 40472756-40472756 on Build GRCh38
rs949314368 Chr.15: 40473132-40473133 on Build GRCh38
rs760493666 Chr.15: 40472931-40472931 on Build GRCh38
rs149213510 Chr.15: 40472847-40472847 on Build GRCh38
rs915919863 Chr.15: 40472849-40472849 on Build GRCh38
rs1022397597 Chr.15: 40472816-40472816 on Build GRCh38
rs186655194 Chr.15: 40472916-40472916 on Build GRCh38
rs907947628 Chr.15: 40473023-40473023 on Build GRCh38
rs899249367 Chr.15: 40472710-40472710 on Build GRCh38
rs911819296 Chr.15: 40472910-40472910 on Build GRCh38
rs987480829 Chr.15: 40472966-40472966 on Build GRCh38
rs540930052 Chr.15: 40473086-40473086 on Build GRCh38
rs968691961 Chr.15: 40473040-40473040 on Build GRCh38
rs191358365 Chr.15: 40472975-40472975 on Build GRCh38
rs930697486 Chr.15: 40472730-40472730 on Build GRCh38
rs1056353530 Chr.15: 40473090-40473090 on Build GRCh38
rs373828117 Chr.15: 40473100-40473100 on Build GRCh38
rs969485354 Chr.15: 40472875-40472875 on Build GRCh38
rs555477789 Chr.15: 40473081-40473081 on Build GRCh38
rs376938893 Chr.15: 40473154-40473154 on Build GRCh38
rs895370012 Chr.15: 40472809-40472809 on Build GRCh38
rs930478630 Chr.15: 40473014-40473014 on Build GRCh38
rs76204446 Chr.15: 40472739-40472739 on Build GRCh38
rs987147201 Chr.15: 40472893-40472893 on Build GRCh38
rs1052246023 Chr.15: 40473093-40473093 on Build GRCh38
rs78703927 Chr.15: 40472742-40472742 on Build GRCh38
rs951635039 Chr.15: 40472962-40472962 on Build GRCh38
rs571878310 Chr.15: 40472764-40472764 on Build GRCh38
rs968453655 Chr.15: 40472827-40472827 on Build GRCh38
rs559444266 Chr.15: 40472815-40472815 on Build GRCh38
rs962870491 Chr.15: 40472922-40472922 on Build GRCh38
rs1013933453 Chr.15: 40472732-40472732 on Build GRCh38
rs925331740 Chr.15: 40473085-40473085 on Build GRCh38

View Full Product Details