Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40767122-40767615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC17
Entrez Gene ID
55192
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Gene Aliases
-
UniGene
Hs.511069
Gene Chromosome Location
Chr.15: 40767448-40807478 on Build GRCh38
Transcripts
XM_017022401.1 NM_005258.2

Gene Symbol
GCHFR
Entrez Gene ID
2644
Gene Name
GTP cyclohydrolase I feedback regulator
Gene Aliases
GFRP, HsT16933, P35
UniGene
Hs.631717
Gene Chromosome Location
Chr.15: 40764087-40767713 on Build GRCh38
Transcripts
XM_017022401.1 NM_005258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74737 c.158C>T p.P53L Chr.15: 40767252-40767252 on Build GRCh38
187351 c.185G>A p.R62H Chr.15: 40767279-40767279 on Build GRCh38
5016466 c.206_207delGC p.S69fs*>16 Chr.15: 40767300-40767301 on Build GRCh38
6994678 c.192C>A p.G64G Chr.15: 40767286-40767286 on Build GRCh38
4686618 c.213delG p.V73fs*>12 Chr.15: 40767307-40767307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746723138 Chr.15: 40767274-40767274 on Build GRCh38
rs376132308 Chr.15: 40767357-40767357 on Build GRCh38
rs757047805 Chr.15: 40767303-40767303 on Build GRCh38
rs139599649 Chr.15: 40767238-40767238 on Build GRCh38
rs146242086 Chr.15: 40767306-40767306 on Build GRCh38
rs941025559 Chr.15: 40767247-40767247 on Build GRCh38
rs149719928 Chr.15: 40767283-40767283 on Build GRCh38
rs1031606375 Chr.15: 40767512-40767512 on Build GRCh38
rs367958027 Chr.15: 40767241-40767241 on Build GRCh38
rs901965817 Chr.15: 40767430-40767430 on Build GRCh38
rs761724887 Chr.15: 40767201-40767201 on Build GRCh38
rs78081362 Chr.15: 40767456-40767456 on Build GRCh38
rs957371665 Chr.15: 40767144-40767144 on Build GRCh38
rs998012862 Chr.15: 40767435-40767435 on Build GRCh38
rs2301176 Chr.15: 40767189-40767189 on Build GRCh38
rs1045955424 Chr.15: 40767302-40767302 on Build GRCh38
rs752258258 Chr.15: 40767383-40767383 on Build GRCh38
rs769713679 Chr.15: 40767236-40767236 on Build GRCh38
rs772366096 Chr.15: 40767323-40767323 on Build GRCh38
rs1020791200 Chr.15: 40767179-40767179 on Build GRCh38
rs772079236 Chr.15: 40767457-40767457 on Build GRCh38
rs376440278 Chr.15: 40767207-40767207 on Build GRCh38
rs770814286 Chr.15: 40767313-40767313 on Build GRCh38
rs137970311 Chr.15: 40767200-40767201 on Build GRCh38
rs374710519 Chr.15: 40767205-40767205 on Build GRCh38
rs984321458 Chr.15: 40767566-40767566 on Build GRCh38
rs757681808 Chr.15: 40767271-40767271 on Build GRCh38
rs749127710 Chr.15: 40767201-40767201 on Build GRCh38
rs754417572 Chr.15: 40767351-40767351 on Build GRCh38
rs745863947 Chr.15: 40767328-40767330 on Build GRCh38
rs759847227 Chr.15: 40767321-40767321 on Build GRCh38
rs527957641 Chr.15: 40767402-40767402 on Build GRCh38
rs747778468 Chr.15: 40767190-40767191 on Build GRCh38
rs970281700 Chr.15: 40767348-40767348 on Build GRCh38
rs927421933 Chr.15: 40767589-40767589 on Build GRCh38
rs977421446 Chr.15: 40767202-40767203 on Build GRCh38
rs774346021 Chr.15: 40767212-40767215 on Build GRCh38
rs960011221 Chr.15: 40767534-40767534 on Build GRCh38
rs571040859 Chr.15: 40767477-40767477 on Build GRCh38
rs754558236 Chr.15: 40767278-40767278 on Build GRCh38
rs149691221 Chr.15: 40767400-40767400 on Build GRCh38
rs950242373 Chr.15: 40767291-40767291 on Build GRCh38
rs774287216 Chr.15: 40767315-40767315 on Build GRCh38
rs150592330 Chr.15: 40767225-40767225 on Build GRCh38
rs370439455 Chr.15: 40767258-40767258 on Build GRCh38
rs745754620 Chr.15: 40767413-40767413 on Build GRCh38
rs918324426 Chr.15: 40767547-40767547 on Build GRCh38
rs775737051 Chr.15: 40767328-40767328 on Build GRCh38
rs73398513 Chr.15: 40767587-40767587 on Build GRCh38
rs770711732 Chr.15: 40767208-40767209 on Build GRCh38
rs766571674 Chr.15: 40767204-40767204 on Build GRCh38
rs764616321 Chr.15: 40767259-40767259 on Build GRCh38
rs558689699 Chr.15: 40767542-40767542 on Build GRCh38
rs200668576 Chr.15: 40767203-40767203 on Build GRCh38
rs922274748 Chr.15: 40767403-40767403 on Build GRCh38
rs953453240 Chr.15: 40767171-40767171 on Build GRCh38
rs568893913 Chr.15: 40767550-40767550 on Build GRCh38
rs1016812849 Chr.15: 40767479-40767479 on Build GRCh38
rs139646493 Chr.15: 40767346-40767346 on Build GRCh38
rs1022115463 Chr.15: 40767268-40767268 on Build GRCh38
rs754646233 Chr.15: 40767375-40767375 on Build GRCh38
rs755145154 Chr.15: 40767206-40767206 on Build GRCh38
rs759815106 Chr.15: 40767248-40767248 on Build GRCh38
rs201305860 Chr.15: 40767398-40767398 on Build GRCh38
rs776591301 Chr.15: 40767193-40767193 on Build GRCh38
rs949868004 Chr.15: 40767563-40767563 on Build GRCh38
rs779227677 Chr.15: 40767272-40767272 on Build GRCh38
rs202144307 Chr.15: 40767332-40767332 on Build GRCh38
rs867123043 Chr.15: 40767387-40767387 on Build GRCh38
rs200059682 Chr.15: 40767208-40767208 on Build GRCh38
rs1008133201 Chr.15: 40767463-40767463 on Build GRCh38
rs768204211 Chr.15: 40767216-40767216 on Build GRCh38
rs768771246 Chr.15: 40767351-40767353 on Build GRCh38
rs998812313 Chr.15: 40767284-40767284 on Build GRCh38
rs749155056 Chr.15: 40767307-40767307 on Build GRCh38
rs985303127 Chr.15: 40767465-40767465 on Build GRCh38
rs953895813 Chr.15: 40767459-40767459 on Build GRCh38
rs531608687 Chr.15: 40767393-40767393 on Build GRCh38
rs765589928 Chr.15: 40767372-40767372 on Build GRCh38
rs371155421 Chr.15: 40767214-40767214 on Build GRCh38
rs1014236671 Chr.15: 40767539-40767539 on Build GRCh38
rs904568715 Chr.15: 40767498-40767498 on Build GRCh38
rs562588825 Chr.15: 40767376-40767376 on Build GRCh38
rs544341876 Chr.15: 40767156-40767156 on Build GRCh38
rs1052000044 Chr.15: 40767447-40767447 on Build GRCh38
rs776744478 Chr.15: 40767400-40767400 on Build GRCh38
rs752762590 Chr.15: 40767254-40767254 on Build GRCh38
rs532506013 Chr.15: 40767270-40767270 on Build GRCh38
rs538786189 Chr.15: 40767510-40767510 on Build GRCh38
rs537459148 Chr.15: 40767580-40767580 on Build GRCh38
rs750728810 Chr.15: 40767373-40767373 on Build GRCh38
rs772903316 Chr.15: 40767237-40767237 on Build GRCh38

View Full Product Details