Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40770655-40771155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf62
Entrez Gene ID
643338
Gene Name
chromosome 15 open reading frame 62
Gene Aliases
-
UniGene
Hs.631715
Gene Chromosome Location
Chr.15: 40769961-40772449 on Build GRCh38
Transcripts
NM_001130448.2 NM_018163.2 XM_017022401.1

Gene Symbol
DNAJC17
Entrez Gene ID
55192
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Gene Aliases
-
UniGene
Hs.511069
Gene Chromosome Location
Chr.15: 40767448-40807478 on Build GRCh38
Transcripts
NM_001130448.2 NM_018163.2 XM_017022401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4408552 c.494T>C p.L165P Chr.15: 40770989-40770989 on Build GRCh38
5576461 c.360C>T p.L120L Chr.15: 40770855-40770855 on Build GRCh38
5599162 c.255C>T p.L85L Chr.15: 40770750-40770750 on Build GRCh38
7225277 c.229C>T p.L77L Chr.15: 40770724-40770724 on Build GRCh38
4982978 c.264G>A p.E88E Chr.15: 40770759-40770759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759509900 Chr.15: 40770792-40770792 on Build GRCh38
rs745895277 Chr.15: 40771113-40771113 on Build GRCh38
rs767218189 Chr.15: 40770803-40770803 on Build GRCh38
rs896124019 Chr.15: 40771114-40771114 on Build GRCh38
rs75415891 Chr.15: 40770719-40770719 on Build GRCh38
rs957009159 Chr.15: 40770923-40770923 on Build GRCh38
rs771505072 Chr.15: 40770703-40770703 on Build GRCh38
rs752284418 Chr.15: 40770934-40770935 on Build GRCh38
rs1004542929 Chr.15: 40770833-40770833 on Build GRCh38
rs995852064 Chr.15: 40770702-40770702 on Build GRCh38
rs866077258 Chr.15: 40770933-40770934 on Build GRCh38
rs1014564978 Chr.15: 40770834-40770834 on Build GRCh38
rs1033000862 Chr.15: 40770897-40770897 on Build GRCh38
rs772565723 Chr.15: 40771040-40771040 on Build GRCh38
rs1036298009 Chr.15: 40771112-40771112 on Build GRCh38
rs1028762448 Chr.15: 40770902-40770902 on Build GRCh38
rs779038030 Chr.15: 40770965-40770965 on Build GRCh38
rs200470384 Chr.15: 40770683-40770683 on Build GRCh38
rs777934673 Chr.15: 40770889-40770889 on Build GRCh38
rs749320471 Chr.15: 40770695-40770695 on Build GRCh38
rs751507060 Chr.15: 40770782-40770782 on Build GRCh38
rs1023401420 Chr.15: 40770850-40770850 on Build GRCh38
rs143048843 Chr.15: 40770809-40770809 on Build GRCh38
rs372111000 Chr.15: 40770771-40770771 on Build GRCh38
rs375299630 Chr.15: 40770975-40770975 on Build GRCh38
rs567542126 Chr.15: 40770718-40770718 on Build GRCh38
rs747454578 Chr.15: 40770755-40770756 on Build GRCh38
rs1050103881 Chr.15: 40771131-40771131 on Build GRCh38
rs766269846 Chr.15: 40771128-40771128 on Build GRCh38
rs757333431 Chr.15: 40770955-40770955 on Build GRCh38
rs887740654 Chr.15: 40770819-40770819 on Build GRCh38
rs781024377 Chr.15: 40770681-40770681 on Build GRCh38
rs748434048 Chr.15: 40770682-40770682 on Build GRCh38
rs559698327 Chr.15: 40771071-40771071 on Build GRCh38
rs966817956 Chr.15: 40770741-40770741 on Build GRCh38
rs762927380 Chr.15: 40771116-40771116 on Build GRCh38
rs979645873 Chr.15: 40770873-40770873 on Build GRCh38
rs1055120023 Chr.15: 40770754-40770754 on Build GRCh38
rs577702432 Chr.15: 40770835-40770835 on Build GRCh38
rs940549861 Chr.15: 40771110-40771110 on Build GRCh38
rs368691469 Chr.15: 40770755-40770755 on Build GRCh38
rs980062598 Chr.15: 40770691-40770691 on Build GRCh38
rs761562699 Chr.15: 40770745-40770745 on Build GRCh38
rs771398272 Chr.15: 40770830-40770830 on Build GRCh38
rs1866170 Chr.15: 40771046-40771046 on Build GRCh38
rs996055169 Chr.15: 40770776-40770776 on Build GRCh38
rs916679806 Chr.15: 40771047-40771047 on Build GRCh38
rs374302461 Chr.15: 40770845-40770845 on Build GRCh38
rs573561742 Chr.15: 40771027-40771027 on Build GRCh38
rs543144516 Chr.15: 40770843-40770843 on Build GRCh38
rs749795660 Chr.15: 40770800-40770800 on Build GRCh38
rs969946655 Chr.15: 40770857-40770857 on Build GRCh38
rs978276658 Chr.15: 40771005-40771005 on Build GRCh38
rs769514449 Chr.15: 40770753-40770753 on Build GRCh38
rs1040192289 Chr.15: 40771098-40771098 on Build GRCh38
rs111945653 Chr.15: 40770742-40770742 on Build GRCh38
rs754777607 Chr.15: 40770680-40770680 on Build GRCh38
rs372401206 Chr.15: 40771008-40771008 on Build GRCh38
rs746255696 Chr.15: 40770901-40770901 on Build GRCh38
rs779330480 Chr.15: 40770713-40770713 on Build GRCh38
rs939466563 Chr.15: 40770746-40770746 on Build GRCh38
rs368474634 Chr.15: 40770846-40770846 on Build GRCh38
rs931805171 Chr.15: 40771076-40771076 on Build GRCh38
rs772918999 Chr.15: 40771093-40771093 on Build GRCh38
rs528364220 Chr.15: 40771129-40771129 on Build GRCh38
rs988358573 Chr.15: 40770999-40770999 on Build GRCh38
rs1013284819 Chr.15: 40771115-40771115 on Build GRCh38
rs887614358 Chr.15: 40771096-40771096 on Build GRCh38
rs751482016 Chr.15: 40770679-40770679 on Build GRCh38
rs115347345 Chr.15: 40770724-40770724 on Build GRCh38
rs747335753 Chr.15: 40771069-40771069 on Build GRCh38

View Full Product Details