Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40774272-40774781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC17
Entrez Gene ID
55192
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Gene Aliases
-
UniGene
Hs.511069
Gene Chromosome Location
Chr.15: 40767448-40807478 on Build GRCh38
Transcripts
NM_018163.2 XM_017022401.1 XM_017022402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5924027 c.601-8C>T p.? Chr.15: 40774444-40774444 on Build GRCh38
280532 c.613C>A p.L205I Chr.15: 40774424-40774424 on Build GRCh38
5612547 c.615C>G p.L205L Chr.15: 40774422-40774422 on Build GRCh38
2269821 c.681G>A p.A227A Chr.15: 40774356-40774356 on Build GRCh38
6647279 c.669C>T p.T223T Chr.15: 40774368-40774368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763278115 Chr.15: 40774410-40774410 on Build GRCh38
rs928565250 Chr.15: 40774632-40774632 on Build GRCh38
rs568044849 Chr.15: 40774493-40774493 on Build GRCh38
rs1031030824 Chr.15: 40774526-40774526 on Build GRCh38
rs769697281 Chr.15: 40774394-40774394 on Build GRCh38
rs373577170 Chr.15: 40774332-40774332 on Build GRCh38
rs973934668 Chr.15: 40774344-40774344 on Build GRCh38
rs764528384 Chr.15: 40774472-40774472 on Build GRCh38
rs751628805 Chr.15: 40774324-40774324 on Build GRCh38
rs754145086 Chr.15: 40774474-40774474 on Build GRCh38
rs951363310 Chr.15: 40774605-40774605 on Build GRCh38
rs759727236 Chr.15: 40774448-40774448 on Build GRCh38
rs373037192 Chr.15: 40774321-40774321 on Build GRCh38
rs192475171 Chr.15: 40774584-40774584 on Build GRCh38
rs528262207 Chr.15: 40774732-40774732 on Build GRCh38
rs954887813 Chr.15: 40774300-40774300 on Build GRCh38
rs763263748 Chr.15: 40774320-40774320 on Build GRCh38
rs370650505 Chr.15: 40774338-40774338 on Build GRCh38
rs148868042 Chr.15: 40774368-40774368 on Build GRCh38
rs758068795 Chr.15: 40774483-40774483 on Build GRCh38
rs576423471 Chr.15: 40774621-40774621 on Build GRCh38
rs1026796061 Chr.15: 40774600-40774600 on Build GRCh38
rs1043168938 Chr.15: 40774498-40774498 on Build GRCh38
rs199700166 Chr.15: 40774463-40774463 on Build GRCh38
rs8023337 Chr.15: 40774585-40774585 on Build GRCh38
rs879380613 Chr.15: 40774653-40774653 on Build GRCh38
rs772815785 Chr.15: 40774402-40774402 on Build GRCh38
rs756553381 Chr.15: 40774339-40774339 on Build GRCh38
rs919764701 Chr.15: 40774349-40774349 on Build GRCh38
rs954716334 Chr.15: 40774532-40774532 on Build GRCh38
rs117485355 Chr.15: 40774427-40774427 on Build GRCh38
rs1008013623 Chr.15: 40774574-40774574 on Build GRCh38
rs764917108 Chr.15: 40774305-40774305 on Build GRCh38
rs903382509 Chr.15: 40774511-40774511 on Build GRCh38
rs537866068 Chr.15: 40774484-40774484 on Build GRCh38
rs1052020189 Chr.15: 40774733-40774733 on Build GRCh38
rs753307625 Chr.15: 40774456-40774456 on Build GRCh38
rs757980457 Chr.15: 40774365-40774365 on Build GRCh38
rs999034758 Chr.15: 40774513-40774513 on Build GRCh38
rs781237478 Chr.15: 40774385-40774385 on Build GRCh38
rs547758557 Chr.15: 40774488-40774488 on Build GRCh38
rs1024508501 Chr.15: 40774508-40774508 on Build GRCh38
rs982681365 Chr.15: 40774631-40774631 on Build GRCh38
rs541743713 Chr.15: 40774723-40774723 on Build GRCh38
rs1055529608 Chr.15: 40774451-40774451 on Build GRCh38
rs533582779 Chr.15: 40774755-40774755 on Build GRCh38
rs1046031412 Chr.15: 40774727-40774727 on Build GRCh38
rs556625852 Chr.15: 40774609-40774609 on Build GRCh38
rs938374697 Chr.15: 40774437-40774437 on Build GRCh38
rs1034447570 Chr.15: 40774551-40774551 on Build GRCh38
rs777990539 Chr.15: 40774346-40774346 on Build GRCh38
rs991529478 Chr.15: 40774669-40774669 on Build GRCh38
rs767475648 Chr.15: 40774453-40774453 on Build GRCh38
rs192719522 Chr.15: 40774690-40774690 on Build GRCh38
rs910652077 Chr.15: 40774340-40774340 on Build GRCh38
rs750145696 Chr.15: 40774356-40774356 on Build GRCh38
rs773539913 Chr.15: 40774313-40774313 on Build GRCh38
rs201311218 Chr.15: 40774325-40774325 on Build GRCh38
rs189449509 Chr.15: 40774689-40774689 on Build GRCh38
rs755408685 Chr.15: 40774673-40774673 on Build GRCh38
rs890737882 Chr.15: 40774743-40774743 on Build GRCh38
rs553589133 Chr.15: 40774554-40774554 on Build GRCh38
rs748888446 Chr.15: 40774724-40774724 on Build GRCh38
rs947116805 Chr.15: 40774710-40774710 on Build GRCh38
rs200233100 Chr.15: 40774389-40774389 on Build GRCh38
rs865899185 Chr.15: 40774408-40774408 on Build GRCh38
rs758944562 Chr.15: 40774499-40774499 on Build GRCh38
rs963910079 Chr.15: 40774343-40774343 on Build GRCh38
rs768613436 Chr.15: 40774382-40774382 on Build GRCh38
rs3784289 Chr.15: 40774677-40774677 on Build GRCh38
rs936332999 Chr.15: 40774693-40774693 on Build GRCh38
rs942551321 Chr.15: 40774611-40774611 on Build GRCh38
rs36041389 Chr.15: 40774519-40774519 on Build GRCh38
rs1017679584 Chr.15: 40774576-40774576 on Build GRCh38
rs1011290990 Chr.15: 40774490-40774490 on Build GRCh38
rs771225527 Chr.15: 40774411-40774411 on Build GRCh38
rs1047029408 Chr.15: 40774419-40774419 on Build GRCh38
rs868306657 Chr.15: 40774367-40774367 on Build GRCh38
rs548038695 Chr.15: 40774366-40774366 on Build GRCh38
rs377185052 Chr.15: 40774328-40774328 on Build GRCh38
rs929757273 Chr.15: 40774295-40774295 on Build GRCh38
rs1052142592 Chr.15: 40774708-40774708 on Build GRCh38
rs751924842 Chr.15: 40774606-40774606 on Build GRCh38

View Full Product Details