Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40872123-40872621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOV
Entrez Gene ID
171177
Gene Name
ras homolog family member V
Gene Aliases
ARHV, CHP, WRCH2
UniGene
Hs.447901
Gene Chromosome Location
Chr.15: 40872214-40874289 on Build GRCh38
Transcripts
NM_133639.3

SNP Detail

SNP ID Chromosome Location
rs917922339 Chr.15: 40872573-40872573 on Build GRCh38
rs534251992 Chr.15: 40872243-40872243 on Build GRCh38
rs2279578 Chr.15: 40872146-40872146 on Build GRCh38
rs918145270 Chr.15: 40872436-40872436 on Build GRCh38
rs957517282 Chr.15: 40872176-40872176 on Build GRCh38
rs941941009 Chr.15: 40872599-40872599 on Build GRCh38
rs941826632 Chr.15: 40872293-40872293 on Build GRCh38
rs529916738 Chr.15: 40872315-40872315 on Build GRCh38
rs975745374 Chr.15: 40872302-40872302 on Build GRCh38
rs1020609750 Chr.15: 40872276-40872276 on Build GRCh38
rs940952426 Chr.15: 40872393-40872393 on Build GRCh38
rs897609849 Chr.15: 40872308-40872308 on Build GRCh38
rs537573453 Chr.15: 40872444-40872444 on Build GRCh38
rs909359647 Chr.15: 40872358-40872358 on Build GRCh38
rs201156724 Chr.15: 40872517-40872518 on Build GRCh38
rs953074115 Chr.15: 40872334-40872334 on Build GRCh38
rs568852689 Chr.15: 40872579-40872579 on Build GRCh38
rs35579673 Chr.15: 40872480-40872480 on Build GRCh38
rs965766098 Chr.15: 40872301-40872301 on Build GRCh38
rs758006976 Chr.15: 40872515-40872515 on Build GRCh38
rs368628454 Chr.15: 40872210-40872210 on Build GRCh38
rs969380589 Chr.15: 40872589-40872589 on Build GRCh38
rs949451999 Chr.15: 40872445-40872445 on Build GRCh38
rs554249439 Chr.15: 40872592-40872592 on Build GRCh38
rs778115288 Chr.15: 40872541-40872541 on Build GRCh38
rs777553907 Chr.15: 40872526-40872526 on Build GRCh38
rs1036644114 Chr.15: 40872402-40872402 on Build GRCh38
rs537516437 Chr.15: 40872591-40872591 on Build GRCh38
rs984960898 Chr.15: 40872339-40872339 on Build GRCh38
rs755829415 Chr.15: 40872481-40872481 on Build GRCh38
rs186631248 Chr.15: 40872517-40872517 on Build GRCh38
rs1036148042 Chr.15: 40872300-40872300 on Build GRCh38
rs956799773 Chr.15: 40872549-40872549 on Build GRCh38
rs1010127390 Chr.15: 40872244-40872244 on Build GRCh38
rs538952741 Chr.15: 40872529-40872529 on Build GRCh38
rs558747627 Chr.15: 40872544-40872544 on Build GRCh38
rs546874942 Chr.15: 40872471-40872471 on Build GRCh38
rs1036178274 Chr.15: 40872600-40872600 on Build GRCh38
rs551871721 Chr.15: 40872555-40872555 on Build GRCh38
rs905291453 Chr.15: 40872466-40872466 on Build GRCh38
rs1045034672 Chr.15: 40872446-40872446 on Build GRCh38

View Full Product Details