Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40872928-40873430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOV
Entrez Gene ID
171177
Gene Name
ras homolog family member V
Gene Aliases
ARHV, CHP, WRCH2
UniGene
Hs.447901
Gene Chromosome Location
Chr.15: 40872214-40874289 on Build GRCh38
Transcripts
NM_133639.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054428 c.562G>A p.A188T Chr.15: 40873207-40873207 on Build GRCh38
961311 c.536G>A p.R179Q Chr.15: 40873233-40873233 on Build GRCh38
4054429 c.443G>T p.R148M Chr.15: 40873326-40873326 on Build GRCh38
7140755 c.604A>G p.I202V Chr.15: 40873165-40873165 on Build GRCh38
3936736 c.435C>T p.A145A Chr.15: 40873334-40873334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943098449 Chr.15: 40873051-40873051 on Build GRCh38
rs775398629 Chr.15: 40873355-40873356 on Build GRCh38
rs376857228 Chr.15: 40873136-40873136 on Build GRCh38
rs910357949 Chr.15: 40873376-40873376 on Build GRCh38
rs913869128 Chr.15: 40873322-40873322 on Build GRCh38
rs933116426 Chr.15: 40872972-40872972 on Build GRCh38
rs760798205 Chr.15: 40873106-40873106 on Build GRCh38
rs971957892 Chr.15: 40873296-40873296 on Build GRCh38
rs773181647 Chr.15: 40873042-40873042 on Build GRCh38
rs201047149 Chr.15: 40873008-40873008 on Build GRCh38
rs146446256 Chr.15: 40873057-40873057 on Build GRCh38
rs936715765 Chr.15: 40872955-40872955 on Build GRCh38
rs768690223 Chr.15: 40873403-40873403 on Build GRCh38
rs747688932 Chr.15: 40873034-40873034 on Build GRCh38
rs1050357314 Chr.15: 40873108-40873108 on Build GRCh38
rs776123479 Chr.15: 40873023-40873023 on Build GRCh38
rs989499063 Chr.15: 40873309-40873309 on Build GRCh38
rs751688269 Chr.15: 40873293-40873293 on Build GRCh38
rs752426836 Chr.15: 40873326-40873326 on Build GRCh38
rs780808040 Chr.15: 40873152-40873152 on Build GRCh38
rs111908877 Chr.15: 40873035-40873035 on Build GRCh38
rs777357106 Chr.15: 40873343-40873343 on Build GRCh38
rs923072276 Chr.15: 40873037-40873037 on Build GRCh38
rs757652620 Chr.15: 40873135-40873135 on Build GRCh38
rs750718965 Chr.15: 40873138-40873138 on Build GRCh38
rs553339259 Chr.15: 40873270-40873270 on Build GRCh38
rs936920351 Chr.15: 40873199-40873199 on Build GRCh38
rs867463553 Chr.15: 40872963-40872963 on Build GRCh38
rs184635660 Chr.15: 40872994-40872994 on Build GRCh38
rs771062342 Chr.15: 40873362-40873362 on Build GRCh38
rs954621412 Chr.15: 40873352-40873352 on Build GRCh38
rs1005924481 Chr.15: 40873137-40873137 on Build GRCh38
rs758738383 Chr.15: 40873291-40873291 on Build GRCh38
rs892639243 Chr.15: 40873214-40873214 on Build GRCh38
rs933126492 Chr.15: 40873040-40873040 on Build GRCh38
rs764547291 Chr.15: 40873112-40873112 on Build GRCh38
rs772021008 Chr.15: 40873377-40873377 on Build GRCh38
rs963315887 Chr.15: 40873294-40873294 on Build GRCh38
rs777109191 Chr.15: 40873177-40873177 on Build GRCh38
rs148901787 Chr.15: 40873289-40873289 on Build GRCh38
rs747658164 Chr.15: 40873165-40873165 on Build GRCh38
rs750843485 Chr.15: 40873290-40873290 on Build GRCh38
rs778983489 Chr.15: 40873367-40873367 on Build GRCh38
rs1042751845 Chr.15: 40873122-40873122 on Build GRCh38
rs780749750 Chr.15: 40873323-40873323 on Build GRCh38
rs532272641 Chr.15: 40873274-40873274 on Build GRCh38
rs755674136 Chr.15: 40873330-40873330 on Build GRCh38
rs774295602 Chr.15: 40873226-40873226 on Build GRCh38
rs762311867 Chr.15: 40873287-40873287 on Build GRCh38
rs897574577 Chr.15: 40873204-40873204 on Build GRCh38
rs774198794 Chr.15: 40873368-40873368 on Build GRCh38
rs754691420 Chr.15: 40873150-40873150 on Build GRCh38
rs759855477 Chr.15: 40873097-40873097 on Build GRCh38
rs140820332 Chr.15: 40873205-40873205 on Build GRCh38
rs966833198 Chr.15: 40873341-40873341 on Build GRCh38
rs748684036 Chr.15: 40873355-40873355 on Build GRCh38
rs753797823 Chr.15: 40873285-40873285 on Build GRCh38
rs747265102 Chr.15: 40873395-40873395 on Build GRCh38
rs769317376 Chr.15: 40873168-40873168 on Build GRCh38
rs774239954 Chr.15: 40873080-40873080 on Build GRCh38
rs775096024 Chr.15: 40873381-40873381 on Build GRCh38
rs752789848 Chr.15: 40873105-40873105 on Build GRCh38
rs764252736 Chr.15: 40873279-40873279 on Build GRCh38
rs771739683 Chr.15: 40873261-40873261 on Build GRCh38
rs149655270 Chr.15: 40873211-40873211 on Build GRCh38
rs868042128 Chr.15: 40873016-40873016 on Build GRCh38
rs767785165 Chr.15: 40873104-40873104 on Build GRCh38
rs922661224 Chr.15: 40873361-40873361 on Build GRCh38
rs145624117 Chr.15: 40873244-40873244 on Build GRCh38
rs989882526 Chr.15: 40872983-40872983 on Build GRCh38
rs754397050 Chr.15: 40873118-40873118 on Build GRCh38
rs1002326673 Chr.15: 40873277-40873277 on Build GRCh38
rs766309376 Chr.15: 40873059-40873059 on Build GRCh38
rs568713452 Chr.15: 40873354-40873354 on Build GRCh38
rs970895564 Chr.15: 40873255-40873255 on Build GRCh38
rs945414642 Chr.15: 40873033-40873033 on Build GRCh38

View Full Product Details