Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41191339-41191738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD1
Entrez Gene ID
161829
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
EXDL1
UniGene
Hs.307999
Gene Chromosome Location
Chr.15: 41182728-41231074 on Build GRCh38
Transcripts
NM_001286441.1 NM_152596.3 XM_006720415.3 XM_011521298.2 XM_011521299.2 XM_011521300.2 XM_011521301.2 XM_011521302.2 XM_017021970.1 XM_017021971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3501029 c.547-1G>A p.? Chr.15: 41191586-41191586 on Build GRCh38
2149064 c.578C>T p.T193M Chr.15: 41191554-41191554 on Build GRCh38
3815998 c.637C>G p.Q213E Chr.15: 41191495-41191495 on Build GRCh38
1163083 c.600C>A p.C200* Chr.15: 41191532-41191532 on Build GRCh38
1129217 c.648T>C p.P216P Chr.15: 41191484-41191484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774082866 Chr.15: 41191591-41191591 on Build GRCh38
rs926883345 Chr.15: 41191389-41191389 on Build GRCh38
rs773574004 Chr.15: 41191412-41191412 on Build GRCh38
rs760005212 Chr.15: 41191428-41191428 on Build GRCh38
rs776138815 Chr.15: 41191402-41191403 on Build GRCh38
rs926859613 Chr.15: 41191646-41191646 on Build GRCh38
rs1031629428 Chr.15: 41191713-41191713 on Build GRCh38
rs554765300 Chr.15: 41191576-41191576 on Build GRCh38
rs763729518 Chr.15: 41191403-41191403 on Build GRCh38
rs1046781318 Chr.15: 41191693-41191693 on Build GRCh38
rs375397761 Chr.15: 41191512-41191512 on Build GRCh38
rs199958393 Chr.15: 41191425-41191425 on Build GRCh38
rs769416031 Chr.15: 41191409-41191409 on Build GRCh38
rs887399512 Chr.15: 41191626-41191626 on Build GRCh38
rs780243280 Chr.15: 41191633-41191633 on Build GRCh38
rs183852185 Chr.15: 41191699-41191699 on Build GRCh38
rs770075401 Chr.15: 41191590-41191590 on Build GRCh38
rs747074437 Chr.15: 41191634-41191634 on Build GRCh38
rs761118111 Chr.15: 41191441-41191441 on Build GRCh38
rs752117599 Chr.15: 41191610-41191610 on Build GRCh38
rs754909536 Chr.15: 41191393-41191393 on Build GRCh38
rs112551797 Chr.15: 41191433-41191433 on Build GRCh38
rs1000114497 Chr.15: 41191710-41191710 on Build GRCh38
rs779204242 Chr.15: 41191499-41191499 on Build GRCh38
rs368109202 Chr.15: 41191614-41191614 on Build GRCh38
rs1971131 Chr.15: 41191484-41191484 on Build GRCh38
rs758019610 Chr.15: 41191465-41191465 on Build GRCh38
rs916897575 Chr.15: 41191684-41191691 on Build GRCh38
rs779212525 Chr.15: 41191572-41191572 on Build GRCh38
rs940248686 Chr.15: 41191639-41191639 on Build GRCh38
rs780605963 Chr.15: 41191654-41191654 on Build GRCh38
rs373773895 Chr.15: 41191417-41191417 on Build GRCh38
rs869300710 Chr.15: 41191435-41191436 on Build GRCh38
rs759088753 Chr.15: 41191543-41191543 on Build GRCh38
rs2412609 Chr.15: 41191438-41191438 on Build GRCh38
rs761035017 Chr.15: 41191518-41191518 on Build GRCh38
rs747959655 Chr.15: 41191403-41191403 on Build GRCh38
rs751075526 Chr.15: 41191470-41191470 on Build GRCh38
rs981005807 Chr.15: 41191387-41191387 on Build GRCh38
rs747404964 Chr.15: 41191686-41191686 on Build GRCh38
rs771248251 Chr.15: 41191493-41191493 on Build GRCh38
rs781200606 Chr.15: 41191482-41191482 on Build GRCh38
rs776072903 Chr.15: 41191516-41191516 on Build GRCh38
rs929674496 Chr.15: 41191692-41191692 on Build GRCh38
rs553335991 Chr.15: 41191698-41191698 on Build GRCh38
rs772212655 Chr.15: 41191577-41191577 on Build GRCh38
rs771158183 Chr.15: 41191423-41191423 on Build GRCh38
rs756099101 Chr.15: 41191551-41191551 on Build GRCh38
rs146356287 Chr.15: 41191585-41191585 on Build GRCh38
rs200607195 Chr.15: 41191438-41191439 on Build GRCh38
rs780915519 Chr.15: 41191398-41191398 on Build GRCh38
rs750703175 Chr.15: 41191625-41191625 on Build GRCh38
rs746442964 Chr.15: 41191515-41191515 on Build GRCh38
rs960954507 Chr.15: 41191651-41191651 on Build GRCh38
rs765911191 Chr.15: 41191468-41191468 on Build GRCh38
rs975547791 Chr.15: 41191581-41191581 on Build GRCh38
rs750249885 Chr.15: 41191621-41191622 on Build GRCh38
rs368618868 Chr.15: 41191531-41191531 on Build GRCh38
rs752508056 Chr.15: 41191483-41191483 on Build GRCh38
rs777751851 Chr.15: 41191553-41191553 on Build GRCh38
rs751899316 Chr.15: 41191714-41191714 on Build GRCh38
rs397702536 Chr.15: 41191434-41191435 on Build GRCh38
rs754928093 Chr.15: 41191481-41191481 on Build GRCh38
rs754992784 Chr.15: 41191635-41191635 on Build GRCh38
rs568236772 Chr.15: 41191580-41191580 on Build GRCh38
rs895956024 Chr.15: 41191681-41191681 on Build GRCh38
rs200349978 Chr.15: 41191554-41191554 on Build GRCh38
rs777664003 Chr.15: 41191486-41191486 on Build GRCh38
rs145939883 Chr.15: 41191514-41191514 on Build GRCh38
rs768068402 Chr.15: 41191437-41191437 on Build GRCh38
rs570889742 Chr.15: 41191487-41191487 on Build GRCh38
rs1013407800 Chr.15: 41191478-41191478 on Build GRCh38
rs7168916 Chr.15: 41191442-41191442 on Build GRCh38
rs758812571 Chr.15: 41191645-41191645 on Build GRCh38
rs946879244 Chr.15: 41191365-41191365 on Build GRCh38
rs767106867 Chr.15: 41191603-41191603 on Build GRCh38
rs749482910 Chr.15: 41191413-41191413 on Build GRCh38
rs769055082 Chr.15: 41191520-41191520 on Build GRCh38
rs375492102 Chr.15: 41191597-41191597 on Build GRCh38
rs149328050 Chr.15: 41191624-41191624 on Build GRCh38
rs750096333 Chr.15: 41191444-41191444 on Build GRCh38
rs1029625783 Chr.15: 41191557-41191557 on Build GRCh38
rs764986360 Chr.15: 41191590-41191590 on Build GRCh38
rs769143182 Chr.15: 41191583-41191583 on Build GRCh38
rs762178927 Chr.15: 41191588-41191588 on Build GRCh38
rs138628477 Chr.15: 41191525-41191525 on Build GRCh38
rs11371504 Chr.15: 41191433-41191434 on Build GRCh38
rs770447274 Chr.15: 41191455-41191456 on Build GRCh38
rs372591403 Chr.15: 41191545-41191545 on Build GRCh38
rs753775128 Chr.15: 41191615-41191615 on Build GRCh38
rs202157007 Chr.15: 41191542-41191542 on Build GRCh38
rs759727171 Chr.15: 41191546-41191546 on Build GRCh38
rs577472074 Chr.15: 41191561-41191561 on Build GRCh38
rs758515599 Chr.15: 41191629-41191629 on Build GRCh38
rs545492696 Chr.15: 41191708-41191708 on Build GRCh38
rs762656438 Chr.15: 41191529-41191529 on Build GRCh38
rs757124878 Chr.15: 41191617-41191617 on Build GRCh38
rs747587519 Chr.15: 41191582-41191582 on Build GRCh38

View Full Product Details