Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41919124-41919625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD4
Entrez Gene ID
30844
Gene Name
EH domain containing 4
Gene Aliases
PAST4
UniGene
Hs.143703
Gene Chromosome Location
Chr.15: 41899440-41972557 on Build GRCh38
Transcripts
NM_139265.3 XM_017022100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680472 c.763G>A p.V255I Chr.15: 41919371-41919371 on Build GRCh38
1372879 c.812G>T p.R271L Chr.15: 41919322-41919322 on Build GRCh38
3764525 c.814C>T p.R272W Chr.15: 41919320-41919320 on Build GRCh38
1372880 c.643G>A p.D215N Chr.15: 41919491-41919491 on Build GRCh38
5849165 c.551G>A p.R184K Chr.15: 41919583-41919583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1704396 Chr.15: 41919288-41919288 on Build GRCh38
rs376548348 Chr.15: 41919447-41919447 on Build GRCh38
rs554458708 Chr.15: 41919372-41919372 on Build GRCh38
rs752622839 Chr.15: 41919240-41919240 on Build GRCh38
rs550766410 Chr.15: 41919298-41919298 on Build GRCh38
rs760724939 Chr.15: 41919308-41919308 on Build GRCh38
rs766831165 Chr.15: 41919331-41919331 on Build GRCh38
rs779307032 Chr.15: 41919174-41919174 on Build GRCh38
rs202148169 Chr.15: 41919279-41919279 on Build GRCh38
rs1036075854 Chr.15: 41919196-41919196 on Build GRCh38
rs576091991 Chr.15: 41919482-41919482 on Build GRCh38
rs200388523 Chr.15: 41919233-41919233 on Build GRCh38
rs748265279 Chr.15: 41919350-41919350 on Build GRCh38
rs202010701 Chr.15: 41919207-41919207 on Build GRCh38
rs909781941 Chr.15: 41919302-41919302 on Build GRCh38
rs16972253 Chr.15: 41919465-41919465 on Build GRCh38
rs760109255 Chr.15: 41919245-41919245 on Build GRCh38
rs768382018 Chr.15: 41919480-41919480 on Build GRCh38
rs755252808 Chr.15: 41919434-41919434 on Build GRCh38
rs780434875 Chr.15: 41919267-41919267 on Build GRCh38
rs555167068 Chr.15: 41919348-41919348 on Build GRCh38
rs145101817 Chr.15: 41919588-41919588 on Build GRCh38
rs182386459 Chr.15: 41919222-41919222 on Build GRCh38
rs1014049100 Chr.15: 41919261-41919261 on Build GRCh38
rs772825170 Chr.15: 41919485-41919485 on Build GRCh38
rs759886582 Chr.15: 41919539-41919539 on Build GRCh38
rs373780104 Chr.15: 41919567-41919567 on Build GRCh38
rs781477004 Chr.15: 41919277-41919277 on Build GRCh38
rs762190346 Chr.15: 41919213-41919213 on Build GRCh38
rs577358797 Chr.15: 41919390-41919390 on Build GRCh38
rs778505288 Chr.15: 41919265-41919265 on Build GRCh38
rs746208043 Chr.15: 41919175-41919175 on Build GRCh38
rs200624712 Chr.15: 41919555-41919555 on Build GRCh38
rs17850065 Chr.15: 41919468-41919468 on Build GRCh38
rs200198158 Chr.15: 41919179-41919179 on Build GRCh38
rs756950488 Chr.15: 41919256-41919256 on Build GRCh38
rs753423229 Chr.15: 41919161-41919161 on Build GRCh38
rs764891494 Chr.15: 41919320-41919320 on Build GRCh38
rs764656471 Chr.15: 41919388-41919388 on Build GRCh38
rs369130040 Chr.15: 41919297-41919297 on Build GRCh38
rs985217597 Chr.15: 41919600-41919600 on Build GRCh38
rs368177481 Chr.15: 41919189-41919189 on Build GRCh38
rs371789052 Chr.15: 41919371-41919371 on Build GRCh38
rs764333028 Chr.15: 41919448-41919448 on Build GRCh38
rs771140821 Chr.15: 41919211-41919211 on Build GRCh38
rs774762467 Chr.15: 41919436-41919436 on Build GRCh38
rs768609604 Chr.15: 41919384-41919384 on Build GRCh38
rs369650545 Chr.15: 41919477-41919477 on Build GRCh38
rs569000123 Chr.15: 41919255-41919255 on Build GRCh38
rs761315340 Chr.15: 41919541-41919541 on Build GRCh38
rs149790926 Chr.15: 41919337-41919337 on Build GRCh38
rs867694581 Chr.15: 41919603-41919603 on Build GRCh38
rs772495218 Chr.15: 41919187-41919187 on Build GRCh38
rs763727588 Chr.15: 41919251-41919251 on Build GRCh38
rs201137614 Chr.15: 41919211-41919211 on Build GRCh38
rs775406627 Chr.15: 41919374-41919374 on Build GRCh38
rs750544880 Chr.15: 41919582-41919582 on Build GRCh38
rs779208576 Chr.15: 41919578-41919578 on Build GRCh38
rs753099915 Chr.15: 41919512-41919512 on Build GRCh38
rs751987742 Chr.15: 41919427-41919427 on Build GRCh38
rs558285342 Chr.15: 41919286-41919286 on Build GRCh38
rs759263318 Chr.15: 41919295-41919295 on Build GRCh38
rs768155622 Chr.15: 41919440-41919440 on Build GRCh38
rs776501059 Chr.15: 41919386-41919386 on Build GRCh38
rs761272807 Chr.15: 41919319-41919319 on Build GRCh38
rs376774893 Chr.15: 41919266-41919266 on Build GRCh38
rs761326048 Chr.15: 41919387-41919387 on Build GRCh38
rs975694777 Chr.15: 41919554-41919554 on Build GRCh38
rs778059179 Chr.15: 41919461-41919461 on Build GRCh38
rs1002148237 Chr.15: 41919250-41919250 on Build GRCh38
rs768061598 Chr.15: 41919507-41919507 on Build GRCh38
rs1009727368 Chr.15: 41919223-41919223 on Build GRCh38
rs561586372 Chr.15: 41919150-41919150 on Build GRCh38
rs769035690 Chr.15: 41919273-41919273 on Build GRCh38
rs150673276 Chr.15: 41919491-41919491 on Build GRCh38
rs145717011 Chr.15: 41919349-41919349 on Build GRCh38
rs267604199 Chr.15: 41919317-41919317 on Build GRCh38
rs370042939 Chr.15: 41919492-41919492 on Build GRCh38
rs940865551 Chr.15: 41919154-41919154 on Build GRCh38
rs921197506 Chr.15: 41919561-41919561 on Build GRCh38
rs762722037 Chr.15: 41919399-41919399 on Build GRCh38
rs756899518 Chr.15: 41919166-41919166 on Build GRCh38
rs1055383890 Chr.15: 41919398-41919398 on Build GRCh38
rs201628120 Chr.15: 41919312-41919312 on Build GRCh38
rs773821490 Chr.15: 41919502-41919502 on Build GRCh38
rs141371705 Chr.15: 41919481-41919481 on Build GRCh38
rs962553408 Chr.15: 41919552-41919552 on Build GRCh38
rs774330337 Chr.15: 41919225-41919225 on Build GRCh38
rs750016209 Chr.15: 41919322-41919322 on Build GRCh38
rs201972261 Chr.15: 41919376-41919376 on Build GRCh38
rs962646790 Chr.15: 41919315-41919315 on Build GRCh38
rs762500242 Chr.15: 41919562-41919562 on Build GRCh38
rs753981380 Chr.15: 41919558-41919558 on Build GRCh38
rs1019433317 Chr.15: 41919533-41919533 on Build GRCh38
rs757538412 Chr.15: 41919476-41919476 on Build GRCh38
rs945101313 Chr.15: 41919590-41919590 on Build GRCh38
rs143820519 Chr.15: 41919407-41919407 on Build GRCh38
rs746841619 Chr.15: 41919479-41919479 on Build GRCh38
rs148918551 Chr.15: 41919373-41919373 on Build GRCh38
rs776158221 Chr.15: 41919316-41919316 on Build GRCh38
rs767381628 Chr.15: 41919239-41919239 on Build GRCh38
rs756209468 Chr.15: 41919363-41919363 on Build GRCh38
rs759988164 Chr.15: 41919503-41919503 on Build GRCh38
rs758690965 Chr.15: 41919330-41919330 on Build GRCh38
rs747077546 Chr.15: 41919194-41919194 on Build GRCh38
rs758611545 Chr.15: 41919587-41919587 on Build GRCh38

View Full Product Details