Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41972024-41972522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD4
Entrez Gene ID
30844
Gene Name
EH domain containing 4
Gene Aliases
PAST4
UniGene
Hs.143703
Gene Chromosome Location
Chr.15: 41899440-41972557 on Build GRCh38
Transcripts
NM_139265.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5825154 c.203G>A p.G68D Chr.15: 41972292-41972292 on Build GRCh38
4680473 c.49G>A p.G17S Chr.15: 41972446-41972446 on Build GRCh38
5960556 c.169G>A p.D57N Chr.15: 41972326-41972326 on Build GRCh38
6372895 c.113C>T p.P38L Chr.15: 41972382-41972382 on Build GRCh38
6258537 c.184C>A p.P62T Chr.15: 41972311-41972311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766921131 Chr.15: 41972410-41972410 on Build GRCh38
rs755803967 Chr.15: 41972217-41972217 on Build GRCh38
rs1704413 Chr.15: 41972233-41972233 on Build GRCh38
rs1030387718 Chr.15: 41972104-41972104 on Build GRCh38
rs546332029 Chr.15: 41972320-41972320 on Build GRCh38
rs954884434 Chr.15: 41972113-41972113 on Build GRCh38
rs779295433 Chr.15: 41972230-41972230 on Build GRCh38
rs755530312 Chr.15: 41972420-41972420 on Build GRCh38
rs781568692 Chr.15: 41972348-41972348 on Build GRCh38
rs569187521 Chr.15: 41972446-41972446 on Build GRCh38
rs1010803265 Chr.15: 41972243-41972243 on Build GRCh38
rs767326565 Chr.15: 41972265-41972265 on Build GRCh38
rs375610735 Chr.15: 41972299-41972299 on Build GRCh38
rs759567413 Chr.15: 41972264-41972264 on Build GRCh38
rs750727862 Chr.15: 41972316-41972316 on Build GRCh38
rs1033563600 Chr.15: 41972075-41972075 on Build GRCh38
rs763701164 Chr.15: 41972225-41972225 on Build GRCh38
rs906247107 Chr.15: 41972200-41972200 on Build GRCh38
rs144645917 Chr.15: 41972342-41972342 on Build GRCh38
rs1003841692 Chr.15: 41972173-41972173 on Build GRCh38
rs370378699 Chr.15: 41972438-41972438 on Build GRCh38
rs756895395 Chr.15: 41972310-41972310 on Build GRCh38
rs1017018419 Chr.15: 41972179-41972179 on Build GRCh38
rs761185974 Chr.15: 41972488-41972488 on Build GRCh38
rs977929886 Chr.15: 41972096-41972096 on Build GRCh38
rs929465796 Chr.15: 41972192-41972192 on Build GRCh38
rs768556393 Chr.15: 41972458-41972458 on Build GRCh38
rs750197821 Chr.15: 41972236-41972252 on Build GRCh38
rs925356986 Chr.15: 41972091-41972091 on Build GRCh38
rs143664668 Chr.15: 41972370-41972370 on Build GRCh38
rs955573958 Chr.15: 41972212-41972212 on Build GRCh38
rs1020773954 Chr.15: 41972087-41972087 on Build GRCh38
rs919486523 Chr.15: 41972449-41972449 on Build GRCh38
rs893684791 Chr.15: 41972079-41972079 on Build GRCh38
rs776942412 Chr.15: 41972244-41972244 on Build GRCh38
rs925296980 Chr.15: 41972245-41972245 on Build GRCh38
rs763177768 Chr.15: 41972408-41972408 on Build GRCh38
rs747108421 Chr.15: 41972334-41972334 on Build GRCh38
rs577508454 Chr.15: 41972455-41972455 on Build GRCh38
rs942296282 Chr.15: 41972133-41972133 on Build GRCh38
rs149243465 Chr.15: 41972279-41972279 on Build GRCh38
rs1008362756 Chr.15: 41972499-41972499 on Build GRCh38
rs376352612 Chr.15: 41972435-41972435 on Build GRCh38
rs1010775511 Chr.15: 41972086-41972086 on Build GRCh38
rs770828488 Chr.15: 41972254-41972254 on Build GRCh38
rs530853023 Chr.15: 41972228-41972228 on Build GRCh38
rs762795018 Chr.15: 41972209-41972209 on Build GRCh38
rs745610526 Chr.15: 41972355-41972355 on Build GRCh38
rs759389891 Chr.15: 41972211-41972211 on Build GRCh38
rs774005655 Chr.15: 41972261-41972261 on Build GRCh38
rs1001796278 Chr.15: 41972074-41972074 on Build GRCh38
rs890879713 Chr.15: 41972482-41972482 on Build GRCh38
rs531948733 Chr.15: 41972354-41972354 on Build GRCh38
rs982261896 Chr.15: 41972476-41972476 on Build GRCh38
rs745692865 Chr.15: 41972444-41972444 on Build GRCh38
rs752045229 Chr.15: 41972414-41972414 on Build GRCh38
rs754227036 Chr.15: 41972226-41972226 on Build GRCh38
rs758980285 Chr.15: 41972319-41972319 on Build GRCh38
rs781472901 Chr.15: 41972427-41972427 on Build GRCh38
rs780227126 Chr.15: 41972238-41972238 on Build GRCh38
rs748441147 Chr.15: 41972351-41972351 on Build GRCh38
rs764761753 Chr.15: 41972400-41972400 on Build GRCh38
rs760185499 Chr.15: 41972366-41972366 on Build GRCh38
rs777813884 Chr.15: 41972441-41972441 on Build GRCh38
rs763752339 Chr.15: 41972297-41972297 on Build GRCh38
rs1017644098 Chr.15: 41972419-41972419 on Build GRCh38
rs990933743 Chr.15: 41972294-41972294 on Build GRCh38
rs779824169 Chr.15: 41972452-41972452 on Build GRCh38
rs902359778 Chr.15: 41972286-41972286 on Build GRCh38
rs747053552 Chr.15: 41972241-41972241 on Build GRCh38
rs949933282 Chr.15: 41972124-41972124 on Build GRCh38
rs769509081 Chr.15: 41972503-41972503 on Build GRCh38
rs776103121 Chr.15: 41972387-41972387 on Build GRCh38
rs546233298 Chr.15: 41972092-41972092 on Build GRCh38
rs866229721 Chr.15: 41972480-41972480 on Build GRCh38
rs775353018 Chr.15: 41972365-41972365 on Build GRCh38
rs751453834 Chr.15: 41972210-41972210 on Build GRCh38
rs772073305 Chr.15: 41972357-41972357 on Build GRCh38
rs565161677 Chr.15: 41972193-41972193 on Build GRCh38
rs867733492 Chr.15: 41972478-41972478 on Build GRCh38
rs769886976 Chr.15: 41972353-41972353 on Build GRCh38
rs910555901 Chr.15: 41972132-41972132 on Build GRCh38
rs1053227358 Chr.15: 41972186-41972186 on Build GRCh38
rs886770437 Chr.15: 41972135-41972135 on Build GRCh38
rs752127600 Chr.15: 41972216-41972216 on Build GRCh38
rs780086905 Chr.15: 41972435-41972451 on Build GRCh38
rs776154364 Chr.15: 41972475-41972475 on Build GRCh38
rs761571939 Chr.15: 41972392-41972392 on Build GRCh38
rs919441324 Chr.15: 41972190-41972190 on Build GRCh38
rs986092479 Chr.15: 41972125-41972125 on Build GRCh38
rs765689035 Chr.15: 41972315-41972315 on Build GRCh38
rs758813808 Chr.15: 41972234-41972234 on Build GRCh38
rs749108456 Chr.15: 41972249-41972249 on Build GRCh38
rs760134279 Chr.15: 41972293-41972293 on Build GRCh38
rs986208843 Chr.15: 41972399-41972399 on Build GRCh38
rs529909896 Chr.15: 41972307-41972307 on Build GRCh38
rs758173638 Chr.15: 41972434-41972462 on Build GRCh38
rs996538069 Chr.15: 41972469-41972469 on Build GRCh38
rs867363499 Chr.15: 41972277-41972277 on Build GRCh38
rs973635772 Chr.15: 41972149-41972149 on Build GRCh38
rs761401443 Chr.15: 41972046-41972046 on Build GRCh38

View Full Product Details