Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41972387-41972891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD4
Entrez Gene ID
30844
Gene Name
EH domain containing 4
Gene Aliases
PAST4
UniGene
Hs.143703
Gene Chromosome Location
Chr.15: 41899440-41972557 on Build GRCh38
Transcripts
NM_139265.3 NR_120334.1

Gene Symbol
PLA2G4E-AS1
Entrez Gene ID
101928388
Gene Name
PLA2G4E antisense RNA 1
Gene Aliases
-
UniGene
Hs.738043
Gene Chromosome Location
Chr.15: 41972763-41999094 on Build GRCh38
Transcripts
NM_139265.3 NR_120334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623101 c.107T>C p.V36A Chr.15: 41972388-41972388 on Build GRCh38
4680473 c.49G>A p.G17S Chr.15: 41972446-41972446 on Build GRCh38
6397273 c.22C>A p.Q8K Chr.15: 41972473-41972473 on Build GRCh38
4606899 c.83T>C p.L28P Chr.15: 41972412-41972412 on Build GRCh38
4455149 c.67A>G p.T23A Chr.15: 41972428-41972428 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112854755 Chr.15: 41972754-41972754 on Build GRCh38
rs529824049 Chr.15: 41972800-41972800 on Build GRCh38
rs766921131 Chr.15: 41972410-41972410 on Build GRCh38
rs1038720385 Chr.15: 41972609-41972609 on Build GRCh38
rs1021554660 Chr.15: 41972508-41972508 on Build GRCh38
rs559802461 Chr.15: 41972846-41972846 on Build GRCh38
rs752846838 Chr.15: 41972538-41972538 on Build GRCh38
rs890879713 Chr.15: 41972482-41972482 on Build GRCh38
rs868401099 Chr.15: 41972662-41972662 on Build GRCh38
rs939933890 Chr.15: 41972598-41972598 on Build GRCh38
rs937796482 Chr.15: 41972534-41972534 on Build GRCh38
rs982261896 Chr.15: 41972476-41972476 on Build GRCh38
rs755530312 Chr.15: 41972420-41972420 on Build GRCh38
rs745692865 Chr.15: 41972444-41972444 on Build GRCh38
rs569187521 Chr.15: 41972446-41972446 on Build GRCh38
rs752045229 Chr.15: 41972414-41972414 on Build GRCh38
rs553939692 Chr.15: 41972692-41972692 on Build GRCh38
rs181118001 Chr.15: 41972594-41972594 on Build GRCh38
rs538197690 Chr.15: 41972545-41972545 on Build GRCh38
rs751512778 Chr.15: 41972807-41972807 on Build GRCh38
rs781472901 Chr.15: 41972427-41972427 on Build GRCh38
rs569047985 Chr.15: 41972810-41972810 on Build GRCh38
rs751951523 Chr.15: 41972520-41972520 on Build GRCh38
rs889612820 Chr.15: 41972574-41972574 on Build GRCh38
rs575600160 Chr.15: 41972783-41972783 on Build GRCh38
rs768098366 Chr.15: 41972531-41972531 on Build GRCh38
rs777813884 Chr.15: 41972441-41972441 on Build GRCh38
rs1017644098 Chr.15: 41972419-41972419 on Build GRCh38
rs187166173 Chr.15: 41972775-41972775 on Build GRCh38
rs772682943 Chr.15: 41972507-41972507 on Build GRCh38
rs779824169 Chr.15: 41972452-41972452 on Build GRCh38
rs973881284 Chr.15: 41972862-41972862 on Build GRCh38
rs928164738 Chr.15: 41972699-41972699 on Build GRCh38
rs769509081 Chr.15: 41972503-41972503 on Build GRCh38
rs760162321 Chr.15: 41972521-41972521 on Build GRCh38
rs761908471 Chr.15: 41972553-41972553 on Build GRCh38
rs370378699 Chr.15: 41972438-41972438 on Build GRCh38
rs961538571 Chr.15: 41972852-41972852 on Build GRCh38
rs749975273 Chr.15: 41972725-41972725 on Build GRCh38
rs74533604 Chr.15: 41972835-41972835 on Build GRCh38
rs980118180 Chr.15: 41972514-41972514 on Build GRCh38
rs866229721 Chr.15: 41972480-41972480 on Build GRCh38
rs577446313 Chr.15: 41972701-41972701 on Build GRCh38
rs933955568 Chr.15: 41972770-41972770 on Build GRCh38
rs575168974 Chr.15: 41972568-41972568 on Build GRCh38
rs761185974 Chr.15: 41972488-41972488 on Build GRCh38
rs766282598 Chr.15: 41972758-41972758 on Build GRCh38
rs768556393 Chr.15: 41972458-41972458 on Build GRCh38
rs867733492 Chr.15: 41972478-41972478 on Build GRCh38
rs1007669250 Chr.15: 41972576-41972576 on Build GRCh38
rs374725303 Chr.15: 41972510-41972510 on Build GRCh38
rs545877668 Chr.15: 41972583-41972583 on Build GRCh38
rs766916424 Chr.15: 41972515-41972515 on Build GRCh38
rs946389910 Chr.15: 41972743-41972743 on Build GRCh38
rs534423659 Chr.15: 41972657-41972657 on Build GRCh38
rs994812530 Chr.15: 41972658-41972658 on Build GRCh38
rs919486523 Chr.15: 41972449-41972449 on Build GRCh38
rs528253250 Chr.15: 41972779-41972779 on Build GRCh38
rs780086905 Chr.15: 41972435-41972451 on Build GRCh38
rs959612936 Chr.15: 41972756-41972756 on Build GRCh38
rs776154364 Chr.15: 41972475-41972475 on Build GRCh38
rs970806729 Chr.15: 41972567-41972567 on Build GRCh38
rs1042706647 Chr.15: 41972558-41972558 on Build GRCh38
rs1026984342 Chr.15: 41972663-41972663 on Build GRCh38
rs914958899 Chr.15: 41972731-41972731 on Build GRCh38
rs959395582 Chr.15: 41972557-41972557 on Build GRCh38
rs1017676542 Chr.15: 41972651-41972651 on Build GRCh38
rs1001118374 Chr.15: 41972737-41972737 on Build GRCh38
rs763177768 Chr.15: 41972408-41972408 on Build GRCh38
rs982763855 Chr.15: 41972686-41972686 on Build GRCh38
rs577508454 Chr.15: 41972455-41972455 on Build GRCh38
rs998509519 Chr.15: 41972565-41972565 on Build GRCh38
rs1039143862 Chr.15: 41972816-41972816 on Build GRCh38
rs1008362756 Chr.15: 41972499-41972499 on Build GRCh38
rs990543864 Chr.15: 41972712-41972712 on Build GRCh38
rs376352612 Chr.15: 41972435-41972435 on Build GRCh38
rs983964971 Chr.15: 41972798-41972798 on Build GRCh38
rs919592235 Chr.15: 41972634-41972634 on Build GRCh38
rs899208503 Chr.15: 41972821-41972821 on Build GRCh38
rs74803080 Chr.15: 41972787-41972787 on Build GRCh38
rs908388807 Chr.15: 41972572-41972572 on Build GRCh38
rs555138007 Chr.15: 41972564-41972564 on Build GRCh38
rs568511899 Chr.15: 41972632-41972632 on Build GRCh38
rs753777822 Chr.15: 41972675-41972675 on Build GRCh38
rs951035096 Chr.15: 41972664-41972664 on Build GRCh38
rs758173638 Chr.15: 41972434-41972462 on Build GRCh38
rs995327685 Chr.15: 41972839-41972839 on Build GRCh38
rs996538069 Chr.15: 41972469-41972469 on Build GRCh38
rs112553819 Chr.15: 41972808-41972808 on Build GRCh38
rs890847141 Chr.15: 41972689-41972689 on Build GRCh38

View Full Product Details