Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22074012-22074524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42
Entrez Gene ID
998
Gene Name
cell division cycle 42
Gene Aliases
CDC42Hs, G25K, TKS
UniGene
Hs.467637
Gene Chromosome Location
Chr.1: 22052627-22092943 on Build GRCh38
Transcripts
NM_001039802.1 NM_001791.3 NM_044472.2

SNP Detail

SNP ID Chromosome Location
rs11582456 Chr.1: 22074499-22074499 on Build GRCh38
rs960052059 Chr.1: 22074368-22074368 on Build GRCh38
rs750375794 Chr.1: 22074437-22074437 on Build GRCh38
rs529022968 Chr.1: 22074223-22074223 on Build GRCh38
rs1038831065 Chr.1: 22074430-22074430 on Build GRCh38
rs182370731 Chr.1: 22074366-22074366 on Build GRCh38
rs1049211609 Chr.1: 22074385-22074386 on Build GRCh38
rs916143447 Chr.1: 22074156-22074156 on Build GRCh38
rs145168930 Chr.1: 22074202-22074202 on Build GRCh38
rs10917138 Chr.1: 22074474-22074474 on Build GRCh38
rs536688494 Chr.1: 22074469-22074470 on Build GRCh38
rs968841594 Chr.1: 22074341-22074341 on Build GRCh38
rs990557867 Chr.1: 22074227-22074227 on Build GRCh38
rs889673673 Chr.1: 22074304-22074304 on Build GRCh38
rs1027405079 Chr.1: 22074171-22074171 on Build GRCh38
rs1036775242 Chr.1: 22074239-22074239 on Build GRCh38
rs747942405 Chr.1: 22074096-22074096 on Build GRCh38
rs997834077 Chr.1: 22074277-22074277 on Build GRCh38
rs887527563 Chr.1: 22074173-22074173 on Build GRCh38
rs1017800848 Chr.1: 22074180-22074180 on Build GRCh38
rs541567626 Chr.1: 22074374-22074374 on Build GRCh38
rs142167150 Chr.1: 22074470-22074470 on Build GRCh38
rs938674564 Chr.1: 22074136-22074136 on Build GRCh38
rs966224060 Chr.1: 22074187-22074187 on Build GRCh38
rs16826422 Chr.1: 22074338-22074338 on Build GRCh38
rs1021726244 Chr.1: 22074071-22074071 on Build GRCh38
rs775279134 Chr.1: 22074133-22074133 on Build GRCh38
rs977454694 Chr.1: 22074122-22074122 on Build GRCh38
rs570741605 Chr.1: 22074435-22074435 on Build GRCh38
rs572470358 Chr.1: 22074373-22074373 on Build GRCh38
rs6690757 Chr.1: 22074143-22074143 on Build GRCh38
rs769229674 Chr.1: 22074095-22074095 on Build GRCh38
rs922334949 Chr.1: 22074498-22074498 on Build GRCh38
rs924583093 Chr.1: 22074134-22074134 on Build GRCh38
rs764026091 Chr.1: 22074493-22074494 on Build GRCh38
rs939218881 Chr.1: 22074317-22074317 on Build GRCh38
rs989706986 Chr.1: 22074048-22074048 on Build GRCh38
rs997722767 Chr.1: 22074448-22074448 on Build GRCh38
rs563915290 Chr.1: 22074153-22074153 on Build GRCh38
rs977941392 Chr.1: 22074356-22074356 on Build GRCh38
rs946530968 Chr.1: 22074079-22074079 on Build GRCh38
rs1010760076 Chr.1: 22074327-22074327 on Build GRCh38
rs897478505 Chr.1: 22074163-22074164 on Build GRCh38
rs547300874 Chr.1: 22074240-22074240 on Build GRCh38
rs940201332 Chr.1: 22074484-22074484 on Build GRCh38
rs16826417 Chr.1: 22074179-22074179 on Build GRCh38
rs10799732 Chr.1: 22074278-22074278 on Build GRCh38
rs887507076 Chr.1: 22074407-22074407 on Build GRCh38
rs759626692 Chr.1: 22074457-22074457 on Build GRCh38
rs527284865 Chr.1: 22074041-22074041 on Build GRCh38
rs919207668 Chr.1: 22074345-22074345 on Build GRCh38
rs914678413 Chr.1: 22074057-22074057 on Build GRCh38
rs997753850 Chr.1: 22074217-22074217 on Build GRCh38
rs531467374 Chr.1: 22074167-22074167 on Build GRCh38
rs564411104 Chr.1: 22074425-22074425 on Build GRCh38
rs888221086 Chr.1: 22074219-22074219 on Build GRCh38
rs189094415 Chr.1: 22074496-22074496 on Build GRCh38

View Full Product Details