Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22081631-22082126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42
Entrez Gene ID
998
Gene Name
cell division cycle 42
Gene Aliases
CDC42Hs, G25K, TKS
UniGene
Hs.467637
Gene Chromosome Location
Chr.1: 22052627-22092943 on Build GRCh38
Transcripts
NM_001039802.1 NM_001791.3 NM_044472.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1645656 c.298A>G p.T100A Chr.1: 22081788-22081788 on Build GRCh38
4527075 c.269G>A p.G90E Chr.1: 22081759-22081759 on Build GRCh38
6624271 c.247G>A p.A83T Chr.1: 22081737-22081737 on Build GRCh38
6624272 c.121G>A p.A41T Chr.1: 22081737-22081737 on Build GRCh38
464004 c.172A>G p.T58A Chr.1: 22081788-22081788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769175616 Chr.1: 22081822-22081822 on Build GRCh38
rs367815612 Chr.1: 22081702-22081702 on Build GRCh38
rs753694054 Chr.1: 22082057-22082057 on Build GRCh38
rs1000490730 Chr.1: 22082066-22082066 on Build GRCh38
rs979453189 Chr.1: 22082056-22082056 on Build GRCh38
rs779470928 Chr.1: 22081811-22081811 on Build GRCh38
rs927299363 Chr.1: 22081665-22081665 on Build GRCh38
rs941360407 Chr.1: 22081950-22081950 on Build GRCh38
rs763869197 Chr.1: 22081677-22081677 on Build GRCh38
rs1057181069 Chr.1: 22082075-22082075 on Build GRCh38
rs189291053 Chr.1: 22081989-22081989 on Build GRCh38
rs761281005 Chr.1: 22081881-22081881 on Build GRCh38
rs774711630 Chr.1: 22081787-22081787 on Build GRCh38
rs927333512 Chr.1: 22081937-22081937 on Build GRCh38
rs553106950 Chr.1: 22081910-22081910 on Build GRCh38
rs1006163764 Chr.1: 22081906-22081906 on Build GRCh38
rs564097500 Chr.1: 22082102-22082102 on Build GRCh38
rs17837976 Chr.1: 22081719-22081719 on Build GRCh38
rs372237865 Chr.1: 22082016-22082016 on Build GRCh38
rs759963199 Chr.1: 22081802-22081802 on Build GRCh38
rs754911082 Chr.1: 22082077-22082077 on Build GRCh38
rs780801100 Chr.1: 22081846-22081846 on Build GRCh38
rs956795304 Chr.1: 22081854-22081854 on Build GRCh38
rs2056976 Chr.1: 22081972-22081972 on Build GRCh38
rs771636441 Chr.1: 22081748-22081748 on Build GRCh38
rs753546831 Chr.1: 22081817-22081817 on Build GRCh38
rs17837978 Chr.1: 22081893-22081893 on Build GRCh38
rs1057518022 Chr.1: 22081740-22081740 on Build GRCh38
rs930161362 Chr.1: 22081784-22081784 on Build GRCh38
rs1051350409 Chr.1: 22082020-22082020 on Build GRCh38
rs141108559 Chr.1: 22081745-22081745 on Build GRCh38
rs745356882 Chr.1: 22081694-22081694 on Build GRCh38
rs572007861 Chr.1: 22081993-22081993 on Build GRCh38
rs760388457 Chr.1: 22082028-22082028 on Build GRCh38
rs753362067 Chr.1: 22081680-22081680 on Build GRCh38
rs17837977 Chr.1: 22081840-22081840 on Build GRCh38
rs573373328 Chr.1: 22081938-22081938 on Build GRCh38
rs1052563821 Chr.1: 22082074-22082074 on Build GRCh38
rs2056975 Chr.1: 22082034-22082034 on Build GRCh38
rs201633260 Chr.1: 22081730-22081730 on Build GRCh38
rs574782905 Chr.1: 22081855-22081855 on Build GRCh38
rs974440269 Chr.1: 22081966-22081966 on Build GRCh38
rs538327935 Chr.1: 22081887-22081887 on Build GRCh38
rs1051667781 Chr.1: 22081803-22081803 on Build GRCh38
rs374060866 Chr.1: 22081683-22081683 on Build GRCh38
rs886372726 Chr.1: 22081897-22081897 on Build GRCh38
rs542813349 Chr.1: 22081858-22081858 on Build GRCh38
rs41300114 Chr.1: 22081699-22081699 on Build GRCh38
rs372440772 Chr.1: 22081828-22081828 on Build GRCh38
rs774888813 Chr.1: 22081835-22081835 on Build GRCh38
rs748279391 Chr.1: 22081720-22081720 on Build GRCh38
rs988952555 Chr.1: 22081962-22081962 on Build GRCh38
rs754833753 Chr.1: 22081841-22081841 on Build GRCh38
rs895914625 Chr.1: 22082098-22082098 on Build GRCh38
rs749999939 Chr.1: 22081831-22081831 on Build GRCh38
rs779979617 Chr.1: 22081836-22081836 on Build GRCh38
rs1030938847 Chr.1: 22081848-22081848 on Build GRCh38
rs368453240 Chr.1: 22081815-22081815 on Build GRCh38
rs892126370 Chr.1: 22082092-22082092 on Build GRCh38
rs1039443242 Chr.1: 22082038-22082038 on Build GRCh38
rs918852532 Chr.1: 22081971-22081971 on Build GRCh38
rs778342210 Chr.1: 22081688-22081688 on Build GRCh38
rs370073568 Chr.1: 22081707-22081707 on Build GRCh38
rs1030496948 Chr.1: 22081948-22081948 on Build GRCh38
rs201214894 Chr.1: 22081706-22081707 on Build GRCh38
rs758370940 Chr.1: 22081698-22081698 on Build GRCh38
rs754183369 Chr.1: 22081696-22081697 on Build GRCh38
rs773477556 Chr.1: 22081736-22081736 on Build GRCh38
rs764401254 Chr.1: 22081671-22081672 on Build GRCh38

View Full Product Details