Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23371418-23371964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF436-AS1
Entrez Gene ID
148898
Gene Name
ZNF436 antisense RNA 1
Gene Aliases
C1orf213
UniGene
Hs.61884
Gene Chromosome Location
Chr.1: 23368971-23371839 on Build GRCh38
Transcripts
NR_033690.1 NR_033691.1

SNP Detail

SNP ID Chromosome Location
rs756006291 Chr.1: 23371564-23371564 on Build GRCh38
rs931967112 Chr.1: 23371750-23371750 on Build GRCh38
rs754984 Chr.1: 23371627-23371627 on Build GRCh38
rs181265378 Chr.1: 23371584-23371584 on Build GRCh38
rs1034513711 Chr.1: 23371938-23371938 on Build GRCh38
rs377732804 Chr.1: 23371522-23371522 on Build GRCh38
rs779823235 Chr.1: 23371789-23371789 on Build GRCh38
rs984082902 Chr.1: 23371503-23371503 on Build GRCh38
rs116911595 Chr.1: 23371505-23371505 on Build GRCh38
rs751503234 Chr.1: 23371445-23371445 on Build GRCh38
rs373405708 Chr.1: 23371632-23371632 on Build GRCh38
rs756691877 Chr.1: 23371454-23371455 on Build GRCh38
rs921995282 Chr.1: 23371748-23371748 on Build GRCh38
rs922676199 Chr.1: 23371917-23371917 on Build GRCh38
rs1025571064 Chr.1: 23371787-23371787 on Build GRCh38
rs535701935 Chr.1: 23371820-23371820 on Build GRCh38
rs907969919 Chr.1: 23371688-23371688 on Build GRCh38
rs977233455 Chr.1: 23371853-23371853 on Build GRCh38
rs552590253 Chr.1: 23371531-23371531 on Build GRCh38
rs988301550 Chr.1: 23371930-23371930 on Build GRCh38
rs1053126055 Chr.1: 23371611-23371611 on Build GRCh38
rs895632705 Chr.1: 23371720-23371720 on Build GRCh38
rs767505456 Chr.1: 23371539-23371539 on Build GRCh38
rs993981854 Chr.1: 23371746-23371746 on Build GRCh38
rs772879107 Chr.1: 23371513-23371513 on Build GRCh38
rs374669495 Chr.1: 23371520-23371520 on Build GRCh38
rs772627172 Chr.1: 23371502-23371502 on Build GRCh38
rs1018269501 Chr.1: 23371844-23371844 on Build GRCh38
rs752530185 Chr.1: 23371544-23371544 on Build GRCh38
rs935498584 Chr.1: 23371926-23371926 on Build GRCh38
rs371215069 Chr.1: 23371529-23371529 on Build GRCh38
rs1035569997 Chr.1: 23371686-23371686 on Build GRCh38
rs964200205 Chr.1: 23371845-23371845 on Build GRCh38
rs549967994 Chr.1: 23371729-23371729 on Build GRCh38
rs764390527 Chr.1: 23371508-23371508 on Build GRCh38
rs951224395 Chr.1: 23371673-23371673 on Build GRCh38
rs188450257 Chr.1: 23371482-23371482 on Build GRCh38
rs747980087 Chr.1: 23371465-23371465 on Build GRCh38
rs1010379790 Chr.1: 23371878-23371878 on Build GRCh38
rs560499467 Chr.1: 23371506-23371506 on Build GRCh38
rs755916068 Chr.1: 23371481-23371481 on Build GRCh38
rs370366451 Chr.1: 23371494-23371494 on Build GRCh38
rs557930989 Chr.1: 23371635-23371635 on Build GRCh38
rs754934848 Chr.1: 23371447-23371447 on Build GRCh38
rs568284272 Chr.1: 23371779-23371779 on Build GRCh38
rs905652652 Chr.1: 23371642-23371642 on Build GRCh38
rs746419450 Chr.1: 23371500-23371500 on Build GRCh38
rs1001675912 Chr.1: 23371658-23371658 on Build GRCh38
rs114152668 Chr.1: 23371492-23371492 on Build GRCh38
rs894024098 Chr.1: 23371618-23371618 on Build GRCh38
rs1041840806 Chr.1: 23371912-23371912 on Build GRCh38
rs780919518 Chr.1: 23371456-23371456 on Build GRCh38

View Full Product Details