Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24358466-24358974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
UniGene
Hs.657920
Gene Chromosome Location
Chr.1: 24319322-24364482 on Build GRCh38
Transcripts
NM_198174.2 NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Gene Symbol
STPG1
Entrez Gene ID
90529
Gene Name
sperm tail PG-rich repeat containing 1
Gene Aliases
C1orf201, MAPO2
UniGene
Hs.403187
Gene Chromosome Location
Chr.1: 24356999-24415533 on Build GRCh38
Transcripts
NM_198174.2 NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5429832 c.859C>G p.L287V Chr.1: 24358548-24358548 on Build GRCh38
906867 c.843G>T p.K281N Chr.1: 24358564-24358564 on Build GRCh38
4599973 c.971A>T p.Y324F Chr.1: 24358577-24358577 on Build GRCh38
3486507 c.962C>T p.S321F Chr.1: 24358586-24358586 on Build GRCh38
1584338 c.984G>T p.K328N Chr.1: 24358564-24358564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377750621 Chr.1: 24358510-24358510 on Build GRCh38
rs761261576 Chr.1: 24358638-24358638 on Build GRCh38
rs774237692 Chr.1: 24358534-24358534 on Build GRCh38
rs749799146 Chr.1: 24358755-24358755 on Build GRCh38
rs371334407 Chr.1: 24358532-24358532 on Build GRCh38
rs765488575 Chr.1: 24358555-24358555 on Build GRCh38
rs779394383 Chr.1: 24358490-24358490 on Build GRCh38
rs1019577093 Chr.1: 24358570-24358570 on Build GRCh38
rs748457981 Chr.1: 24358526-24358526 on Build GRCh38
rs753421357 Chr.1: 24358553-24358553 on Build GRCh38
rs372838863 Chr.1: 24358639-24358639 on Build GRCh38
rs761576155 Chr.1: 24358554-24358554 on Build GRCh38
rs763240698 Chr.1: 24358642-24358642 on Build GRCh38
rs768740128 Chr.1: 24358514-24358514 on Build GRCh38
rs548676646 Chr.1: 24358816-24358816 on Build GRCh38
rs114811766 Chr.1: 24358752-24358752 on Build GRCh38
rs750873234 Chr.1: 24358500-24358500 on Build GRCh38
rs919744204 Chr.1: 24358725-24358725 on Build GRCh38
rs1015542060 Chr.1: 24358925-24358925 on Build GRCh38
rs780484873 Chr.1: 24358572-24358572 on Build GRCh38
rs976008075 Chr.1: 24358592-24358592 on Build GRCh38
rs559119091 Chr.1: 24358693-24358693 on Build GRCh38
rs781207949 Chr.1: 24358521-24358521 on Build GRCh38
rs943702905 Chr.1: 24358769-24358769 on Build GRCh38
rs745599379 Chr.1: 24358612-24358612 on Build GRCh38
rs370081789 Chr.1: 24358650-24358650 on Build GRCh38
rs1019563746 Chr.1: 24358858-24358858 on Build GRCh38
rs115184896 Chr.1: 24358798-24358798 on Build GRCh38
rs373190951 Chr.1: 24358505-24358505 on Build GRCh38
rs139587004 Chr.1: 24358541-24358541 on Build GRCh38
rs769989170 Chr.1: 24358772-24358772 on Build GRCh38
rs997372475 Chr.1: 24358899-24358899 on Build GRCh38
rs868073747 Chr.1: 24358548-24358548 on Build GRCh38
rs775581295 Chr.1: 24358545-24358545 on Build GRCh38
rs924442277 Chr.1: 24358599-24358599 on Build GRCh38
rs758787696 Chr.1: 24358571-24358571 on Build GRCh38
rs956166221 Chr.1: 24358950-24358950 on Build GRCh38
rs1055053151 Chr.1: 24358809-24358809 on Build GRCh38
rs775524801 Chr.1: 24358645-24358645 on Build GRCh38
rs759435827 Chr.1: 24358535-24358535 on Build GRCh38
rs763729229 Chr.1: 24358552-24358552 on Build GRCh38
rs1002859624 Chr.1: 24358924-24358924 on Build GRCh38
rs752772283 Chr.1: 24358661-24358661 on Build GRCh38
rs760384931 Chr.1: 24358912-24358912 on Build GRCh38
rs771996325 Chr.1: 24358613-24358613 on Build GRCh38
rs985142636 Chr.1: 24358749-24358749 on Build GRCh38
rs748283519 Chr.1: 24358582-24358582 on Build GRCh38
rs1006935288 Chr.1: 24358839-24358839 on Build GRCh38
rs199684576 Chr.1: 24358503-24358503 on Build GRCh38
rs766030527 Chr.1: 24358946-24358946 on Build GRCh38
rs58160842 Chr.1: 24358867-24358867 on Build GRCh38
rs74449398 Chr.1: 24358780-24358780 on Build GRCh38
rs377422598 Chr.1: 24358640-24358640 on Build GRCh38
rs768030098 Chr.1: 24358627-24358627 on Build GRCh38
rs775562958 Chr.1: 24358615-24358615 on Build GRCh38
rs757560497 Chr.1: 24358489-24358489 on Build GRCh38
rs769162044 Chr.1: 24358762-24358762 on Build GRCh38
rs111484075 Chr.1: 24358691-24358691 on Build GRCh38
rs113386084 Chr.1: 24358584-24358584 on Build GRCh38
rs577417902 Chr.1: 24358742-24358742 on Build GRCh38
rs1015519093 Chr.1: 24358685-24358685 on Build GRCh38
rs781379228 Chr.1: 24358581-24358581 on Build GRCh38
rs897356067 Chr.1: 24358508-24358508 on Build GRCh38
rs897301259 Chr.1: 24358767-24358767 on Build GRCh38
rs755035519 Chr.1: 24358578-24358578 on Build GRCh38
rs781115517 Chr.1: 24358656-24358656 on Build GRCh38
rs760194663 Chr.1: 24358546-24358546 on Build GRCh38
rs573583719 Chr.1: 24358588-24358588 on Build GRCh38
rs146480764 Chr.1: 24358560-24358560 on Build GRCh38
rs752036833 Chr.1: 24358646-24358646 on Build GRCh38
rs765074232 Chr.1: 24358573-24358573 on Build GRCh38
rs367604358 Chr.1: 24358626-24358626 on Build GRCh38
rs776246644 Chr.1: 24358634-24358634 on Build GRCh38
rs1044460988 Chr.1: 24358881-24358881 on Build GRCh38
rs767936084 Chr.1: 24358660-24358660 on Build GRCh38
rs777879742 Chr.1: 24358589-24358589 on Build GRCh38

View Full Product Details