Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24379574-24380068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STPG1
Entrez Gene ID
90529
Gene Name
sperm tail PG-rich repeat containing 1
Gene Aliases
C1orf201, MAPO2
UniGene
Hs.403187
Gene Chromosome Location
Chr.1: 24356999-24415533 on Build GRCh38
Transcripts
NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4947944 c.62C>T p.S21L Chr.1: 24379762-24379762 on Build GRCh38
1687223 c.245C>T p.S82L Chr.1: 24379729-24379729 on Build GRCh38
5380344 c.43A>C p.N15H Chr.1: 24379781-24379781 on Build GRCh38
5475665 c.192G>A p.A64A Chr.1: 24379782-24379782 on Build GRCh38
1152063 c.445G>A p.A149T Chr.1: 24379670-24379670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751484558 Chr.1: 24379778-24379778 on Build GRCh38
rs899615319 Chr.1: 24379972-24379972 on Build GRCh38
rs755595406 Chr.1: 24379862-24379862 on Build GRCh38
rs374777670 Chr.1: 24379686-24379686 on Build GRCh38
rs763334400 Chr.1: 24379604-24379604 on Build GRCh38
rs886885794 Chr.1: 24380025-24380025 on Build GRCh38
rs113054161 Chr.1: 24379677-24379677 on Build GRCh38
rs772868550 Chr.1: 24379715-24379715 on Build GRCh38
rs759999066 Chr.1: 24379742-24379742 on Build GRCh38
rs199840433 Chr.1: 24379764-24379764 on Build GRCh38
rs386629582 Chr.1: 24379970-24379971 on Build GRCh38
rs779764404 Chr.1: 24379773-24379773 on Build GRCh38
rs754179559 Chr.1: 24379770-24379770 on Build GRCh38
rs3765428 Chr.1: 24379623-24379623 on Build GRCh38
rs752070224 Chr.1: 24379615-24379615 on Build GRCh38
rs111326791 Chr.1: 24379761-24379761 on Build GRCh38
rs368278367 Chr.1: 24379769-24379769 on Build GRCh38
rs1064842 Chr.1: 24379779-24379779 on Build GRCh38
rs112031952 Chr.1: 24379618-24379618 on Build GRCh38
rs148922346 Chr.1: 24379814-24379814 on Build GRCh38
rs577828197 Chr.1: 24379918-24379918 on Build GRCh38
rs746979234 Chr.1: 24379660-24379660 on Build GRCh38
rs145625712 Chr.1: 24379654-24379654 on Build GRCh38
rs772399596 Chr.1: 24379811-24379811 on Build GRCh38
rs927377209 Chr.1: 24379650-24379650 on Build GRCh38
rs1020495964 Chr.1: 24380041-24380041 on Build GRCh38
rs147676822 Chr.1: 24379999-24379999 on Build GRCh38
rs774897144 Chr.1: 24379730-24379730 on Build GRCh38
rs754819557 Chr.1: 24379827-24379827 on Build GRCh38
rs899542323 Chr.1: 24379785-24379785 on Build GRCh38
rs914001055 Chr.1: 24379907-24379907 on Build GRCh38
rs781133070 Chr.1: 24379781-24379781 on Build GRCh38
rs958808172 Chr.1: 24379887-24379887 on Build GRCh38
rs771292787 Chr.1: 24379798-24379798 on Build GRCh38
rs35124440 Chr.1: 24379665-24379665 on Build GRCh38
rs966670401 Chr.1: 24379915-24379915 on Build GRCh38
rs949880769 Chr.1: 24380010-24380010 on Build GRCh38
rs1003893894 Chr.1: 24379794-24379794 on Build GRCh38
rs41307808 Chr.1: 24379839-24379839 on Build GRCh38
rs575725742 Chr.1: 24379998-24379998 on Build GRCh38
rs751266236 Chr.1: 24379826-24379826 on Build GRCh38
rs925892042 Chr.1: 24379953-24379953 on Build GRCh38
rs746264799 Chr.1: 24379809-24379809 on Build GRCh38
rs1045476993 Chr.1: 24380013-24380013 on Build GRCh38
rs751583945 Chr.1: 24379676-24379676 on Build GRCh38
rs375539520 Chr.1: 24379866-24379866 on Build GRCh38
rs371742348 Chr.1: 24379806-24379806 on Build GRCh38
rs145294929 Chr.1: 24379822-24379822 on Build GRCh38
rs41300112 Chr.1: 24379784-24379784 on Build GRCh38
rs11538188 Chr.1: 24379728-24379728 on Build GRCh38
rs758200725 Chr.1: 24379771-24379771 on Build GRCh38
rs1142057 Chr.1: 24379802-24379802 on Build GRCh38
rs986824096 Chr.1: 24379906-24379906 on Build GRCh38
rs892144037 Chr.1: 24380019-24380019 on Build GRCh38
rs139161643 Chr.1: 24379821-24379821 on Build GRCh38
rs3765431 Chr.1: 24379920-24379920 on Build GRCh38
rs752934316 Chr.1: 24379621-24379621 on Build GRCh38
rs913867063 Chr.1: 24379641-24379641 on Build GRCh38
rs778161649 Chr.1: 24379651-24379651 on Build GRCh38
rs138910562 Chr.1: 24379782-24379782 on Build GRCh38
rs112889972 Chr.1: 24379928-24379928 on Build GRCh38
rs41311987 Chr.1: 24379608-24379608 on Build GRCh38
rs748985785 Chr.1: 24379790-24379790 on Build GRCh38
rs763295960 Chr.1: 24379717-24379717 on Build GRCh38
rs764327921 Chr.1: 24379817-24379817 on Build GRCh38
rs929908247 Chr.1: 24379912-24379912 on Build GRCh38
rs149733917 Chr.1: 24379652-24379652 on Build GRCh38
rs546813317 Chr.1: 24379755-24379755 on Build GRCh38
rs3835230 Chr.1: 24379948-24379949 on Build GRCh38
rs747689812 Chr.1: 24379691-24379691 on Build GRCh38
rs553113072 Chr.1: 24379916-24379916 on Build GRCh38
rs886748786 Chr.1: 24379788-24379788 on Build GRCh38
rs147041081 Chr.1: 24379816-24379816 on Build GRCh38
rs780036113 Chr.1: 24379655-24379655 on Build GRCh38
rs979267957 Chr.1: 24379646-24379646 on Build GRCh38
rs776972860 Chr.1: 24379820-24379820 on Build GRCh38
rs777985540 Chr.1: 24379637-24379637 on Build GRCh38
rs142007584 Chr.1: 24379729-24379729 on Build GRCh38
rs3765432 Chr.1: 24379971-24379971 on Build GRCh38
rs767380057 Chr.1: 24379828-24379828 on Build GRCh38
rs397770183 Chr.1: 24379949-24379950 on Build GRCh38
rs551667574 Chr.1: 24379617-24379617 on Build GRCh38

View Full Product Details