Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24391474-24391984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STPG1
Entrez Gene ID
90529
Gene Name
sperm tail PG-rich repeat containing 1
Gene Aliases
C1orf201, MAPO2
UniGene
Hs.403187
Gene Chromosome Location
Chr.1: 24356999-24415533 on Build GRCh38
Transcripts
NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574515 c.186G>T p.K62N Chr.1: 24391564-24391564 on Build GRCh38
3741162 c.179C>G p.P60R Chr.1: 24391571-24391571 on Build GRCh38
1341003 c.150_151insA p.G51fs*4 Chr.1: 24391599-24391600 on Build GRCh38
5111083 c.80_81insA p.G28fs*4 Chr.1: 24391599-24391600 on Build GRCh38
4526032 c.136G>A p.E46K Chr.1: 24391614-24391614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933215118 Chr.1: 24391507-24391507 on Build GRCh38
rs138074662 Chr.1: 24391508-24391508 on Build GRCh38
rs373120395 Chr.1: 24391839-24391839 on Build GRCh38
rs397764285 Chr.1: 24391837-24391838 on Build GRCh38
rs755987462 Chr.1: 24391742-24391742 on Build GRCh38
rs376293143 Chr.1: 24391841-24391841 on Build GRCh38
rs749389338 Chr.1: 24391592-24391592 on Build GRCh38
rs773073076 Chr.1: 24391883-24391883 on Build GRCh38
rs1016372147 Chr.1: 24391876-24391876 on Build GRCh38
rs1013795171 Chr.1: 24391811-24391811 on Build GRCh38
rs569938583 Chr.1: 24391823-24391823 on Build GRCh38
rs755402455 Chr.1: 24391548-24391548 on Build GRCh38
rs868391249 Chr.1: 24391599-24391599 on Build GRCh38
rs941896426 Chr.1: 24391790-24391790 on Build GRCh38
rs747680990 Chr.1: 24391779-24391779 on Build GRCh38
rs537276257 Chr.1: 24391846-24391846 on Build GRCh38
rs555429968 Chr.1: 24391861-24391861 on Build GRCh38
rs768552853 Chr.1: 24391625-24391625 on Build GRCh38
rs397698281 Chr.1: 24391838-24391839 on Build GRCh38
rs760457476 Chr.1: 24391616-24391616 on Build GRCh38
rs774559373 Chr.1: 24391615-24391615 on Build GRCh38
rs890147097 Chr.1: 24391906-24391906 on Build GRCh38
rs1057196266 Chr.1: 24391746-24391746 on Build GRCh38
rs540134776 Chr.1: 24391649-24391649 on Build GRCh38
rs937064752 Chr.1: 24391698-24391698 on Build GRCh38
rs890677007 Chr.1: 24391557-24391557 on Build GRCh38
rs773918134 Chr.1: 24391756-24391756 on Build GRCh38
rs3215501 Chr.1: 24391828-24391829 on Build GRCh38
rs143632495 Chr.1: 24391549-24391549 on Build GRCh38
rs776410379 Chr.1: 24391670-24391670 on Build GRCh38
rs913070575 Chr.1: 24391542-24391542 on Build GRCh38
rs150334986 Chr.1: 24391497-24391497 on Build GRCh38
rs992806398 Chr.1: 24391673-24391673 on Build GRCh38
rs557896595 Chr.1: 24391793-24391793 on Build GRCh38
rs575245628 Chr.1: 24391941-24391941 on Build GRCh38
rs970594736 Chr.1: 24391697-24391697 on Build GRCh38
rs1034476759 Chr.1: 24391627-24391627 on Build GRCh38
rs541019651 Chr.1: 24391940-24391940 on Build GRCh38
rs2294520 Chr.1: 24391585-24391585 on Build GRCh38
rs758467038 Chr.1: 24391569-24391569 on Build GRCh38
rs1010495514 Chr.1: 24391561-24391561 on Build GRCh38
rs999583404 Chr.1: 24391796-24391796 on Build GRCh38
rs79437859 Chr.1: 24391957-24391957 on Build GRCh38
rs958948077 Chr.1: 24391662-24391662 on Build GRCh38
rs566198611 Chr.1: 24391606-24391606 on Build GRCh38
rs868195907 Chr.1: 24391858-24391858 on Build GRCh38
rs1006788835 Chr.1: 24391871-24391871 on Build GRCh38
rs904675316 Chr.1: 24391944-24391944 on Build GRCh38
rs560219 Chr.1: 24391589-24391589 on Build GRCh38
rs779885929 Chr.1: 24391827-24391827 on Build GRCh38
rs917248855 Chr.1: 24391696-24391696 on Build GRCh38
rs553341381 Chr.1: 24391945-24391945 on Build GRCh38
rs147211882 Chr.1: 24391882-24391882 on Build GRCh38
rs751424903 Chr.1: 24391717-24391717 on Build GRCh38
rs771081237 Chr.1: 24391601-24391601 on Build GRCh38
rs567405959 Chr.1: 24391562-24391562 on Build GRCh38

View Full Product Details