Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74209268-74209764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC33
Entrez Gene ID
80125
Gene Name
coiled-coil domain containing 33
Gene Aliases
CT61, HP11097
UniGene
Hs.383206
Gene Chromosome Location
Chr.15: 74202705-74336472 on Build GRCh38
Transcripts
XM_017022629.1 NM_001199040.1 XM_011521883.1 XM_017022479.1

Gene Symbol
STRA6
Entrez Gene ID
64220
Gene Name
stimulated by retinoic acid 6
Gene Aliases
MCOPCB8, MCOPS9, PP14296
UniGene
Hs.24553
Gene Chromosome Location
Chr.15: 74179466-74212267 on Build GRCh38
Transcripts
XM_017022629.1 NM_001199040.1 XM_011521883.1 XM_017022479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6572658 c.40C>T p.L14F Chr.15: 74209415-74209415 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs968025009 Chr.15: 74209692-74209692 on Build GRCh38
rs557196110 Chr.15: 74209422-74209422 on Build GRCh38
rs1049288702 Chr.15: 74209629-74209629 on Build GRCh38
rs901926459 Chr.15: 74209665-74209665 on Build GRCh38
rs1033358757 Chr.15: 74209707-74209707 on Build GRCh38
rs765756437 Chr.15: 74209392-74209392 on Build GRCh38
rs777923813 Chr.15: 74209611-74209611 on Build GRCh38
rs967327093 Chr.15: 74209410-74209410 on Build GRCh38
rs749059658 Chr.15: 74209500-74209500 on Build GRCh38
rs750020524 Chr.15: 74209444-74209444 on Build GRCh38
rs184924574 Chr.15: 74209398-74209398 on Build GRCh38
rs539628058 Chr.15: 74209431-74209431 on Build GRCh38
rs558111203 Chr.15: 74209433-74209433 on Build GRCh38
rs575482209 Chr.15: 74209430-74209430 on Build GRCh38
rs543873906 Chr.15: 74209680-74209680 on Build GRCh38
rs994494829 Chr.15: 74209397-74209397 on Build GRCh38
rs917001577 Chr.15: 74209445-74209445 on Build GRCh38
rs901003424 Chr.15: 74209321-74209321 on Build GRCh38
rs929796437 Chr.15: 74209722-74209722 on Build GRCh38
rs978665180 Chr.15: 74209420-74209420 on Build GRCh38
rs913464692 Chr.15: 74209635-74209635 on Build GRCh38
rs978762022 Chr.15: 74209653-74209653 on Build GRCh38
rs1008160462 Chr.15: 74209642-74209642 on Build GRCh38
rs1000136287 Chr.15: 74209706-74209706 on Build GRCh38
rs756564766 Chr.15: 74209574-74209574 on Build GRCh38
rs755321735 Chr.15: 74209394-74209394 on Build GRCh38
rs755758636 Chr.15: 74209461-74209461 on Build GRCh38
rs796228399 Chr.15: 74209522-74209522 on Build GRCh38
rs767163302 Chr.15: 74209435-74209435 on Build GRCh38
rs918923242 Chr.15: 74209570-74209570 on Build GRCh38
rs1009653188 Chr.15: 74209679-74209679 on Build GRCh38
rs901008817 Chr.15: 74209718-74209718 on Build GRCh38
rs970519664 Chr.15: 74209618-74209618 on Build GRCh38
rs1035992395 Chr.15: 74209514-74209514 on Build GRCh38
rs1041095399 Chr.15: 74209693-74209693 on Build GRCh38
rs868614438 Chr.15: 74209609-74209609 on Build GRCh38
rs1023286831 Chr.15: 74209441-74209441 on Build GRCh38
rs960203632 Chr.15: 74209582-74209582 on Build GRCh38
rs969152679 Chr.15: 74209567-74209567 on Build GRCh38
rs540262742 Chr.15: 74209505-74209505 on Build GRCh38
rs1019429367 Chr.15: 74209349-74209349 on Build GRCh38
rs573682061 Chr.15: 74209644-74209644 on Build GRCh38
rs138676653 Chr.15: 74209643-74209643 on Build GRCh38
rs180854138 Chr.15: 74209391-74209391 on Build GRCh38
rs1047440352 Chr.15: 74209725-74209725 on Build GRCh38
rs911214416 Chr.15: 74209676-74209676 on Build GRCh38
rs779863045 Chr.15: 74209484-74209484 on Build GRCh38
rs930930789 Chr.15: 74209617-74209617 on Build GRCh38
rs562755960 Chr.15: 74209740-74209740 on Build GRCh38
rs771112698 Chr.15: 74209595-74209595 on Build GRCh38
rs573051017 Chr.15: 74209477-74209477 on Build GRCh38
rs748687572 Chr.15: 74209573-74209573 on Build GRCh38

View Full Product Details