Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74243613-74244104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC33
Entrez Gene ID
80125
Gene Name
coiled-coil domain containing 33
Gene Aliases
CT61, HP11097
UniGene
Hs.383206
Gene Chromosome Location
Chr.15: 74202705-74336472 on Build GRCh38
Transcripts
NM_025055.4 XM_005254692.1 XM_011522085.2 XM_011522086.1 XM_011522087.2 XM_011522088.1 XM_017022623.1 XM_017022624.1 XM_017022625.1 XM_017022626.1 XM_017022627.1 XM_017022628.1 XM_017022629.1 XM_017022630.1 XM_017022631.1 XM_017022633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4254062 c.99G>T p.K33N Chr.15: 74244062-74244062 on Build GRCh38
6377925 c.100A>G p.K34E Chr.15: 74244063-74244063 on Build GRCh38
5849751 c.41A>C p.E14A Chr.15: 74244004-74244004 on Build GRCh38
6622075 c.680C>A p.P227H Chr.15: 74244034-74244034 on Build GRCh38
7175220 c.649-9G>A p.? Chr.15: 74243994-74243994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779233514 Chr.15: 74243774-74243774 on Build GRCh38
rs776485585 Chr.15: 74244067-74244067 on Build GRCh38
rs397774110 Chr.15: 74244063-74244065 on Build GRCh38
rs751712968 Chr.15: 74243936-74243937 on Build GRCh38
rs1032082529 Chr.15: 74243956-74243956 on Build GRCh38
rs754114257 Chr.15: 74243980-74243980 on Build GRCh38
rs766433450 Chr.15: 74244080-74244080 on Build GRCh38
rs746354219 Chr.15: 74244010-74244010 on Build GRCh38
rs35899977 Chr.15: 74244062-74244064 on Build GRCh38
rs755274774 Chr.15: 74243757-74243757 on Build GRCh38
rs892258036 Chr.15: 74243828-74243828 on Build GRCh38
rs866989530 Chr.15: 74244066-74244068 on Build GRCh38
rs1056310571 Chr.15: 74243780-74243780 on Build GRCh38
rs752582732 Chr.15: 74243851-74243851 on Build GRCh38
rs1046616450 Chr.15: 74243653-74243653 on Build GRCh38
rs370325525 Chr.15: 74244042-74244042 on Build GRCh38
rs565856732 Chr.15: 74244059-74244059 on Build GRCh38
rs773127194 Chr.15: 74243691-74243692 on Build GRCh38
rs748718388 Chr.15: 74243778-74243778 on Build GRCh38
rs373400480 Chr.15: 74243641-74243641 on Build GRCh38
rs67845844 Chr.15: 74243945-74243945 on Build GRCh38
rs779286933 Chr.15: 74244005-74244005 on Build GRCh38
rs781766558 Chr.15: 74243944-74243945 on Build GRCh38
rs879061768 Chr.15: 74243946-74243946 on Build GRCh38
rs796916684 Chr.15: 74243944-74243945 on Build GRCh38
rs766633022 Chr.15: 74243978-74243978 on Build GRCh38
rs930069996 Chr.15: 74243733-74243733 on Build GRCh38
rs991511295 Chr.15: 74243634-74243634 on Build GRCh38
rs990520806 Chr.15: 74243996-74243996 on Build GRCh38
rs78218275 Chr.15: 74243957-74243957 on Build GRCh38
rs767307838 Chr.15: 74243974-74243974 on Build GRCh38
rs1040070599 Chr.15: 74243776-74243776 on Build GRCh38
rs199782040 Chr.15: 74244063-74244063 on Build GRCh38
rs10552820 Chr.15: 74244060-74244062 on Build GRCh38
rs1048507505 Chr.15: 74243772-74243772 on Build GRCh38
rs79769980 Chr.15: 74243961-74243962 on Build GRCh38
rs997359301 Chr.15: 74243803-74243803 on Build GRCh38
rs77688424 Chr.15: 74243637-74243637 on Build GRCh38
rs749657011 Chr.15: 74243741-74243741 on Build GRCh38
rs753598268 Chr.15: 74243773-74243773 on Build GRCh38
rs779356834 Chr.15: 74243959-74243960 on Build GRCh38
rs753882238 Chr.15: 74244083-74244083 on Build GRCh38
rs1017796462 Chr.15: 74244067-74244067 on Build GRCh38
rs756080402 Chr.15: 74243976-74243976 on Build GRCh38
rs750546260 Chr.15: 74243975-74243975 on Build GRCh38
rs760643974 Chr.15: 74244075-74244075 on Build GRCh38
rs756292982 Chr.15: 74243945-74243946 on Build GRCh38
rs774938868 Chr.15: 74243954-74243954 on Build GRCh38
rs769521729 Chr.15: 74243948-74243948 on Build GRCh38
rs146921233 Chr.15: 74244025-74244025 on Build GRCh38
rs755214067 Chr.15: 74243670-74243670 on Build GRCh38
rs754093141 Chr.15: 74243945-74243947 on Build GRCh38
rs1045153490 Chr.15: 74243711-74243711 on Build GRCh38
rs758020026 Chr.15: 74244047-74244047 on Build GRCh38
rs373604550 Chr.15: 74244029-74244029 on Build GRCh38
rs201189801 Chr.15: 74244059-74244061 on Build GRCh38
rs748493188 Chr.15: 74244006-74244006 on Build GRCh38
rs776257067 Chr.15: 74243940-74243940 on Build GRCh38
rs767493515 Chr.15: 74244054-74244054 on Build GRCh38
rs926708503 Chr.15: 74243739-74243739 on Build GRCh38
rs561499906 Chr.15: 74243649-74243649 on Build GRCh38
rs756379727 Chr.15: 74243686-74243686 on Build GRCh38
rs764039686 Chr.15: 74243958-74243958 on Build GRCh38
rs377200305 Chr.15: 74244041-74244041 on Build GRCh38
rs774285235 Chr.15: 74243959-74243959 on Build GRCh38
rs756924909 Chr.15: 74243904-74243904 on Build GRCh38
rs780098643 Chr.15: 74243723-74243723 on Build GRCh38
rs767976943 Chr.15: 74243944-74243944 on Build GRCh38
rs755185146 Chr.15: 74243981-74243981 on Build GRCh38
rs757788244 Chr.15: 74243945-74243948 on Build GRCh38
rs758969893 Chr.15: 74244013-74244013 on Build GRCh38
rs531931469 Chr.15: 74243654-74243654 on Build GRCh38
rs775127307 Chr.15: 74244028-74244028 on Build GRCh38
rs778105434 Chr.15: 74244014-74244014 on Build GRCh38
rs778321793 Chr.15: 74243972-74243972 on Build GRCh38
rs745324786 Chr.15: 74243945-74243945 on Build GRCh38
rs768132160 Chr.15: 74244063-74244064 on Build GRCh38
rs772539624 Chr.15: 74243939-74243939 on Build GRCh38
rs781455124 Chr.15: 74243674-74243674 on Build GRCh38
rs398027949 Chr.15: 74243959-74243959 on Build GRCh38
rs1009492880 Chr.15: 74244000-74244000 on Build GRCh38
rs900434320 Chr.15: 74243779-74243779 on Build GRCh38
rs201484902 Chr.15: 74244062-74244062 on Build GRCh38
rs189088694 Chr.15: 74244015-74244015 on Build GRCh38

View Full Product Details