Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74279836-74280334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC33
Entrez Gene ID
80125
Gene Name
coiled-coil domain containing 33
Gene Aliases
CT61, HP11097
UniGene
Hs.383206
Gene Chromosome Location
Chr.15: 74202705-74336472 on Build GRCh38
Transcripts
NM_025055.4 XM_005254692.1 XM_011522085.2 XM_011522086.1 XM_011522087.2 XM_011522088.1 XM_011522089.2 XM_017022623.1 XM_017022624.1 XM_017022625.1 XM_017022626.1 XM_017022627.1 XM_017022628.1 XM_017022629.1 XM_017022630.1 XM_017022631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7311478 c.1396C>T p.Q466* Chr.15: 74279990-74279990 on Build GRCh38
5027999 c.845C>G p.S282* Chr.15: 74280048-74280048 on Build GRCh38
5580547 c.1459G>A p.D487N Chr.15: 74280053-74280053 on Build GRCh38
1640336 c.889+4C>T p.? Chr.15: 74280096-74280096 on Build GRCh38
5584721 c.1373C>T p.S458F Chr.15: 74279967-74279967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994848205 Chr.15: 74280185-74280185 on Build GRCh38
rs932174441 Chr.15: 74280086-74280086 on Build GRCh38
rs351165 Chr.15: 74279901-74279901 on Build GRCh38
rs921019290 Chr.15: 74280052-74280052 on Build GRCh38
rs201820602 Chr.15: 74279883-74279883 on Build GRCh38
rs975098154 Chr.15: 74280310-74280310 on Build GRCh38
rs775097538 Chr.15: 74280138-74280138 on Build GRCh38
rs913942170 Chr.15: 74280287-74280287 on Build GRCh38
rs376118208 Chr.15: 74280026-74280026 on Build GRCh38
rs751306190 Chr.15: 74279960-74279960 on Build GRCh38
rs1024059861 Chr.15: 74279965-74279965 on Build GRCh38
rs562483815 Chr.15: 74280273-74280273 on Build GRCh38
rs946710571 Chr.15: 74280289-74280289 on Build GRCh38
rs749016637 Chr.15: 74280106-74280107 on Build GRCh38
rs74023464 Chr.15: 74279871-74279871 on Build GRCh38
rs751361397 Chr.15: 74280070-74280070 on Build GRCh38
rs777562257 Chr.15: 74280063-74280064 on Build GRCh38
rs761079944 Chr.15: 74280040-74280040 on Build GRCh38
rs1024112220 Chr.15: 74280248-74280248 on Build GRCh38
rs554269697 Chr.15: 74280274-74280274 on Build GRCh38
rs894569325 Chr.15: 74280219-74280219 on Build GRCh38
rs768094124 Chr.15: 74279917-74279917 on Build GRCh38
rs545000745 Chr.15: 74280308-74280308 on Build GRCh38
rs764842007 Chr.15: 74280058-74280058 on Build GRCh38
rs1032872057 Chr.15: 74280269-74280269 on Build GRCh38
rs763697038 Chr.15: 74280063-74280063 on Build GRCh38
rs765926271 Chr.15: 74280027-74280027 on Build GRCh38
rs1043698410 Chr.15: 74280311-74280311 on Build GRCh38
rs755978836 Chr.15: 74280088-74280088 on Build GRCh38
rs541229249 Chr.15: 74280049-74280049 on Build GRCh38
rs769001580 Chr.15: 74280107-74280107 on Build GRCh38
rs1052455480 Chr.15: 74279905-74279905 on Build GRCh38
rs114872868 Chr.15: 74280249-74280249 on Build GRCh38
rs759027301 Chr.15: 74279927-74279927 on Build GRCh38
rs752125536 Chr.15: 74280060-74280060 on Build GRCh38
rs781042993 Chr.15: 74279972-74279972 on Build GRCh38
rs1010187826 Chr.15: 74279929-74279930 on Build GRCh38
rs773826161 Chr.15: 74280039-74280039 on Build GRCh38
rs749283081 Chr.15: 74280097-74280097 on Build GRCh38
rs756989407 Chr.15: 74280072-74280072 on Build GRCh38
rs749567762 Chr.15: 74279984-74279984 on Build GRCh38
rs1036584814 Chr.15: 74280112-74280112 on Build GRCh38
rs185714663 Chr.15: 74279977-74279977 on Build GRCh38
rs962008296 Chr.15: 74280110-74280110 on Build GRCh38
rs1056318812 Chr.15: 74280191-74280191 on Build GRCh38
rs1014717398 Chr.15: 74280238-74280238 on Build GRCh38
rs777594904 Chr.15: 74279959-74279959 on Build GRCh38
rs752459365 Chr.15: 74279937-74279937 on Build GRCh38
rs368483325 Chr.15: 74280079-74280079 on Build GRCh38
rs764639507 Chr.15: 74279928-74279928 on Build GRCh38
rs769976545 Chr.15: 74279979-74279979 on Build GRCh38
rs920849949 Chr.15: 74280141-74280141 on Build GRCh38
rs572431031 Chr.15: 74280091-74280091 on Build GRCh38
rs199609536 Chr.15: 74280032-74280032 on Build GRCh38
rs554268267 Chr.15: 74280014-74280014 on Build GRCh38
rs188873868 Chr.15: 74280266-74280266 on Build GRCh38
rs780955188 Chr.15: 74280077-74280077 on Build GRCh38
rs757220070 Chr.15: 74279961-74279961 on Build GRCh38
rs779997632 Chr.15: 74279983-74279983 on Build GRCh38
rs555268456 Chr.15: 74280140-74280140 on Build GRCh38
rs762561864 Chr.15: 74280036-74280036 on Build GRCh38
rs762617304 Chr.15: 74280061-74280061 on Build GRCh38
rs375511346 Chr.15: 74280128-74280128 on Build GRCh38
rs773418759 Chr.15: 74280120-74280120 on Build GRCh38
rs1013938969 Chr.15: 74280096-74280096 on Build GRCh38
rs757955091 Chr.15: 74279938-74279938 on Build GRCh38
rs11853660 Chr.15: 74280221-74280221 on Build GRCh38
rs114214926 Chr.15: 74280117-74280117 on Build GRCh38
rs62003683 Chr.15: 74279878-74279878 on Build GRCh38
rs753399872 Chr.15: 74279923-74279923 on Build GRCh38
rs771294414 Chr.15: 74280133-74280133 on Build GRCh38
rs577579152 Chr.15: 74279910-74279910 on Build GRCh38
rs779216280 Chr.15: 74280114-74280114 on Build GRCh38
rs977285629 Chr.15: 74279990-74279990 on Build GRCh38
rs750594576 Chr.15: 74280215-74280215 on Build GRCh38

View Full Product Details