Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74338486-74338985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
UniGene
Hs.303980
Gene Chromosome Location
Chr.15: 74337762-74367740 on Build GRCh38
Transcripts
NM_000781.2 NM_001099773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849763 c.1411G>A p.E471K Chr.15: 74338594-74338594 on Build GRCh38
3887488 c.1351C>T p.R451W Chr.15: 74338654-74338654 on Build GRCh38
4578491 c.1290C>T p.F430F Chr.15: 74338715-74338715 on Build GRCh38
271189 c.1401C>T p.I467I Chr.15: 74338604-74338604 on Build GRCh38
6638991 c.1291G>A p.D431N Chr.15: 74338714-74338714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763985808 Chr.15: 74338597-74338597 on Build GRCh38
rs749515942 Chr.15: 74338616-74338616 on Build GRCh38
rs980016533 Chr.15: 74338637-74338637 on Build GRCh38
rs146830942 Chr.15: 74338670-74338670 on Build GRCh38
rs762412759 Chr.15: 74338735-74338735 on Build GRCh38
rs141235847 Chr.15: 74338612-74338612 on Build GRCh38
rs900306752 Chr.15: 74338714-74338714 on Build GRCh38
rs140651916 Chr.15: 74338807-74338807 on Build GRCh38
rs766351201 Chr.15: 74338774-74338774 on Build GRCh38
rs923914796 Chr.15: 74338954-74338954 on Build GRCh38
rs991398839 Chr.15: 74338847-74338847 on Build GRCh38
rs1051487436 Chr.15: 74338726-74338726 on Build GRCh38
rs376136531 Chr.15: 74338531-74338531 on Build GRCh38
rs755975808 Chr.15: 74338690-74338690 on Build GRCh38
rs781028010 Chr.15: 74338604-74338604 on Build GRCh38
rs759185471 Chr.15: 74338555-74338555 on Build GRCh38
rs368047178 Chr.15: 74338609-74338609 on Build GRCh38
rs761206886 Chr.15: 74338650-74338650 on Build GRCh38
rs182055225 Chr.15: 74338939-74338939 on Build GRCh38
rs747431545 Chr.15: 74338817-74338817 on Build GRCh38
rs779043334 Chr.15: 74338710-74338710 on Build GRCh38
rs749547595 Chr.15: 74338673-74338673 on Build GRCh38
rs774511568 Chr.15: 74338520-74338520 on Build GRCh38
rs535782968 Chr.15: 74338627-74338627 on Build GRCh38
rs760627282 Chr.15: 74338765-74338765 on Build GRCh38
rs538118268 Chr.15: 74338659-74338659 on Build GRCh38
rs1007830066 Chr.15: 74338862-74338862 on Build GRCh38
rs778230506 Chr.15: 74338662-74338662 on Build GRCh38
rs750370186 Chr.15: 74338682-74338682 on Build GRCh38
rs955973655 Chr.15: 74338924-74338924 on Build GRCh38
rs773652136 Chr.15: 74338644-74338644 on Build GRCh38
rs775102947 Chr.15: 74338654-74338654 on Build GRCh38
rs147405344 Chr.15: 74338715-74338715 on Build GRCh38
rs978196986 Chr.15: 74338941-74338941 on Build GRCh38
rs778216958 Chr.15: 74338729-74338729 on Build GRCh38
rs376845671 Chr.15: 74338755-74338755 on Build GRCh38
rs758061011 Chr.15: 74338591-74338591 on Build GRCh38
rs778308676 Chr.15: 74338811-74338811 on Build GRCh38
rs372523428 Chr.15: 74338610-74338610 on Build GRCh38
rs970788582 Chr.15: 74338892-74338892 on Build GRCh38
rs571183316 Chr.15: 74338709-74338709 on Build GRCh38
rs748510040 Chr.15: 74338618-74338618 on Build GRCh38
rs780138488 Chr.15: 74338696-74338696 on Build GRCh38
rs995747544 Chr.15: 74338800-74338800 on Build GRCh38
rs576685829 Chr.15: 74338910-74338910 on Build GRCh38
rs771393499 Chr.15: 74338820-74338820 on Build GRCh38
rs773746289 Chr.15: 74338540-74338540 on Build GRCh38
rs757055824 Chr.15: 74338603-74338603 on Build GRCh38
rs761467521 Chr.15: 74338739-74338739 on Build GRCh38
rs768205181 Chr.15: 74338736-74338736 on Build GRCh38
rs755072877 Chr.15: 74338795-74338795 on Build GRCh38
rs189901407 Chr.15: 74338784-74338784 on Build GRCh38
rs918002510 Chr.15: 74338608-74338608 on Build GRCh38
rs914444285 Chr.15: 74338580-74338580 on Build GRCh38
rs567242447 Chr.15: 74338530-74338530 on Build GRCh38
rs1019309635 Chr.15: 74338552-74338552 on Build GRCh38
rs778995024 Chr.15: 74338801-74338801 on Build GRCh38
rs746015534 Chr.15: 74338836-74338836 on Build GRCh38
rs1004621798 Chr.15: 74338741-74338741 on Build GRCh38
rs774799229 Chr.15: 74338617-74338617 on Build GRCh38
rs748390637 Chr.15: 74338711-74338711 on Build GRCh38
rs1054131477 Chr.15: 74338678-74338678 on Build GRCh38
rs555167028 Chr.15: 74338909-74338909 on Build GRCh38
rs953733004 Chr.15: 74338520-74338521 on Build GRCh38
rs753866013 Chr.15: 74338703-74338703 on Build GRCh38
rs767327902 Chr.15: 74338680-74338680 on Build GRCh38
rs764826737 Chr.15: 74338557-74338557 on Build GRCh38
rs771520081 Chr.15: 74338730-74338730 on Build GRCh38
rs121912814 Chr.15: 74338761-74338761 on Build GRCh38
rs752867507 Chr.15: 74338803-74338803 on Build GRCh38
rs565141497 Chr.15: 74338962-74338962 on Build GRCh38
rs879245016 Chr.15: 74338762-74338762 on Build GRCh38
rs764844923 Chr.15: 74338653-74338653 on Build GRCh38
rs752413112 Chr.15: 74338583-74338583 on Build GRCh38
rs753531363 Chr.15: 74338715-74338717 on Build GRCh38
rs543226650 Chr.15: 74338852-74338852 on Build GRCh38
rs890065135 Chr.15: 74338824-74338824 on Build GRCh38
rs777258935 Chr.15: 74338734-74338734 on Build GRCh38
rs1025181635 Chr.15: 74338872-74338872 on Build GRCh38
rs767217819 Chr.15: 74338754-74338754 on Build GRCh38

View Full Product Details