Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74671377-74671882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
UniGene
Hs.682454
Gene Chromosome Location
Chr.15: 74630558-74696045 on Build GRCh38
Transcripts
NM_001142443.1 NM_001142444.1 NM_025083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69238 c.165-2A>G p.? Chr.15: 74671776-74671776 on Build GRCh38
3503955 c.439G>A p.E147K Chr.15: 74671500-74671500 on Build GRCh38
5383827 c.351C>T p.I117I Chr.15: 74671588-74671588 on Build GRCh38
5383828 c.324C>T p.A108A Chr.15: 74671615-74671615 on Build GRCh38
3503956 c.234C>T p.D78D Chr.15: 74671705-74671705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762672355 Chr.15: 74671469-74671469 on Build GRCh38
rs759094705 Chr.15: 74671433-74671433 on Build GRCh38
rs777978304 Chr.15: 74671635-74671635 on Build GRCh38
rs759682563 Chr.15: 74671735-74671735 on Build GRCh38
rs561864930 Chr.15: 74671405-74671405 on Build GRCh38
rs757001010 Chr.15: 74671606-74671606 on Build GRCh38
rs748562304 Chr.15: 74671515-74671515 on Build GRCh38
rs750261315 Chr.15: 74671488-74671488 on Build GRCh38
rs1009263319 Chr.15: 74671419-74671419 on Build GRCh38
rs948455779 Chr.15: 74671795-74671795 on Build GRCh38
rs928321229 Chr.15: 74671821-74671821 on Build GRCh38
rs774119590 Chr.15: 74671682-74671682 on Build GRCh38
rs778414721 Chr.15: 74671520-74671520 on Build GRCh38
rs779114659 Chr.15: 74671584-74671584 on Build GRCh38
rs1040929364 Chr.15: 74671832-74671832 on Build GRCh38
rs758859891 Chr.15: 74671625-74671625 on Build GRCh38
rs148327581 Chr.15: 74671544-74671544 on Build GRCh38
rs780091375 Chr.15: 74671501-74671501 on Build GRCh38
rs760503949 Chr.15: 74671571-74671571 on Build GRCh38
rs902125780 Chr.15: 74671849-74671849 on Build GRCh38
rs1003838369 Chr.15: 74671466-74671466 on Build GRCh38
rs758517938 Chr.15: 74671755-74671755 on Build GRCh38
rs748819356 Chr.15: 74671677-74671677 on Build GRCh38
rs544178799 Chr.15: 74671472-74671472 on Build GRCh38
rs767211729 Chr.15: 74671552-74671552 on Build GRCh38
rs754592279 Chr.15: 74671780-74671780 on Build GRCh38
rs746813483 Chr.15: 74671818-74671818 on Build GRCh38
rs542158660 Chr.15: 74671551-74671551 on Build GRCh38
rs77655432 Chr.15: 74671753-74671753 on Build GRCh38
rs753606943 Chr.15: 74671582-74671583 on Build GRCh38
rs773815619 Chr.15: 74671547-74671547 on Build GRCh38
rs376915971 Chr.15: 74671413-74671413 on Build GRCh38
rs752863254 Chr.15: 74671598-74671598 on Build GRCh38
rs202013828 Chr.15: 74671783-74671783 on Build GRCh38
rs771651776 Chr.15: 74671809-74671809 on Build GRCh38
rs917872534 Chr.15: 74671589-74671589 on Build GRCh38
rs1007558701 Chr.15: 74671594-74671594 on Build GRCh38
rs761464346 Chr.15: 74671474-74671474 on Build GRCh38
rs746372718 Chr.15: 74671771-74671771 on Build GRCh38
rs906829547 Chr.15: 74671410-74671410 on Build GRCh38
rs767501602 Chr.15: 74671475-74671475 on Build GRCh38
rs147438242 Chr.15: 74671683-74671683 on Build GRCh38
rs779063604 Chr.15: 74671513-74671513 on Build GRCh38
rs919162871 Chr.15: 74671698-74671698 on Build GRCh38
rs1022854363 Chr.15: 74671574-74671574 on Build GRCh38
rs749503885 Chr.15: 74671404-74671404 on Build GRCh38
rs939926716 Chr.15: 74671603-74671604 on Build GRCh38
rs968108909 Chr.15: 74671586-74671586 on Build GRCh38
rs766482884 Chr.15: 74671580-74671580 on Build GRCh38
rs763893555 Chr.15: 74671407-74671408 on Build GRCh38
rs760420471 Chr.15: 74671707-74671707 on Build GRCh38
rs747870951 Chr.15: 74671786-74671786 on Build GRCh38
rs764338631 Chr.15: 74671759-74671759 on Build GRCh38
rs376760239 Chr.15: 74671503-74671503 on Build GRCh38
rs766322816 Chr.15: 74671709-74671709 on Build GRCh38
rs771598019 Chr.15: 74671652-74671652 on Build GRCh38
rs113901995 Chr.15: 74671509-74671509 on Build GRCh38
rs747461655 Chr.15: 74671525-74671525 on Build GRCh38
rs751892786 Chr.15: 74671760-74671760 on Build GRCh38
rs753948126 Chr.15: 74671583-74671583 on Build GRCh38
rs377587628 Chr.15: 74671421-74671421 on Build GRCh38
rs756283540 Chr.15: 74671496-74671496 on Build GRCh38
rs774671939 Chr.15: 74671408-74671408 on Build GRCh38
rs750428769 Chr.15: 74671566-74671566 on Build GRCh38
rs370722720 Chr.15: 74671440-74671440 on Build GRCh38
rs757635961 Chr.15: 74671765-74671765 on Build GRCh38
rs776488328 Chr.15: 74671727-74671727 on Build GRCh38
rs546850263 Chr.15: 74671839-74671839 on Build GRCh38
rs765222222 Chr.15: 74671737-74671737 on Build GRCh38
rs753017071 Chr.15: 74671751-74671751 on Build GRCh38
rs979152587 Chr.15: 74671473-74671473 on Build GRCh38
rs772846860 Chr.15: 74671686-74671686 on Build GRCh38
rs772261057 Chr.15: 74671518-74671518 on Build GRCh38
rs552199641 Chr.15: 74671667-74671667 on Build GRCh38
rs781765253 Chr.15: 74671769-74671769 on Build GRCh38
rs772868320 Chr.15: 74671812-74671812 on Build GRCh38
rs142875618 Chr.15: 74671680-74671680 on Build GRCh38
rs764023124 Chr.15: 74671470-74671470 on Build GRCh38
rs967809528 Chr.15: 74671448-74671448 on Build GRCh38
rs200931517 Chr.15: 74671543-74671543 on Build GRCh38
rs767215558 Chr.15: 74671545-74671545 on Build GRCh38
rs775342225 Chr.15: 74671454-74671454 on Build GRCh38
rs909012007 Chr.15: 74671725-74671725 on Build GRCh38
rs755158567 Chr.15: 74671512-74671512 on Build GRCh38
rs747591587 Chr.15: 74671646-74671646 on Build GRCh38
rs769477794 Chr.15: 74671531-74671531 on Build GRCh38
rs373160392 Chr.15: 74671416-74671416 on Build GRCh38

View Full Product Details