Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74801013-74801512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSK
Entrez Gene ID
1445
Gene Name
c-src tyrosine kinase
Gene Aliases
-
UniGene
Hs.77793
Gene Chromosome Location
Chr.15: 74782064-74803198 on Build GRCh38
Transcripts
NM_001127190.1 NM_004383.2 XM_005254165.4 XM_017021925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4507694 c.753C>T p.L251L Chr.15: 74801042-74801042 on Build GRCh38
6636185 c.766G>A p.V256M Chr.15: 74801055-74801055 on Build GRCh38
5572512 c.803A>G p.Y268C Chr.15: 74801092-74801092 on Build GRCh38
4056921 c.781G>A p.G261R Chr.15: 74801070-74801070 on Build GRCh38
232104 c.785T>G p.L262R Chr.15: 74801074-74801074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758389871 Chr.15: 74801146-74801146 on Build GRCh38
rs754759286 Chr.15: 74801070-74801070 on Build GRCh38
rs181902017 Chr.15: 74801484-74801484 on Build GRCh38
rs753454254 Chr.15: 74801123-74801123 on Build GRCh38
rs759767225 Chr.15: 74801104-74801104 on Build GRCh38
rs112518573 Chr.15: 74801433-74801433 on Build GRCh38
rs930322089 Chr.15: 74801360-74801360 on Build GRCh38
rs541407225 Chr.15: 74801125-74801125 on Build GRCh38
rs998727243 Chr.15: 74801448-74801448 on Build GRCh38
rs922302783 Chr.15: 74801411-74801411 on Build GRCh38
rs752533008 Chr.15: 74801137-74801137 on Build GRCh38
rs1031624443 Chr.15: 74801189-74801189 on Build GRCh38
rs912776734 Chr.15: 74801262-74801262 on Build GRCh38
rs918906141 Chr.15: 74801356-74801356 on Build GRCh38
rs367785871 Chr.15: 74801476-74801476 on Build GRCh38
rs778720882 Chr.15: 74801075-74801075 on Build GRCh38
rs987395570 Chr.15: 74801261-74801261 on Build GRCh38
rs33991421 Chr.15: 74801081-74801081 on Build GRCh38
rs770081932 Chr.15: 74801107-74801107 on Build GRCh38
rs777666651 Chr.15: 74801147-74801147 on Build GRCh38
rs1028386141 Chr.15: 74801480-74801480 on Build GRCh38
rs371619768 Chr.15: 74801122-74801122 on Build GRCh38
rs543257080 Chr.15: 74801339-74801339 on Build GRCh38
rs35896486 Chr.15: 74801457-74801457 on Build GRCh38
rs560867771 Chr.15: 74801221-74801221 on Build GRCh38
rs954210108 Chr.15: 74801242-74801242 on Build GRCh38
rs1012214297 Chr.15: 74801260-74801260 on Build GRCh38
rs970960202 Chr.15: 74801264-74801264 on Build GRCh38
rs961632046 Chr.15: 74801282-74801282 on Build GRCh38
rs35465927 Chr.15: 74801439-74801439 on Build GRCh38
rs542531751 Chr.15: 74801200-74801200 on Build GRCh38
rs149657503 Chr.15: 74801054-74801054 on Build GRCh38
rs970971603 Chr.15: 74801036-74801036 on Build GRCh38
rs201236561 Chr.15: 74801136-74801136 on Build GRCh38
rs371828294 Chr.15: 74801152-74801152 on Build GRCh38
rs139139283 Chr.15: 74801469-74801469 on Build GRCh38
rs565511204 Chr.15: 74801409-74801409 on Build GRCh38
rs778434377 Chr.15: 74801387-74801387 on Build GRCh38
rs369845435 Chr.15: 74801471-74801471 on Build GRCh38
rs34983171 Chr.15: 74801401-74801401 on Build GRCh38
rs752623253 Chr.15: 74801164-74801164 on Build GRCh38
rs564626792 Chr.15: 74801378-74801378 on Build GRCh38
rs770046997 Chr.15: 74801474-74801474 on Build GRCh38
rs771931241 Chr.15: 74801082-74801082 on Build GRCh38
rs984706402 Chr.15: 74801372-74801372 on Build GRCh38
rs753705973 Chr.15: 74801057-74801057 on Build GRCh38
rs763472074 Chr.15: 74801043-74801043 on Build GRCh38
rs894292975 Chr.15: 74801139-74801139 on Build GRCh38
rs35058771 Chr.15: 74801192-74801192 on Build GRCh38
rs1000456620 Chr.15: 74801229-74801229 on Build GRCh38
rs773675209 Chr.15: 74801485-74801485 on Build GRCh38
rs771193750 Chr.15: 74801095-74801095 on Build GRCh38
rs942338674 Chr.15: 74801144-74801144 on Build GRCh38
rs1045981266 Chr.15: 74801434-74801434 on Build GRCh38
rs16953877 Chr.15: 74801239-74801239 on Build GRCh38
rs918730317 Chr.15: 74801320-74801320 on Build GRCh38
rs781597035 Chr.15: 74801486-74801486 on Build GRCh38
rs749945630 Chr.15: 74801049-74801049 on Build GRCh38
rs201794218 Chr.15: 74801109-74801109 on Build GRCh38
rs746077965 Chr.15: 74801154-74801154 on Build GRCh38
rs766534464 Chr.15: 74801055-74801055 on Build GRCh38
rs749999050 Chr.15: 74801116-74801116 on Build GRCh38
rs769338558 Chr.15: 74801488-74801488 on Build GRCh38
rs757348109 Chr.15: 74801151-74801151 on Build GRCh38
rs577460849 Chr.15: 74801113-74801113 on Build GRCh38
rs930375989 Chr.15: 74801069-74801069 on Build GRCh38
rs1000000020 Chr.15: 74801449-74801449 on Build GRCh38
rs371737390 Chr.15: 74801493-74801493 on Build GRCh38
rs747080333 Chr.15: 74801086-74801086 on Build GRCh38
rs958117974 Chr.15: 74801258-74801258 on Build GRCh38
rs776689754 Chr.15: 74801103-74801103 on Build GRCh38
rs764681458 Chr.15: 74801115-74801115 on Build GRCh38
rs368354202 Chr.15: 74801110-74801110 on Build GRCh38
rs2229730 Chr.15: 74801048-74801048 on Build GRCh38
rs113763792 Chr.15: 74801124-74801124 on Build GRCh38
rs893126639 Chr.15: 74801458-74801458 on Build GRCh38
rs755021838 Chr.15: 74801148-74801148 on Build GRCh38
rs375310133 Chr.15: 74801191-74801191 on Build GRCh38
rs777897715 Chr.15: 74801085-74801085 on Build GRCh38
rs760229578 Chr.15: 74801120-74801120 on Build GRCh38
rs189278716 Chr.15: 74801405-74801405 on Build GRCh38
rs1048771822 Chr.15: 74801365-74801365 on Build GRCh38
rs1054698328 Chr.15: 74801455-74801455 on Build GRCh38
rs756893514 Chr.15: 74801299-74801299 on Build GRCh38
rs907116754 Chr.15: 74801157-74801157 on Build GRCh38
rs973017990 Chr.15: 74801292-74801292 on Build GRCh38

View Full Product Details