Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74929005-74929514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX5A
Entrez Gene ID
9377
Gene Name
cytochrome c oxidase subunit 5A
Gene Aliases
COX, COX-VA, VA
UniGene
Hs.401903
Gene Chromosome Location
Chr.15: 74920275-74938154 on Build GRCh38
Transcripts
NM_004255.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233505 c.164G>A p.R55H Chr.15: 74929169-74929169 on Build GRCh38
4921222 c.172A>G p.T58A Chr.15: 74929161-74929161 on Build GRCh38
964940 c.115C>T p.R39C Chr.15: 74929218-74929218 on Build GRCh38
4935325 c.120C>G p.C40W Chr.15: 74929213-74929213 on Build GRCh38
4961835 c.213T>A p.R71R Chr.15: 74929120-74929120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575567611 Chr.15: 74929139-74929139 on Build GRCh38
rs775306455 Chr.15: 74929204-74929204 on Build GRCh38
rs375830469 Chr.15: 74929188-74929188 on Build GRCh38
rs912640190 Chr.15: 74929329-74929329 on Build GRCh38
rs1051375831 Chr.15: 74929365-74929365 on Build GRCh38
rs751665674 Chr.15: 74929134-74929134 on Build GRCh38
rs372049492 Chr.15: 74929085-74929085 on Build GRCh38
rs749742231 Chr.15: 74929074-74929074 on Build GRCh38
rs958559739 Chr.15: 74929489-74929489 on Build GRCh38
rs561670311 Chr.15: 74929322-74929322 on Build GRCh38
rs932862668 Chr.15: 74929363-74929363 on Build GRCh38
rs899083955 Chr.15: 74929475-74929475 on Build GRCh38
rs774638701 Chr.15: 74929217-74929217 on Build GRCh38
rs369677783 Chr.15: 74929461-74929461 on Build GRCh38
rs994999479 Chr.15: 74929413-74929413 on Build GRCh38
rs372249618 Chr.15: 74929205-74929205 on Build GRCh38
rs765729197 Chr.15: 74929243-74929243 on Build GRCh38
rs200811470 Chr.15: 74929218-74929218 on Build GRCh38
rs562162336 Chr.15: 74929368-74929368 on Build GRCh38
rs542641042 Chr.15: 74929122-74929122 on Build GRCh38
rs138526218 Chr.15: 74929172-74929172 on Build GRCh38
rs755523610 Chr.15: 74929236-74929236 on Build GRCh38
rs543822451 Chr.15: 74929366-74929366 on Build GRCh38
rs756774888 Chr.15: 74929261-74929261 on Build GRCh38
rs768682367 Chr.15: 74929212-74929212 on Build GRCh38
rs113828200 Chr.15: 74929295-74929295 on Build GRCh38
rs917232481 Chr.15: 74929252-74929252 on Build GRCh38
rs140256516 Chr.15: 74929221-74929221 on Build GRCh38
rs757639315 Chr.15: 74929077-74929078 on Build GRCh38
rs750854500 Chr.15: 74929169-74929169 on Build GRCh38
rs886159166 Chr.15: 74929388-74929388 on Build GRCh38
rs143289611 Chr.15: 74929201-74929201 on Build GRCh38
rs780696817 Chr.15: 74929190-74929190 on Build GRCh38
rs1008605085 Chr.15: 74929371-74929371 on Build GRCh38
rs745528482 Chr.15: 74929265-74929265 on Build GRCh38
rs779754155 Chr.15: 74929276-74929276 on Build GRCh38
rs767844945 Chr.15: 74929150-74929150 on Build GRCh38
rs760914811 Chr.15: 74929232-74929232 on Build GRCh38
rs112721341 Chr.15: 74929189-74929189 on Build GRCh38
rs879870652 Chr.15: 74929473-74929473 on Build GRCh38
rs773308546 Chr.15: 74929231-74929231 on Build GRCh38
rs766352622 Chr.15: 74929077-74929077 on Build GRCh38
rs528877564 Chr.15: 74929340-74929340 on Build GRCh38
rs1803316 Chr.15: 74929195-74929195 on Build GRCh38
rs746494422 Chr.15: 74929184-74929184 on Build GRCh38
rs780396486 Chr.15: 74929170-74929170 on Build GRCh38
rs1056837565 Chr.15: 74929386-74929387 on Build GRCh38
rs912599455 Chr.15: 74929034-74929034 on Build GRCh38
rs991377815 Chr.15: 74929242-74929242 on Build GRCh38
rs201693225 Chr.15: 74929116-74929116 on Build GRCh38
rs900021525 Chr.15: 74929362-74929362 on Build GRCh38
rs759247989 Chr.15: 74929097-74929097 on Build GRCh38
rs900050999 Chr.15: 74929053-74929053 on Build GRCh38
rs150174803 Chr.15: 74929121-74929121 on Build GRCh38
rs546135039 Chr.15: 74929183-74929183 on Build GRCh38
rs766679274 Chr.15: 74929233-74929233 on Build GRCh38
rs986920949 Chr.15: 74929321-74929321 on Build GRCh38
rs755767527 Chr.15: 74929273-74929273 on Build GRCh38
rs1004974032 Chr.15: 74929435-74929435 on Build GRCh38
rs751128223 Chr.15: 74929249-74929249 on Build GRCh38
rs760880460 Chr.15: 74929343-74929343 on Build GRCh38
rs1027414282 Chr.15: 74929442-74929442 on Build GRCh38
rs200382369 Chr.15: 74929196-74929196 on Build GRCh38
rs200848101 Chr.15: 74929088-74929088 on Build GRCh38
rs764291064 Chr.15: 74929132-74929132 on Build GRCh38
rs755698460 Chr.15: 74929350-74929350 on Build GRCh38
rs897540513 Chr.15: 74929377-74929377 on Build GRCh38
rs938834728 Chr.15: 74929382-74929382 on Build GRCh38
rs954375210 Chr.15: 74929099-74929099 on Build GRCh38
rs761019932 Chr.15: 74929215-74929215 on Build GRCh38
rs780408288 Chr.15: 74929070-74929070 on Build GRCh38
rs754212993 Chr.15: 74929235-74929235 on Build GRCh38
rs777229415 Chr.15: 74929078-74929078 on Build GRCh38
rs11553377 Chr.15: 74929185-74929185 on Build GRCh38
rs77302218 Chr.15: 74929373-74929373 on Build GRCh38
rs779206976 Chr.15: 74929084-74929084 on Build GRCh38
rs924753874 Chr.15: 74929339-74929339 on Build GRCh38
rs761900789 Chr.15: 74929336-74929351 on Build GRCh38
rs1016607099 Chr.15: 74929199-74929199 on Build GRCh38
rs1044532641 Chr.15: 74929108-74929108 on Build GRCh38
rs1002612732 Chr.15: 74929123-74929123 on Build GRCh38
rs769828984 Chr.15: 74929111-74929111 on Build GRCh38

View Full Product Details