Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75205919-75206424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614408 c.31G>A p.G11R Chr.15: 75206079-75206079 on Build GRCh38
6614407 c.31G>A p.G11R Chr.15: 75206079-75206079 on Build GRCh38
3932158 c.243A>C p.P81P Chr.15: 75206291-75206291 on Build GRCh38
3816757 c.225C>T p.T75T Chr.15: 75206273-75206273 on Build GRCh38
4517112 c.26_27CC>TT p.T9I Chr.15: 75206074-75206075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763343698 Chr.15: 75206386-75206386 on Build GRCh38
rs755746534 Chr.15: 75206332-75206332 on Build GRCh38
rs767069470 Chr.15: 75206390-75206390 on Build GRCh38
rs1873379 Chr.15: 75206403-75206403 on Build GRCh38
rs544130198 Chr.15: 75206048-75206048 on Build GRCh38
rs567148879 Chr.15: 75206150-75206150 on Build GRCh38
rs143381666 Chr.15: 75206274-75206274 on Build GRCh38
rs766910319 Chr.15: 75206291-75206291 on Build GRCh38
rs374891360 Chr.15: 75206194-75206194 on Build GRCh38
rs200766020 Chr.15: 75206336-75206336 on Build GRCh38
rs758404364 Chr.15: 75206019-75206019 on Build GRCh38
rs368173977 Chr.15: 75206056-75206056 on Build GRCh38
rs759535804 Chr.15: 75206107-75206107 on Build GRCh38
rs775434920 Chr.15: 75206182-75206182 on Build GRCh38
rs761045591 Chr.15: 75206357-75206357 on Build GRCh38
rs139089591 Chr.15: 75205989-75205989 on Build GRCh38
rs143806224 Chr.15: 75206106-75206106 on Build GRCh38
rs930511473 Chr.15: 75205966-75205966 on Build GRCh38
rs769529091 Chr.15: 75206176-75206176 on Build GRCh38
rs753096366 Chr.15: 75206314-75206314 on Build GRCh38
rs573923541 Chr.15: 75206018-75206018 on Build GRCh38
rs1009191425 Chr.15: 75206110-75206110 on Build GRCh38
rs781585751 Chr.15: 75206052-75206052 on Build GRCh38
rs771283251 Chr.15: 75206364-75206364 on Build GRCh38
rs767798047 Chr.15: 75206214-75206214 on Build GRCh38
rs759967363 Chr.15: 75206374-75206374 on Build GRCh38
rs370180250 Chr.15: 75206335-75206335 on Build GRCh38
rs1466663 Chr.15: 75206207-75206207 on Build GRCh38
rs771661306 Chr.15: 75206081-75206081 on Build GRCh38
rs771930954 Chr.15: 75206016-75206016 on Build GRCh38
rs772245072 Chr.15: 75206349-75206349 on Build GRCh38
rs544261988 Chr.15: 75206239-75206239 on Build GRCh38
rs747678168 Chr.15: 75206077-75206077 on Build GRCh38
rs765358715 Chr.15: 75206007-75206007 on Build GRCh38
rs554950133 Chr.15: 75206331-75206331 on Build GRCh38
rs148726096 Chr.15: 75206136-75206136 on Build GRCh38
rs958806766 Chr.15: 75205950-75205950 on Build GRCh38
rs749277296 Chr.15: 75206273-75206273 on Build GRCh38
rs765162473 Chr.15: 75206115-75206115 on Build GRCh38
rs746658909 Chr.15: 75206088-75206088 on Build GRCh38
rs769629452 Chr.15: 75206336-75206370 on Build GRCh38
rs770681140 Chr.15: 75206090-75206090 on Build GRCh38
rs373391409 Chr.15: 75206267-75206267 on Build GRCh38
rs748007536 Chr.15: 75206347-75206347 on Build GRCh38
rs1048857623 Chr.15: 75206337-75206337 on Build GRCh38
rs369809955 Chr.15: 75206206-75206206 on Build GRCh38
rs1049390558 Chr.15: 75205979-75205979 on Build GRCh38
rs914974262 Chr.15: 75206069-75206069 on Build GRCh38
rs768466352 Chr.15: 75206104-75206104 on Build GRCh38
rs778943835 Chr.15: 75206345-75206345 on Build GRCh38
rs971431644 Chr.15: 75206216-75206216 on Build GRCh38
rs756581472 Chr.15: 75206222-75206222 on Build GRCh38
rs991436850 Chr.15: 75205994-75205994 on Build GRCh38
rs780644935 Chr.15: 75206062-75206062 on Build GRCh38
rs150862421 Chr.15: 75206170-75206170 on Build GRCh38
rs114540434 Chr.15: 75206340-75206340 on Build GRCh38
rs772007617 Chr.15: 75206284-75206284 on Build GRCh38
rs377419458 Chr.15: 75206310-75206310 on Build GRCh38
rs773414453 Chr.15: 75206354-75206354 on Build GRCh38
rs765476522 Chr.15: 75206384-75206384 on Build GRCh38
rs550458392 Chr.15: 75206183-75206183 on Build GRCh38
rs373115235 Chr.15: 75206259-75206259 on Build GRCh38
rs777215863 Chr.15: 75206240-75206240 on Build GRCh38
rs757626994 Chr.15: 75206317-75206317 on Build GRCh38
rs777316901 Chr.15: 75206146-75206146 on Build GRCh38
rs764124554 Chr.15: 75206201-75206201 on Build GRCh38
rs773612448 Chr.15: 75206285-75206285 on Build GRCh38
rs751754386 Chr.15: 75206047-75206047 on Build GRCh38
rs770613916 Chr.15: 75206159-75206159 on Build GRCh38
rs943315860 Chr.15: 75206006-75206006 on Build GRCh38
rs374040917 Chr.15: 75206034-75206034 on Build GRCh38
rs116549616 Chr.15: 75206341-75206341 on Build GRCh38
rs1024387817 Chr.15: 75206382-75206382 on Build GRCh38
rs774856280 Chr.15: 75206186-75206186 on Build GRCh38
rs984633259 Chr.15: 75206199-75206199 on Build GRCh38
rs780917995 Chr.15: 75206260-75206260 on Build GRCh38
rs865817368 Chr.15: 75206028-75206028 on Build GRCh38
rs756820384 Chr.15: 75206327-75206327 on Build GRCh38
rs746407054 Chr.15: 75206055-75206055 on Build GRCh38
rs750626591 Chr.15: 75206221-75206221 on Build GRCh38
rs141467125 Chr.15: 75206145-75206145 on Build GRCh38
rs751767197 Chr.15: 75206129-75206129 on Build GRCh38
rs754447200 Chr.15: 75206234-75206234 on Build GRCh38
rs774101723 Chr.15: 75206275-75206275 on Build GRCh38
rs201549769 Chr.15: 75206288-75206288 on Build GRCh38
rs370912089 Chr.15: 75206064-75206064 on Build GRCh38
rs759753350 Chr.15: 75206306-75206306 on Build GRCh38
rs146748144 Chr.15: 75206301-75206301 on Build GRCh38
rs924884759 Chr.15: 75206249-75206249 on Build GRCh38
rs562351386 Chr.15: 75206085-75206085 on Build GRCh38
rs527380769 Chr.15: 75206132-75206132 on Build GRCh38
rs761978762 Chr.15: 75206277-75206277 on Build GRCh38
rs75710563 Chr.15: 75206033-75206033 on Build GRCh38
rs199544667 Chr.15: 75206271-75206271 on Build GRCh38
rs564658480 Chr.15: 75206030-75206030 on Build GRCh38
rs147249276 Chr.15: 75206123-75206123 on Build GRCh38
rs780056068 Chr.15: 75206031-75206031 on Build GRCh38
rs554389286 Chr.15: 75206242-75206242 on Build GRCh38
rs559843501 Chr.15: 75206130-75206130 on Build GRCh38
rs377571506 Chr.15: 75206261-75206261 on Build GRCh38
rs780646657 Chr.15: 75206329-75206329 on Build GRCh38
rs749926416 Chr.15: 75206398-75206398 on Build GRCh38
rs756493518 Chr.15: 75206131-75206131 on Build GRCh38
rs149365490 Chr.15: 75206169-75206169 on Build GRCh38
rs776934401 Chr.15: 75206365-75206365 on Build GRCh38

View Full Product Details