Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75682485-75682983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG4
Entrez Gene ID
1464
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16, NG2
UniGene
Hs.513044
Gene Chromosome Location
Chr.15: 75674322-75712848 on Build GRCh38
Transcripts
NM_001897.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015872 c.4749G>A p.S1583S Chr.15: 75682641-75682641 on Build GRCh38
4675167 c.4570C>T p.R1524W Chr.15: 75682921-75682921 on Build GRCh38
4851582 c.4640C>A p.S1547* Chr.15: 75682851-75682851 on Build GRCh38
1939495 c.4559G>A p.R1520Q Chr.15: 75682932-75682932 on Build GRCh38
6384478 c.4564G>T p.V1522L Chr.15: 75682927-75682927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779970315 Chr.15: 75682747-75682747 on Build GRCh38
rs375631511 Chr.15: 75682780-75682780 on Build GRCh38
rs750888913 Chr.15: 75682829-75682829 on Build GRCh38
rs760606618 Chr.15: 75682633-75682633 on Build GRCh38
rs781623258 Chr.15: 75682585-75682585 on Build GRCh38
rs534570676 Chr.15: 75682604-75682604 on Build GRCh38
rs773124996 Chr.15: 75682623-75682623 on Build GRCh38
rs530150831 Chr.15: 75682925-75682925 on Build GRCh38
rs751157044 Chr.15: 75682908-75682908 on Build GRCh38
rs144616541 Chr.15: 75682649-75682649 on Build GRCh38
rs769641390 Chr.15: 75682868-75682868 on Build GRCh38
rs967347047 Chr.15: 75682914-75682915 on Build GRCh38
rs761112800 Chr.15: 75682832-75682832 on Build GRCh38
rs147158109 Chr.15: 75682866-75682866 on Build GRCh38
rs140387476 Chr.15: 75682920-75682920 on Build GRCh38
rs768039791 Chr.15: 75682752-75682752 on Build GRCh38
rs765598057 Chr.15: 75682904-75682904 on Build GRCh38
rs770877552 Chr.15: 75682701-75682701 on Build GRCh38
rs552981109 Chr.15: 75682642-75682642 on Build GRCh38
rs755373115 Chr.15: 75682852-75682852 on Build GRCh38
rs916673436 Chr.15: 75682827-75682827 on Build GRCh38
rs751712851 Chr.15: 75682737-75682737 on Build GRCh38
rs141642595 Chr.15: 75682720-75682720 on Build GRCh38
rs776471631 Chr.15: 75682704-75682704 on Build GRCh38
rs766677030 Chr.15: 75682835-75682835 on Build GRCh38
rs763414673 Chr.15: 75682936-75682936 on Build GRCh38
rs761958703 Chr.15: 75682887-75682887 on Build GRCh38
rs976339783 Chr.15: 75682590-75682590 on Build GRCh38
rs778858827 Chr.15: 75682918-75682918 on Build GRCh38
rs777396298 Chr.15: 75682854-75682854 on Build GRCh38
rs756732390 Chr.15: 75682862-75682862 on Build GRCh38
rs368470206 Chr.15: 75682880-75682880 on Build GRCh38
rs757332982 Chr.15: 75682750-75682750 on Build GRCh38
rs769564688 Chr.15: 75682794-75682794 on Build GRCh38
rs773183480 Chr.15: 75682891-75682891 on Build GRCh38
rs374291734 Chr.15: 75682867-75682867 on Build GRCh38
rs780300019 Chr.15: 75682765-75682765 on Build GRCh38
rs750961628 Chr.15: 75682762-75682762 on Build GRCh38
rs144338087 Chr.15: 75682933-75682933 on Build GRCh38
rs781174070 Chr.15: 75682778-75682778 on Build GRCh38
rs751227189 Chr.15: 75682856-75682856 on Build GRCh38
rs772736007 Chr.15: 75682698-75682698 on Build GRCh38
rs367996596 Chr.15: 75682727-75682727 on Build GRCh38
rs771054582 Chr.15: 75682931-75682931 on Build GRCh38
rs771500047 Chr.15: 75682695-75682695 on Build GRCh38
rs756491921 Chr.15: 75682763-75682763 on Build GRCh38
rs753767013 Chr.15: 75682952-75682952 on Build GRCh38
rs371831713 Chr.15: 75682913-75682913 on Build GRCh38
rs200471186 Chr.15: 75682560-75682560 on Build GRCh38
rs747859506 Chr.15: 75682686-75682686 on Build GRCh38
rs775740360 Chr.15: 75682934-75682934 on Build GRCh38
rs768379105 Chr.15: 75682596-75682596 on Build GRCh38
rs778627105 Chr.15: 75682685-75682685 on Build GRCh38
rs994380451 Chr.15: 75682713-75682713 on Build GRCh38
rs545602898 Chr.15: 75682826-75682826 on Build GRCh38
rs749298439 Chr.15: 75682916-75682916 on Build GRCh38
rs774757053 Chr.15: 75682819-75682819 on Build GRCh38
rs763218968 Chr.15: 75682799-75682799 on Build GRCh38
rs149321023 Chr.15: 75682622-75682622 on Build GRCh38
rs772158757 Chr.15: 75682921-75682921 on Build GRCh38
rs760040347 Chr.15: 75682932-75682932 on Build GRCh38
rs775456912 Chr.15: 75682798-75682798 on Build GRCh38
rs770045405 Chr.15: 75682915-75682915 on Build GRCh38
rs754175918 Chr.15: 75682842-75682842 on Build GRCh38
rs768978972 Chr.15: 75682802-75682802 on Build GRCh38
rs373172676 Chr.15: 75682587-75682587 on Build GRCh38
rs754765879 Chr.15: 75682917-75682917 on Build GRCh38
rs774704296 Chr.15: 75682886-75682886 on Build GRCh38
rs763273863 Chr.15: 75682721-75682721 on Build GRCh38
rs994020897 Chr.15: 75682514-75682514 on Build GRCh38
rs756478440 Chr.15: 75682683-75682683 on Build GRCh38
rs781364010 Chr.15: 75682677-75682677 on Build GRCh38
rs1048701917 Chr.15: 75682873-75682873 on Build GRCh38
rs763302287 Chr.15: 75682751-75682751 on Build GRCh38
rs540165747 Chr.15: 75682911-75682911 on Build GRCh38
rs747628906 Chr.15: 75682617-75682617 on Build GRCh38
rs752302356 Chr.15: 75682768-75682768 on Build GRCh38
rs759879058 Chr.15: 75682900-75682900 on Build GRCh38
rs760853749 Chr.15: 75682896-75682896 on Build GRCh38
rs759296054 Chr.15: 75682644-75682644 on Build GRCh38
rs765988638 Chr.15: 75682951-75682951 on Build GRCh38
rs764282652 Chr.15: 75682666-75682666 on Build GRCh38
rs746897054 Chr.15: 75682700-75682700 on Build GRCh38
rs371145581 Chr.15: 75682774-75682774 on Build GRCh38
rs199703779 Chr.15: 75682714-75682714 on Build GRCh38
rs370078244 Chr.15: 75682673-75682673 on Build GRCh38
rs748250578 Chr.15: 75682750-75682751 on Build GRCh38
rs757639619 Chr.15: 75682561-75682561 on Build GRCh38
rs945497898 Chr.15: 75682616-75682619 on Build GRCh38
rs1006687933 Chr.15: 75682882-75682882 on Build GRCh38
rs564006260 Chr.15: 75682870-75682870 on Build GRCh38
rs201339927 Chr.15: 75682915-75682915 on Build GRCh38
rs572824250 Chr.15: 75682871-75682871 on Build GRCh38
rs936735709 Chr.15: 75682820-75682820 on Build GRCh38
rs758064667 Chr.15: 75682769-75682769 on Build GRCh38
rs574407281 Chr.15: 75682658-75682658 on Build GRCh38
rs112647859 Chr.15: 75682789-75682789 on Build GRCh38
rs750146416 Chr.15: 75682940-75682940 on Build GRCh38
rs552860515 Chr.15: 75682543-75682543 on Build GRCh38
rs754406835 Chr.15: 75682897-75682897 on Build GRCh38
rs765465521 Chr.15: 75682647-75682647 on Build GRCh38
rs148389768 Chr.15: 75682679-75682679 on Build GRCh38
rs746752555 Chr.15: 75682910-75682910 on Build GRCh38
rs747052869 Chr.15: 75682930-75682930 on Build GRCh38
rs557064196 Chr.15: 75682715-75682715 on Build GRCh38
rs755678669 Chr.15: 75682944-75682944 on Build GRCh38
rs553544927 Chr.15: 75682937-75682937 on Build GRCh38
rs754983756 Chr.15: 75682958-75682958 on Build GRCh38
rs752045565 Chr.15: 75682672-75682672 on Build GRCh38
rs761950301 Chr.15: 75682823-75682823 on Build GRCh38
rs375780848 Chr.15: 75682641-75682641 on Build GRCh38
rs745806406 Chr.15: 75682779-75682779 on Build GRCh38
rs1046409826 Chr.15: 75682863-75682863 on Build GRCh38

View Full Product Details