Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75930169-75930682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO22
Entrez Gene ID
26263
Gene Name
F-box protein 22
Gene Aliases
FBX22, FISTC1
UniGene
Hs.591115
Gene Chromosome Location
Chr.15: 75903859-75937513 on Build GRCh38
Transcripts
NM_012170.3 NM_147188.2 NR_037623.1 XM_011521447.2

SNP Detail

SNP ID Chromosome Location
rs771692781 Chr.15: 75930510-75930511 on Build GRCh38
rs61087430 Chr.15: 75930443-75930443 on Build GRCh38
rs572391924 Chr.15: 75930346-75930346 on Build GRCh38
rs368885606 Chr.15: 75930499-75930499 on Build GRCh38
rs755816549 Chr.15: 75930222-75930222 on Build GRCh38
rs1030136934 Chr.15: 75930347-75930347 on Build GRCh38
rs926241240 Chr.15: 75930538-75930538 on Build GRCh38
rs561408224 Chr.15: 75930460-75930460 on Build GRCh38
rs941551498 Chr.15: 75930493-75930496 on Build GRCh38
rs763614594 Chr.15: 75930321-75930321 on Build GRCh38
rs1017990426 Chr.15: 75930252-75930252 on Build GRCh38
rs910055908 Chr.15: 75930475-75930475 on Build GRCh38
rs1001139690 Chr.15: 75930223-75930223 on Build GRCh38
rs941246306 Chr.15: 75930531-75930531 on Build GRCh38
rs139127277 Chr.15: 75930657-75930657 on Build GRCh38
rs918868629 Chr.15: 75930498-75930498 on Build GRCh38
rs964711522 Chr.15: 75930302-75930302 on Build GRCh38
rs937716459 Chr.15: 75930567-75930567 on Build GRCh38
rs185986999 Chr.15: 75930245-75930245 on Build GRCh38
rs976162993 Chr.15: 75930312-75930312 on Build GRCh38
rs542783871 Chr.15: 75930381-75930381 on Build GRCh38
rs954169648 Chr.15: 75930363-75930363 on Build GRCh38
rs3833815 Chr.15: 75930265-75930265 on Build GRCh38
rs974024207 Chr.15: 75930537-75930537 on Build GRCh38
rs948504620 Chr.15: 75930639-75930639 on Build GRCh38
rs335680 Chr.15: 75930494-75930494 on Build GRCh38

View Full Product Details