Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78178222-78178734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSBG1
Entrez Gene ID
23205
Gene Name
acyl-CoA synthetase bubblegum family member 1
Gene Aliases
BG, BG1, BGM, GR-LACS, LPD
UniGene
Hs.655760
Gene Chromosome Location
Chr.15: 78167468-78234707 on Build GRCh38
Transcripts
NM_001199377.1 NM_015162.4 XM_011521390.2 XM_011521391.2 XM_017022025.1 XM_017022026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555115 c.1693C>A p.R565S Chr.15: 78178623-78178623 on Build GRCh38
1742685 c.1615G>T p.A539S Chr.15: 78178701-78178701 on Build GRCh38
118281 c.1620C>T p.I540I Chr.15: 78178696-78178696 on Build GRCh38
4652643 c.1664A>G p.D555G Chr.15: 78178652-78178652 on Build GRCh38
6025631 c.1595T>C p.M532T Chr.15: 78178721-78178721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987612734 Chr.15: 78178615-78178615 on Build GRCh38
rs780783529 Chr.15: 78178675-78178675 on Build GRCh38
rs543911373 Chr.15: 78178672-78178672 on Build GRCh38
rs974813729 Chr.15: 78178370-78178370 on Build GRCh38
rs535512069 Chr.15: 78178443-78178443 on Build GRCh38
rs919912842 Chr.15: 78178408-78178408 on Build GRCh38
rs761399514 Chr.15: 78178577-78178577 on Build GRCh38
rs367875558 Chr.15: 78178632-78178632 on Build GRCh38
rs34921279 Chr.15: 78178692-78178692 on Build GRCh38
rs542486338 Chr.15: 78178282-78178282 on Build GRCh38
rs886449162 Chr.15: 78178392-78178392 on Build GRCh38
rs1031963188 Chr.15: 78178288-78178291 on Build GRCh38
rs1032625969 Chr.15: 78178508-78178508 on Build GRCh38
rs755698104 Chr.15: 78178682-78178682 on Build GRCh38
rs774297634 Chr.15: 78178695-78178695 on Build GRCh38
rs377622035 Chr.15: 78178651-78178651 on Build GRCh38
rs768536234 Chr.15: 78178614-78178614 on Build GRCh38
rs974885778 Chr.15: 78178562-78178562 on Build GRCh38
rs762182242 Chr.15: 78178643-78178643 on Build GRCh38
rs767872050 Chr.15: 78178471-78178471 on Build GRCh38
rs1030673316 Chr.15: 78178527-78178527 on Build GRCh38
rs546980832 Chr.15: 78178403-78178403 on Build GRCh38
rs754413183 Chr.15: 78178658-78178658 on Build GRCh38
rs752077876 Chr.15: 78178393-78178393 on Build GRCh38
rs754604846 Chr.15: 78178652-78178652 on Build GRCh38
rs563873043 Chr.15: 78178286-78178286 on Build GRCh38
rs936447156 Chr.15: 78178705-78178705 on Build GRCh38
rs774009900 Chr.15: 78178617-78178617 on Build GRCh38
rs958291107 Chr.15: 78178549-78178549 on Build GRCh38
rs964152708 Chr.15: 78178624-78178624 on Build GRCh38
rs369035787 Chr.15: 78178591-78178591 on Build GRCh38
rs956653492 Chr.15: 78178295-78178295 on Build GRCh38
rs200911228 Chr.15: 78178608-78178608 on Build GRCh38
rs113950211 Chr.15: 78178297-78178297 on Build GRCh38
rs1009161234 Chr.15: 78178486-78178486 on Build GRCh38
rs779399847 Chr.15: 78178688-78178688 on Build GRCh38
rs773426891 Chr.15: 78178648-78178648 on Build GRCh38
rs1045228393 Chr.15: 78178254-78178255 on Build GRCh38
rs1030186411 Chr.15: 78178352-78178352 on Build GRCh38
rs756909612 Chr.15: 78178673-78178673 on Build GRCh38
rs918166501 Chr.15: 78178666-78178666 on Build GRCh38
rs569097682 Chr.15: 78178472-78178472 on Build GRCh38
rs992881905 Chr.15: 78178276-78178276 on Build GRCh38
rs772450975 Chr.15: 78178706-78178706 on Build GRCh38
rs368163981 Chr.15: 78178351-78178351 on Build GRCh38
rs1014009805 Chr.15: 78178467-78178467 on Build GRCh38
rs967112400 Chr.15: 78178477-78178477 on Build GRCh38
rs775449753 Chr.15: 78178606-78178606 on Build GRCh38
rs539207930 Chr.15: 78178499-78178499 on Build GRCh38
rs374210790 Chr.15: 78178598-78178598 on Build GRCh38
rs141551096 Chr.15: 78178650-78178650 on Build GRCh38
rs1037605472 Chr.15: 78178423-78178423 on Build GRCh38
rs1027048683 Chr.15: 78178649-78178649 on Build GRCh38
rs936848561 Chr.15: 78178360-78178360 on Build GRCh38
rs770560069 Chr.15: 78178586-78178586 on Build GRCh38
rs757750719 Chr.15: 78178409-78178409 on Build GRCh38
rs745318303 Chr.15: 78178592-78178592 on Build GRCh38
rs1034162451 Chr.15: 78178452-78178452 on Build GRCh38
rs918146175 Chr.15: 78178446-78178446 on Build GRCh38
rs531123757 Chr.15: 78178287-78178287 on Build GRCh38
rs779121025 Chr.15: 78178565-78178565 on Build GRCh38
rs767787287 Chr.15: 78178645-78178645 on Build GRCh38
rs572576525 Chr.15: 78178561-78178561 on Build GRCh38
rs190337245 Chr.15: 78178693-78178693 on Build GRCh38
rs907848394 Chr.15: 78178623-78178623 on Build GRCh38
rs552811286 Chr.15: 78178299-78178299 on Build GRCh38
rs557533394 Chr.15: 78178541-78178541 on Build GRCh38
rs1037677742 Chr.15: 78178685-78178685 on Build GRCh38
rs978370772 Chr.15: 78178343-78178343 on Build GRCh38
rs568429149 Chr.15: 78178436-78178436 on Build GRCh38
rs747882724 Chr.15: 78178696-78178696 on Build GRCh38
rs570909866 Chr.15: 78178335-78178335 on Build GRCh38
rs933439788 Chr.15: 78178390-78178390 on Build GRCh38
rs898990135 Chr.15: 78178427-78178427 on Build GRCh38
rs921982747 Chr.15: 78178382-78178382 on Build GRCh38
rs892111385 Chr.15: 78178480-78178480 on Build GRCh38
rs370057969 Chr.15: 78178659-78178659 on Build GRCh38
rs528617009 Chr.15: 78178391-78178391 on Build GRCh38
rs560683383 Chr.15: 78178455-78178455 on Build GRCh38

View Full Product Details