Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78180548-78181046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSBG1
Entrez Gene ID
23205
Gene Name
acyl-CoA synthetase bubblegum family member 1
Gene Aliases
BG, BG1, BGM, GR-LACS, LPD
UniGene
Hs.655760
Gene Chromosome Location
Chr.15: 78167468-78234707 on Build GRCh38
Transcripts
NM_001199377.1 NM_015162.4 XM_011521390.2 XM_011521391.2 XM_017022025.1 XM_017022026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792074 c.1249G>A p.G417S Chr.15: 78180759-78180759 on Build GRCh38
5383872 c.1143G>A p.K381K Chr.15: 78180865-78180865 on Build GRCh38
5817978 c.1125G>T p.V375V Chr.15: 78180883-78180883 on Build GRCh38
7280727 c.1088C>T p.T363M Chr.15: 78180920-78180920 on Build GRCh38
4658966 c.1154G>A p.R385H Chr.15: 78180854-78180854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981984025 Chr.15: 78180688-78180688 on Build GRCh38
rs1008456534 Chr.15: 78180589-78180589 on Build GRCh38
rs761668317 Chr.15: 78180709-78180709 on Build GRCh38
rs946793111 Chr.15: 78180635-78180635 on Build GRCh38
rs17479135 Chr.15: 78180676-78180676 on Build GRCh38
rs778578080 Chr.15: 78180836-78180836 on Build GRCh38
rs193007252 Chr.15: 78180671-78180671 on Build GRCh38
rs988989799 Chr.15: 78180971-78180971 on Build GRCh38
rs889721725 Chr.15: 78180588-78180588 on Build GRCh38
rs368902007 Chr.15: 78180829-78180829 on Build GRCh38
rs34638717 Chr.15: 78180690-78180691 on Build GRCh38
rs746175029 Chr.15: 78180825-78180825 on Build GRCh38
rs776682887 Chr.15: 78180987-78180987 on Build GRCh38
rs763081605 Chr.15: 78180892-78180892 on Build GRCh38
rs922217372 Chr.15: 78180898-78180898 on Build GRCh38
rs752746904 Chr.15: 78180803-78180803 on Build GRCh38
rs915367641 Chr.15: 78180621-78180621 on Build GRCh38
rs372203439 Chr.15: 78180838-78180838 on Build GRCh38
rs1033969390 Chr.15: 78181027-78181027 on Build GRCh38
rs770790995 Chr.15: 78180876-78180876 on Build GRCh38
rs1026719466 Chr.15: 78180843-78180843 on Build GRCh38
rs373127133 Chr.15: 78180735-78180735 on Build GRCh38
rs757842128 Chr.15: 78180918-78180918 on Build GRCh38
rs746484386 Chr.15: 78180979-78180979 on Build GRCh38
rs958613594 Chr.15: 78180786-78180786 on Build GRCh38
rs147846122 Chr.15: 78180919-78180919 on Build GRCh38
rs1002993138 Chr.15: 78181017-78181017 on Build GRCh38
rs765463008 Chr.15: 78180802-78180802 on Build GRCh38
rs202151408 Chr.15: 78180740-78180740 on Build GRCh38
rs889829083 Chr.15: 78180984-78180984 on Build GRCh38
rs35728145 Chr.15: 78180992-78180992 on Build GRCh38
rs928767641 Chr.15: 78180959-78180959 on Build GRCh38
rs776444526 Chr.15: 78180877-78180877 on Build GRCh38
rs952477396 Chr.15: 78180857-78180857 on Build GRCh38
rs993861297 Chr.15: 78180823-78180823 on Build GRCh38
rs544929691 Chr.15: 78180661-78180661 on Build GRCh38
rs143166464 Chr.15: 78180914-78180914 on Build GRCh38
rs780246786 Chr.15: 78180931-78180931 on Build GRCh38
rs149090840 Chr.15: 78180854-78180854 on Build GRCh38
rs755851172 Chr.15: 78180721-78180721 on Build GRCh38
rs868114336 Chr.15: 78180990-78180990 on Build GRCh38
rs751633265 Chr.15: 78180915-78180915 on Build GRCh38
rs374067302 Chr.15: 78180983-78180983 on Build GRCh38
rs149309741 Chr.15: 78180972-78180972 on Build GRCh38
rs865836208 Chr.15: 78180665-78180665 on Build GRCh38
rs527718769 Chr.15: 78180946-78180946 on Build GRCh38
rs990852272 Chr.15: 78180888-78180888 on Build GRCh38
rs749007684 Chr.15: 78180751-78180751 on Build GRCh38
rs1005025249 Chr.15: 78180985-78180985 on Build GRCh38
rs1040334444 Chr.15: 78180881-78180881 on Build GRCh38
rs17850482 Chr.15: 78180814-78180814 on Build GRCh38
rs553809423 Chr.15: 78180603-78180603 on Build GRCh38
rs544973852 Chr.15: 78180749-78180749 on Build GRCh38
rs773566057 Chr.15: 78180815-78180815 on Build GRCh38
rs754096251 Chr.15: 78180723-78180723 on Build GRCh38
rs771382400 Chr.15: 78180706-78180709 on Build GRCh38
rs764922969 Chr.15: 78180885-78180885 on Build GRCh38
rs561284398 Chr.15: 78181006-78181006 on Build GRCh38
rs753613833 Chr.15: 78180791-78180791 on Build GRCh38
rs1019449220 Chr.15: 78180619-78180619 on Build GRCh38
rs201092202 Chr.15: 78180818-78180818 on Build GRCh38
rs756551163 Chr.15: 78180920-78180920 on Build GRCh38
rs200646960 Chr.15: 78180759-78180759 on Build GRCh38
rs764329696 Chr.15: 78180912-78180912 on Build GRCh38
rs958540235 Chr.15: 78180587-78180587 on Build GRCh38
rs374008679 Chr.15: 78180739-78180739 on Build GRCh38
rs1007997209 Chr.15: 78180600-78180600 on Build GRCh38
rs371236331 Chr.15: 78180809-78180809 on Build GRCh38
rs567144170 Chr.15: 78180649-78180649 on Build GRCh38
rs202065533 Chr.15: 78180777-78180777 on Build GRCh38
rs201408176 Chr.15: 78180719-78180719 on Build GRCh38
rs12915634 Chr.15: 78180967-78180967 on Build GRCh38
rs771462805 Chr.15: 78180745-78180745 on Build GRCh38
rs759686118 Chr.15: 78180794-78180794 on Build GRCh38
rs779137895 Chr.15: 78180981-78180981 on Build GRCh38
rs961079691 Chr.15: 78180629-78180629 on Build GRCh38
rs796851895 Chr.15: 78180680-78180680 on Build GRCh38
rs777970413 Chr.15: 78180811-78180811 on Build GRCh38
rs760571690 Chr.15: 78180783-78180783 on Build GRCh38
rs200102425 Chr.15: 78180736-78180736 on Build GRCh38
rs1053215613 Chr.15: 78180731-78180731 on Build GRCh38
rs771925659 Chr.15: 78180779-78180779 on Build GRCh38
rs574146982 Chr.15: 78180675-78180675 on Build GRCh38
rs753004934 Chr.15: 78180889-78180889 on Build GRCh38
rs761346938 Chr.15: 78180769-78180769 on Build GRCh38
rs746422518 Chr.15: 78180984-78180985 on Build GRCh38
rs781699204 Chr.15: 78180819-78180819 on Build GRCh38
rs906912591 Chr.15: 78180995-78180995 on Build GRCh38
rs372198914 Chr.15: 78180977-78180977 on Build GRCh38
rs772265446 Chr.15: 78180790-78180790 on Build GRCh38
rs565232292 Chr.15: 78180662-78180662 on Build GRCh38
rs755221046 Chr.15: 78180725-78180725 on Build GRCh38
rs773736059 Chr.15: 78180760-78180760 on Build GRCh38
rs760659983 Chr.15: 78180855-78180855 on Build GRCh38

View Full Product Details