Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78340193-78340690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRABP1
Entrez Gene ID
1381
Gene Name
cellular retinoic acid binding protein 1
Gene Aliases
CRABP, CRABP-I, CRABPI, RBP5
UniGene
Hs.346950
Gene Chromosome Location
Chr.15: 78340324-78348230 on Build GRCh38
Transcripts
NM_004378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3816825 c.70+1G>C p.? Chr.15: 78340499-78340499 on Build GRCh38
434365 c.60C>G p.L20L Chr.15: 78340488-78340488 on Build GRCh38
1249092 c.57G>A p.L19L Chr.15: 78340485-78340485 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs555983792 Chr.15: 78340655-78340655 on Build GRCh38
rs918713287 Chr.15: 78340661-78340661 on Build GRCh38
rs143159972 Chr.15: 78340440-78340440 on Build GRCh38
rs929982116 Chr.15: 78340457-78340457 on Build GRCh38
rs746123318 Chr.15: 78340398-78340398 on Build GRCh38
rs765928145 Chr.15: 78340528-78340528 on Build GRCh38
rs964541922 Chr.15: 78340654-78340654 on Build GRCh38
rs770387235 Chr.15: 78340497-78340497 on Build GRCh38
rs373513681 Chr.15: 78340244-78340244 on Build GRCh38
rs772284141 Chr.15: 78340403-78340404 on Build GRCh38
rs753446807 Chr.15: 78340542-78340542 on Build GRCh38
rs1044927788 Chr.15: 78340516-78340516 on Build GRCh38
rs754442605 Chr.15: 78340543-78340543 on Build GRCh38
rs766416239 Chr.15: 78340404-78340404 on Build GRCh38
rs1009267458 Chr.15: 78340354-78340354 on Build GRCh38
rs1030731933 Chr.15: 78340222-78340222 on Build GRCh38
rs1018421210 Chr.15: 78340642-78340642 on Build GRCh38
rs949473733 Chr.15: 78340458-78340458 on Build GRCh38
rs775744719 Chr.15: 78340408-78340410 on Build GRCh38
rs1047117322 Chr.15: 78340269-78340269 on Build GRCh38
rs896603653 Chr.15: 78340286-78340286 on Build GRCh38
rs936575837 Chr.15: 78340507-78340507 on Build GRCh38
rs986929269 Chr.15: 78340377-78340377 on Build GRCh38
rs765642594 Chr.15: 78340412-78340412 on Build GRCh38
rs774249656 Chr.15: 78340381-78340381 on Build GRCh38
rs772346461 Chr.15: 78340387-78340387 on Build GRCh38
rs909835822 Chr.15: 78340491-78340491 on Build GRCh38
rs954485118 Chr.15: 78340366-78340366 on Build GRCh38
rs368691944 Chr.15: 78340520-78340520 on Build GRCh38
rs778704148 Chr.15: 78340459-78340459 on Build GRCh38
rs750025429 Chr.15: 78340420-78340420 on Build GRCh38
rs1003274909 Chr.15: 78340218-78340218 on Build GRCh38
rs982780008 Chr.15: 78340423-78340423 on Build GRCh38
rs768355161 Chr.15: 78340379-78340379 on Build GRCh38
rs767341454 Chr.15: 78340417-78340417 on Build GRCh38
rs760041116 Chr.15: 78340411-78340411 on Build GRCh38
rs974068348 Chr.15: 78340234-78340234 on Build GRCh38
rs758833020 Chr.15: 78340415-78340415 on Build GRCh38
rs553526777 Chr.15: 78340324-78340324 on Build GRCh38
rs1009236268 Chr.15: 78340601-78340601 on Build GRCh38
rs759510356 Chr.15: 78340496-78340496 on Build GRCh38
rs544940375 Chr.15: 78340644-78340644 on Build GRCh38
rs376583344 Chr.15: 78340421-78340421 on Build GRCh38
rs780521599 Chr.15: 78340648-78340648 on Build GRCh38
rs922948391 Chr.15: 78340300-78340300 on Build GRCh38
rs1007572455 Chr.15: 78340626-78340626 on Build GRCh38
rs942949706 Chr.15: 78340230-78340230 on Build GRCh38
rs1030080733 Chr.15: 78340362-78340362 on Build GRCh38
rs1010615021 Chr.15: 78340579-78340579 on Build GRCh38
rs567147658 Chr.15: 78340338-78340338 on Build GRCh38
rs969025956 Chr.15: 78340607-78340607 on Build GRCh38
rs540305387 Chr.15: 78340587-78340587 on Build GRCh38
rs974748977 Chr.15: 78340406-78340406 on Build GRCh38
rs776133213 Chr.15: 78340505-78340505 on Build GRCh38
rs1127472 Chr.15: 78340488-78340488 on Build GRCh38
rs184858686 Chr.15: 78340656-78340656 on Build GRCh38
rs556959596 Chr.15: 78340353-78340353 on Build GRCh38
rs949213086 Chr.15: 78340301-78340301 on Build GRCh38
rs777037680 Chr.15: 78340405-78340405 on Build GRCh38
rs760757894 Chr.15: 78340403-78340403 on Build GRCh38
rs930053485 Chr.15: 78340241-78340241 on Build GRCh38
rs532784055 Chr.15: 78340511-78340511 on Build GRCh38
rs1021880145 Chr.15: 78340238-78340238 on Build GRCh38
rs1018499732 Chr.15: 78340638-78340638 on Build GRCh38
rs773409715 Chr.15: 78340389-78340389 on Build GRCh38
rs764507825 Chr.15: 78340519-78340519 on Build GRCh38
rs777983851 Chr.15: 78340489-78340489 on Build GRCh38
rs754994289 Chr.15: 78340455-78340455 on Build GRCh38
rs761032471 Chr.15: 78340414-78340415 on Build GRCh38
rs748718781 Chr.15: 78340443-78340443 on Build GRCh38
rs770944935 Chr.15: 78340495-78340495 on Build GRCh38
rs1003347022 Chr.15: 78340537-78340537 on Build GRCh38
rs1018867708 Chr.15: 78340399-78340399 on Build GRCh38
rs760418980 Chr.15: 78340522-78340522 on Build GRCh38
rs373846623 Chr.15: 78340527-78340527 on Build GRCh38
rs747184677 Chr.15: 78340494-78340494 on Build GRCh38
rs558021365 Chr.15: 78340425-78340425 on Build GRCh38
rs892180707 Chr.15: 78340570-78340570 on Build GRCh38
rs534754590 Chr.15: 78340237-78340237 on Build GRCh38
rs947953771 Chr.15: 78340500-78340500 on Build GRCh38
rs369671248 Chr.15: 78340606-78340606 on Build GRCh38
rs956370592 Chr.15: 78340224-78340224 on Build GRCh38
rs575521750 Chr.15: 78340373-78340373 on Build GRCh38
rs57304310 Chr.15: 78340296-78340296 on Build GRCh38
rs370214828 Chr.15: 78340413-78340413 on Build GRCh38
rs570603070 Chr.15: 78340216-78340216 on Build GRCh38
rs367904156 Chr.15: 78340582-78340582 on Build GRCh38
rs76956384 Chr.15: 78340337-78340337 on Build GRCh38
rs747906092 Chr.15: 78340382-78340382 on Build GRCh38
rs1007330546 Chr.15: 78340620-78340620 on Build GRCh38
rs778561914 Chr.15: 78340530-78340530 on Build GRCh38
rs748004759 Chr.15: 78340482-78340482 on Build GRCh38
rs1052189064 Chr.15: 78340561-78340561 on Build GRCh38
rs751464363 Chr.15: 78340339-78340339 on Build GRCh38

View Full Product Details