Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78594354-78594879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA3
Entrez Gene ID
1136
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
LNCR2, NACHRA3, PAOD2
UniGene
Hs.89605
Gene Chromosome Location
Chr.15: 78593052-78621295 on Build GRCh38
Transcripts
NM_001166694.1 NR_046313.1 NM_000745.3 NM_001307945.1

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_001166694.1 NR_046313.1 NM_000745.3 NM_001307945.1

SNP Detail

SNP ID Chromosome Location
rs146832909 Chr.15: 78594495-78594495 on Build GRCh38
rs56255741 Chr.15: 78594550-78594550 on Build GRCh38
rs771778152 Chr.15: 78594492-78594492 on Build GRCh38
rs144907889 Chr.15: 78594576-78594576 on Build GRCh38
rs143101371 Chr.15: 78594657-78594657 on Build GRCh38
rs1048558885 Chr.15: 78594679-78594679 on Build GRCh38
rs186039949 Chr.15: 78594806-78594806 on Build GRCh38
rs867081958 Chr.15: 78594505-78594505 on Build GRCh38
rs151172211 Chr.15: 78594818-78594818 on Build GRCh38
rs940345957 Chr.15: 78594387-78594387 on Build GRCh38
rs748360064 Chr.15: 78594635-78594635 on Build GRCh38
rs76678365 Chr.15: 78594551-78594551 on Build GRCh38
rs900023271 Chr.15: 78594455-78594455 on Build GRCh38
rs370931048 Chr.15: 78594398-78594398 on Build GRCh38
rs887240276 Chr.15: 78594699-78594699 on Build GRCh38
rs947246574 Chr.15: 78594754-78594754 on Build GRCh38
rs908797269 Chr.15: 78594592-78594592 on Build GRCh38
rs778870924 Chr.15: 78594631-78594631 on Build GRCh38
rs998815470 Chr.15: 78594477-78594477 on Build GRCh38
rs933141995 Chr.15: 78594399-78594399 on Build GRCh38
rs573649543 Chr.15: 78594503-78594503 on Build GRCh38
rs971738411 Chr.15: 78594608-78594608 on Build GRCh38
rs1043028329 Chr.15: 78594474-78594474 on Build GRCh38
rs1011945599 Chr.15: 78594460-78594460 on Build GRCh38
rs773284952 Chr.15: 78594669-78594669 on Build GRCh38
rs7183983 Chr.15: 78594560-78594560 on Build GRCh38
rs926696481 Chr.15: 78594532-78594532 on Build GRCh38
rs989938518 Chr.15: 78594835-78594835 on Build GRCh38
rs374255078 Chr.15: 78594475-78594475 on Build GRCh38
rs577425311 Chr.15: 78594715-78594715 on Build GRCh38
rs1051444370 Chr.15: 78594387-78594388 on Build GRCh38
rs4887067 Chr.15: 78594605-78594605 on Build GRCh38
rs1050716580 Chr.15: 78594405-78594405 on Build GRCh38
rs539579411 Chr.15: 78594765-78594765 on Build GRCh38
rs55989223 Chr.15: 78594506-78594506 on Build GRCh38
rs1033619194 Chr.15: 78594815-78594815 on Build GRCh38
rs778034799 Chr.15: 78594655-78594657 on Build GRCh38
rs756191800 Chr.15: 78594571-78594572 on Build GRCh38
rs945322580 Chr.15: 78594570-78594570 on Build GRCh38
rs1027538137 Chr.15: 78594473-78594473 on Build GRCh38
rs980846706 Chr.15: 78594509-78594509 on Build GRCh38
rs747116248 Chr.15: 78594696-78594696 on Build GRCh38
rs889018748 Chr.15: 78594448-78594448 on Build GRCh38
rs757566487 Chr.15: 78594730-78594731 on Build GRCh38
rs933090962 Chr.15: 78594627-78594627 on Build GRCh38
rs923004974 Chr.15: 78594620-78594620 on Build GRCh38
rs566370605 Chr.15: 78594397-78594397 on Build GRCh38
rs149128996 Chr.15: 78594528-78594528 on Build GRCh38
rs537201115 Chr.15: 78594561-78594561 on Build GRCh38
rs7163480 Chr.15: 78594829-78594829 on Build GRCh38

View Full Product Details