Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78934787-78935283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSH
Entrez Gene ID
1512
Gene Name
cathepsin H
Gene Aliases
ACC-4, ACC-5, ACC4, ACC5, CPSB
UniGene
Hs.148641
Gene Chromosome Location
Chr.15: 78921750-78945098 on Build GRCh38
Transcripts
NM_001319137.1 NM_004390.4 XM_005254181.2 XM_011521276.1 XM_017021951.1 XM_017021952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4057181 c.358G>A p.V120M Chr.15: 78935025-78935025 on Build GRCh38
5603310 c.349C>T p.P117S Chr.15: 78935034-78935034 on Build GRCh38
5383888 c.391C>T p.P131S Chr.15: 78934992-78934992 on Build GRCh38
4566336 c.352_353CC>TT p.P118F Chr.15: 78935030-78935031 on Build GRCh38
2011397 c.337A>G p.T113A Chr.15: 78935046-78935046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915395345 Chr.15: 78935201-78935201 on Build GRCh38
rs756842363 Chr.15: 78935012-78935012 on Build GRCh38
rs762133290 Chr.15: 78935050-78935050 on Build GRCh38
rs145029663 Chr.15: 78935075-78935075 on Build GRCh38
rs779639457 Chr.15: 78935158-78935158 on Build GRCh38
rs565309151 Chr.15: 78935030-78935030 on Build GRCh38
rs766864044 Chr.15: 78934965-78934965 on Build GRCh38
rs373219279 Chr.15: 78935145-78935147 on Build GRCh38
rs375372809 Chr.15: 78934941-78934941 on Build GRCh38
rs548206226 Chr.15: 78935052-78935052 on Build GRCh38
rs750176850 Chr.15: 78934964-78934965 on Build GRCh38
rs918809456 Chr.15: 78935088-78935088 on Build GRCh38
rs748954987 Chr.15: 78935181-78935181 on Build GRCh38
rs968222892 Chr.15: 78935205-78935205 on Build GRCh38
rs781162371 Chr.15: 78934932-78934932 on Build GRCh38
rs746559833 Chr.15: 78934906-78934906 on Build GRCh38
rs556869954 Chr.15: 78935111-78935111 on Build GRCh38
rs750453161 Chr.15: 78934954-78934954 on Build GRCh38
rs777466079 Chr.15: 78935000-78935000 on Build GRCh38
rs777312265 Chr.15: 78934924-78934924 on Build GRCh38
rs149850795 Chr.15: 78935233-78935233 on Build GRCh38
rs758005960 Chr.15: 78934893-78934893 on Build GRCh38
rs769491003 Chr.15: 78934898-78934898 on Build GRCh38
rs35536541 Chr.15: 78934916-78934917 on Build GRCh38
rs754201172 Chr.15: 78934993-78934993 on Build GRCh38
rs376855196 Chr.15: 78934948-78934948 on Build GRCh38
rs904283744 Chr.15: 78935258-78935258 on Build GRCh38
rs769705580 Chr.15: 78934938-78934938 on Build GRCh38
rs745343457 Chr.15: 78935025-78935025 on Build GRCh38
rs147551459 Chr.15: 78935132-78935132 on Build GRCh38
rs764364344 Chr.15: 78934843-78934843 on Build GRCh38
rs747275706 Chr.15: 78934876-78934876 on Build GRCh38
rs761360105 Chr.15: 78934856-78934857 on Build GRCh38
rs745804659 Chr.15: 78934935-78934935 on Build GRCh38
rs766831718 Chr.15: 78935066-78935066 on Build GRCh38
rs768559983 Chr.15: 78935046-78935046 on Build GRCh38
rs572209103 Chr.15: 78934953-78934953 on Build GRCh38
rs758441356 Chr.15: 78935035-78935036 on Build GRCh38
rs542229489 Chr.15: 78935195-78935195 on Build GRCh38
rs766370610 Chr.15: 78935037-78935038 on Build GRCh38
rs1017799878 Chr.15: 78934821-78934821 on Build GRCh38
rs1037454348 Chr.15: 78935082-78935082 on Build GRCh38
rs561860873 Chr.15: 78935153-78935153 on Build GRCh38
rs777188123 Chr.15: 78935073-78935073 on Build GRCh38
rs372283224 Chr.15: 78935045-78935045 on Build GRCh38
rs868114560 Chr.15: 78935006-78935006 on Build GRCh38
rs74724679 Chr.15: 78935120-78935120 on Build GRCh38
rs751518176 Chr.15: 78935093-78935096 on Build GRCh38
rs768329442 Chr.15: 78935197-78935197 on Build GRCh38
rs1014648535 Chr.15: 78935067-78935067 on Build GRCh38
rs769040305 Chr.15: 78934951-78934951 on Build GRCh38
rs139938364 Chr.15: 78934888-78934889 on Build GRCh38
rs765597891 Chr.15: 78935078-78935078 on Build GRCh38
rs991017389 Chr.15: 78935200-78935200 on Build GRCh38
rs769669640 Chr.15: 78935026-78935026 on Build GRCh38
rs968394955 Chr.15: 78934841-78934841 on Build GRCh38
rs756588280 Chr.15: 78934874-78934874 on Build GRCh38
rs532381869 Chr.15: 78935144-78935144 on Build GRCh38
rs763089706 Chr.15: 78934820-78934820 on Build GRCh38
rs148414677 Chr.15: 78934816-78934816 on Build GRCh38
rs941348474 Chr.15: 78934968-78934968 on Build GRCh38
rs765093951 Chr.15: 78934917-78934917 on Build GRCh38
rs866741335 Chr.15: 78934971-78934971 on Build GRCh38
rs1038756752 Chr.15: 78935228-78935228 on Build GRCh38
rs111717641 Chr.15: 78934812-78934812 on Build GRCh38
rs760107873 Chr.15: 78935064-78935064 on Build GRCh38
rs761955150 Chr.15: 78934854-78934854 on Build GRCh38
rs774233788 Chr.15: 78935048-78935048 on Build GRCh38
rs528091807 Chr.15: 78935051-78935051 on Build GRCh38
rs751739082 Chr.15: 78934853-78934853 on Build GRCh38
rs199936044 Chr.15: 78935093-78935093 on Build GRCh38
rs750883613 Chr.15: 78934873-78934873 on Build GRCh38
rs760782047 Chr.15: 78935055-78935055 on Build GRCh38
rs188780244 Chr.15: 78935223-78935223 on Build GRCh38
rs201187590 Chr.15: 78934931-78934931 on Build GRCh38
rs905752698 Chr.15: 78935099-78935099 on Build GRCh38
rs754194618 Chr.15: 78934889-78934889 on Build GRCh38
rs944742055 Chr.15: 78935232-78935232 on Build GRCh38
rs755301352 Chr.15: 78934998-78934998 on Build GRCh38
rs761122599 Chr.15: 78934956-78934956 on Build GRCh38
rs780571910 Chr.15: 78935015-78935015 on Build GRCh38
rs756301467 Chr.15: 78935117-78935120 on Build GRCh38
rs955962693 Chr.15: 78934880-78934880 on Build GRCh38
rs771961955 Chr.15: 78934862-78934863 on Build GRCh38
rs768206788 Chr.15: 78934859-78934859 on Build GRCh38
rs184387793 Chr.15: 78935147-78935147 on Build GRCh38
rs751197826 Chr.15: 78935001-78935001 on Build GRCh38

View Full Product Details