Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80152925-80153448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAH
Entrez Gene ID
2184
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
-
UniGene
Hs.73875
Gene Chromosome Location
Chr.15: 80152891-80186582 on Build GRCh38
Transcripts
NM_000137.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835989 c.68C>G p.S23W Chr.15: 80153122-80153122 on Build GRCh38
6579064 c.22G>T p.E8* Chr.15: 80153076-80153076 on Build GRCh38
7199130 c.10A>G p.I4V Chr.15: 80153064-80153064 on Build GRCh38
5003530 c.55T>A p.Y19N Chr.15: 80153109-80153109 on Build GRCh38
6232546 c.63C>T p.V21V Chr.15: 80153117-80153117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554422109 Chr.15: 80153173-80153191 on Build GRCh38
rs755880952 Chr.15: 80153168-80153168 on Build GRCh38
rs768074832 Chr.15: 80153029-80153029 on Build GRCh38
rs111985816 Chr.15: 80152970-80152970 on Build GRCh38
rs920635101 Chr.15: 80153066-80153066 on Build GRCh38
rs371740875 Chr.15: 80153005-80153005 on Build GRCh38
rs570210399 Chr.15: 80152969-80152969 on Build GRCh38
rs372271309 Chr.15: 80153161-80153161 on Build GRCh38
rs774230227 Chr.15: 80153225-80153225 on Build GRCh38
rs558321563 Chr.15: 80153176-80153176 on Build GRCh38
rs1051057226 Chr.15: 80153084-80153084 on Build GRCh38
rs754336457 Chr.15: 80153064-80153064 on Build GRCh38
rs768146083 Chr.15: 80153126-80153126 on Build GRCh38
rs1001940632 Chr.15: 80153313-80153313 on Build GRCh38
rs1021218825 Chr.15: 80153413-80153413 on Build GRCh38
rs534453827 Chr.15: 80153170-80153171 on Build GRCh38
rs910647551 Chr.15: 80153022-80153022 on Build GRCh38
rs188046704 Chr.15: 80153143-80153143 on Build GRCh38
rs77616478 Chr.15: 80153089-80153089 on Build GRCh38
rs769838982 Chr.15: 80153020-80153020 on Build GRCh38
rs1057516934 Chr.15: 80153056-80153056 on Build GRCh38
rs199733231 Chr.15: 80153145-80153145 on Build GRCh38
rs371100968 Chr.15: 80153015-80153015 on Build GRCh38
rs777902403 Chr.15: 80153176-80153177 on Build GRCh38
rs377188658 Chr.15: 80153011-80153011 on Build GRCh38
rs960633870 Chr.15: 80152988-80152988 on Build GRCh38
rs767638459 Chr.15: 80153173-80153181 on Build GRCh38
rs567818237 Chr.15: 80153394-80153394 on Build GRCh38
rs1053848682 Chr.15: 80153359-80153359 on Build GRCh38
rs77692007 Chr.15: 80153275-80153275 on Build GRCh38
rs757176902 Chr.15: 80153171-80153175 on Build GRCh38
rs1031291810 Chr.15: 80152974-80152974 on Build GRCh38
rs574266745 Chr.15: 80153235-80153235 on Build GRCh38
rs71699670 Chr.15: 80153171-80153172 on Build GRCh38
rs752679360 Chr.15: 80153173-80153186 on Build GRCh38
rs752897835 Chr.15: 80153075-80153075 on Build GRCh38
rs867200761 Chr.15: 80153133-80153133 on Build GRCh38
rs867498731 Chr.15: 80153210-80153210 on Build GRCh38
rs201528408 Chr.15: 80153035-80153035 on Build GRCh38
rs772895065 Chr.15: 80153137-80153137 on Build GRCh38
rs747338192 Chr.15: 80153138-80153138 on Build GRCh38
rs763173058 Chr.15: 80153160-80153160 on Build GRCh38
rs771057671 Chr.15: 80153215-80153215 on Build GRCh38
rs142522569 Chr.15: 80153021-80153021 on Build GRCh38
rs200127538 Chr.15: 80153032-80153032 on Build GRCh38
rs550679976 Chr.15: 80152962-80152962 on Build GRCh38
rs969092117 Chr.15: 80153291-80153291 on Build GRCh38
rs771218632 Chr.15: 80153141-80153141 on Build GRCh38
rs758685960 Chr.15: 80153096-80153096 on Build GRCh38
rs765242250 Chr.15: 80153071-80153071 on Build GRCh38
rs759608245 Chr.15: 80153070-80153070 on Build GRCh38
rs60184934 Chr.15: 80153173-80153174 on Build GRCh38
rs200319726 Chr.15: 80153147-80153147 on Build GRCh38
rs563018217 Chr.15: 80153277-80153277 on Build GRCh38
rs1053103770 Chr.15: 80153248-80153248 on Build GRCh38
rs187323555 Chr.15: 80153002-80153002 on Build GRCh38
rs756381638 Chr.15: 80153173-80153201 on Build GRCh38
rs869215431 Chr.15: 80153174-80153192 on Build GRCh38
rs778283270 Chr.15: 80153129-80153129 on Build GRCh38
rs57837512 Chr.15: 80153210-80153214 on Build GRCh38
rs376322780 Chr.15: 80153172-80153176 on Build GRCh38
rs200704926 Chr.15: 80153012-80153012 on Build GRCh38
rs759366090 Chr.15: 80153226-80153227 on Build GRCh38
rs753734814 Chr.15: 80153185-80153185 on Build GRCh38
rs762924131 Chr.15: 80153173-80153176 on Build GRCh38
rs371489046 Chr.15: 80153113-80153113 on Build GRCh38
rs1057517341 Chr.15: 80153068-80153068 on Build GRCh38
rs1000181141 Chr.15: 80152963-80152963 on Build GRCh38
rs886051495 Chr.15: 80152988-80152988 on Build GRCh38
rs534184772 Chr.15: 80153211-80153211 on Build GRCh38
rs201702367 Chr.15: 80153228-80153228 on Build GRCh38
rs1057517972 Chr.15: 80153055-80153055 on Build GRCh38
rs751861502 Chr.15: 80153165-80153165 on Build GRCh38
rs958522605 Chr.15: 80153004-80153004 on Build GRCh38
rs1023362108 Chr.15: 80153006-80153006 on Build GRCh38
rs1005965971 Chr.15: 80153348-80153348 on Build GRCh38
rs375149490 Chr.15: 80153039-80153039 on Build GRCh38
rs578112638 Chr.15: 80153192-80153192 on Build GRCh38
rs121965073 Chr.15: 80153101-80153101 on Build GRCh38
rs749196943 Chr.15: 80153114-80153114 on Build GRCh38
rs1024694492 Chr.15: 80152973-80152973 on Build GRCh38
rs532122242 Chr.15: 80152992-80152992 on Build GRCh38
rs766882348 Chr.15: 80153057-80153057 on Build GRCh38
rs775696191 Chr.15: 80153156-80153156 on Build GRCh38
rs761078986 Chr.15: 80153042-80153042 on Build GRCh38
rs962166287 Chr.15: 80153410-80153410 on Build GRCh38
rs923234833 Chr.15: 80153037-80153037 on Build GRCh38
rs747716227 Chr.15: 80153130-80153130 on Build GRCh38
rs1039694386 Chr.15: 80153195-80153195 on Build GRCh38
rs112474268 Chr.15: 80152950-80152950 on Build GRCh38
rs569070594 Chr.15: 80153135-80153135 on Build GRCh38
rs756012361 Chr.15: 80153105-80153105 on Build GRCh38
rs74249373 Chr.15: 80153233-80153233 on Build GRCh38
rs971950108 Chr.15: 80153414-80153414 on Build GRCh38
rs759962349 Chr.15: 80153144-80153144 on Build GRCh38
rs748799603 Chr.15: 80153027-80153027 on Build GRCh38
rs1016052333 Chr.15: 80153399-80153399 on Build GRCh38
rs961937153 Chr.15: 80153023-80153023 on Build GRCh38
rs541677717 Chr.15: 80153223-80153224 on Build GRCh38
rs896748881 Chr.15: 80153264-80153264 on Build GRCh38
rs576624913 Chr.15: 80153288-80153288 on Build GRCh38

View Full Product Details