Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80562953-80563449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNT2
Entrez Gene ID
9915
Gene Name
aryl hydrocarbon receptor nuclear translocator 2
Gene Aliases
WEDAS, bHLHe1
UniGene
Hs.459070
Gene Chromosome Location
Chr.15: 80404350-80597937 on Build GRCh38
Transcripts
NM_014862.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4057241 c.1193C>T p.S398L Chr.15: 80563116-80563116 on Build GRCh38
4663458 c.1269C>T p.P423P Chr.15: 80563192-80563192 on Build GRCh38
1718092 c.1267C>T p.P423S Chr.15: 80563190-80563190 on Build GRCh38
965515 c.1205G>A p.R402Q Chr.15: 80563128-80563128 on Build GRCh38
5799840 c.1172A>C p.K391T Chr.15: 80563095-80563095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760219727 Chr.15: 80563244-80563244 on Build GRCh38
rs751683131 Chr.15: 80563276-80563276 on Build GRCh38
rs758396995 Chr.15: 80563163-80563163 on Build GRCh38
rs767137998 Chr.15: 80563138-80563138 on Build GRCh38
rs112669114 Chr.15: 80563267-80563267 on Build GRCh38
rs370619205 Chr.15: 80563180-80563180 on Build GRCh38
rs190462241 Chr.15: 80563354-80563354 on Build GRCh38
rs755579298 Chr.15: 80563102-80563102 on Build GRCh38
rs571855678 Chr.15: 80563294-80563294 on Build GRCh38
rs769473257 Chr.15: 80563119-80563119 on Build GRCh38
rs548713623 Chr.15: 80563134-80563134 on Build GRCh38
rs1011091822 Chr.15: 80563286-80563286 on Build GRCh38
rs142574625 Chr.15: 80563145-80563145 on Build GRCh38
rs373157030 Chr.15: 80563055-80563055 on Build GRCh38
rs568307746 Chr.15: 80563024-80563024 on Build GRCh38
rs767105573 Chr.15: 80563062-80563062 on Build GRCh38
rs778742835 Chr.15: 80563289-80563289 on Build GRCh38
rs747169199 Chr.15: 80563169-80563169 on Build GRCh38
rs1055243285 Chr.15: 80563248-80563248 on Build GRCh38
rs113524323 Chr.15: 80563016-80563016 on Build GRCh38
rs563959728 Chr.15: 80562980-80562980 on Build GRCh38
rs141035056 Chr.15: 80563213-80563213 on Build GRCh38
rs911597186 Chr.15: 80563061-80563061 on Build GRCh38
rs527417551 Chr.15: 80562994-80562994 on Build GRCh38
rs748273741 Chr.15: 80563110-80563110 on Build GRCh38
rs973929453 Chr.15: 80563111-80563111 on Build GRCh38
rs200026422 Chr.15: 80563260-80563260 on Build GRCh38
rs780523195 Chr.15: 80563080-80563080 on Build GRCh38
rs894384279 Chr.15: 80563302-80563302 on Build GRCh38
rs921936810 Chr.15: 80563389-80563389 on Build GRCh38
rs568395792 Chr.15: 80563344-80563344 on Build GRCh38
rs370267927 Chr.15: 80563264-80563264 on Build GRCh38
rs375154338 Chr.15: 80563279-80563279 on Build GRCh38
rs147773330 Chr.15: 80563122-80563122 on Build GRCh38
rs574251932 Chr.15: 80563339-80563339 on Build GRCh38
rs182072402 Chr.15: 80563368-80563368 on Build GRCh38
rs531624246 Chr.15: 80563311-80563311 on Build GRCh38
rs776157951 Chr.15: 80563382-80563382 on Build GRCh38
rs966567482 Chr.15: 80563355-80563355 on Build GRCh38
rs4778606 Chr.15: 80563332-80563332 on Build GRCh38
rs761315788 Chr.15: 80563137-80563137 on Build GRCh38
rs141193900 Chr.15: 80563128-80563128 on Build GRCh38
rs976103211 Chr.15: 80563369-80563369 on Build GRCh38
rs773964764 Chr.15: 80563117-80563117 on Build GRCh38
rs976514335 Chr.15: 80563172-80563172 on Build GRCh38
rs866542302 Chr.15: 80563191-80563191 on Build GRCh38
rs774284119 Chr.15: 80563233-80563233 on Build GRCh38
rs775387829 Chr.15: 80563227-80563228 on Build GRCh38
rs560227882 Chr.15: 80563010-80563010 on Build GRCh38
rs1026735581 Chr.15: 80563316-80563316 on Build GRCh38
rs760439735 Chr.15: 80563139-80563139 on Build GRCh38
rs113239024 Chr.15: 80562978-80562978 on Build GRCh38
rs771538158 Chr.15: 80563342-80563342 on Build GRCh38
rs990421716 Chr.15: 80563423-80563423 on Build GRCh38
rs779091420 Chr.15: 80563104-80563104 on Build GRCh38
rs1014945272 Chr.15: 80563338-80563338 on Build GRCh38
rs1019924325 Chr.15: 80563002-80563002 on Build GRCh38
rs754379136 Chr.15: 80563116-80563116 on Build GRCh38
rs537962071 Chr.15: 80563235-80563235 on Build GRCh38
rs556296961 Chr.15: 80563395-80563395 on Build GRCh38
rs761405299 Chr.15: 80563045-80563045 on Build GRCh38
rs763564645 Chr.15: 80563039-80563039 on Build GRCh38
rs765758088 Chr.15: 80563263-80563263 on Build GRCh38
rs951231664 Chr.15: 80563327-80563327 on Build GRCh38
rs377428762 Chr.15: 80563036-80563036 on Build GRCh38
rs150964641 Chr.15: 80563151-80563151 on Build GRCh38
rs768511501 Chr.15: 80563224-80563224 on Build GRCh38
rs750731438 Chr.15: 80563067-80563067 on Build GRCh38
rs142862370 Chr.15: 80562997-80562997 on Build GRCh38
rs764058530 Chr.15: 80563268-80563268 on Build GRCh38
rs768601519 Chr.15: 80563133-80563133 on Build GRCh38
rs906883678 Chr.15: 80563022-80563022 on Build GRCh38
rs755505716 Chr.15: 80563146-80563146 on Build GRCh38
rs4778813 Chr.15: 80563402-80563402 on Build GRCh38
rs772356140 Chr.15: 80563114-80563114 on Build GRCh38
rs181670560 Chr.15: 80562983-80562983 on Build GRCh38
rs528834296 Chr.15: 80563042-80563042 on Build GRCh38
rs377684761 Chr.15: 80563075-80563075 on Build GRCh38
rs748139605 Chr.15: 80563234-80563234 on Build GRCh38
rs367632725 Chr.15: 80563246-80563246 on Build GRCh38
rs201779344 Chr.15: 80563243-80563243 on Build GRCh38
rs368438348 Chr.15: 80563280-80563280 on Build GRCh38
rs186761656 Chr.15: 80563201-80563201 on Build GRCh38
rs766617881 Chr.15: 80563142-80563142 on Build GRCh38
rs754752706 Chr.15: 80563288-80563288 on Build GRCh38
rs1034256024 Chr.15: 80563047-80563047 on Build GRCh38
rs754291471 Chr.15: 80563086-80563086 on Build GRCh38
rs932261118 Chr.15: 80563230-80563230 on Build GRCh38

View Full Product Details