Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80695048-80695561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17C
Entrez Gene ID
58489
Gene Name
abhydrolase domain containing 17C
Gene Aliases
FAM108C1
UniGene
Hs.459072
Gene Chromosome Location
Chr.15: 80695311-80755621 on Build GRCh38
Transcripts
NM_021214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959695 c.17C>A p.P6H Chr.15: 80695446-80695446 on Build GRCh38
1206005 c.81C>A p.S27R Chr.15: 80695510-80695510 on Build GRCh38
3754502 c.42T>C p.G14G Chr.15: 80695471-80695471 on Build GRCh38
6233464 c.46C>T p.L16L Chr.15: 80695475-80695475 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs147940975 Chr.15: 80695408-80695408 on Build GRCh38
rs748846703 Chr.15: 80695491-80695491 on Build GRCh38
rs975765618 Chr.15: 80695094-80695094 on Build GRCh38
rs1045640606 Chr.15: 80695459-80695459 on Build GRCh38
rs997153049 Chr.15: 80695312-80695312 on Build GRCh38
rs941991451 Chr.15: 80695149-80695149 on Build GRCh38
rs1030017887 Chr.15: 80695318-80695318 on Build GRCh38
rs563717383 Chr.15: 80695104-80695104 on Build GRCh38
rs956454154 Chr.15: 80695086-80695086 on Build GRCh38
rs936842930 Chr.15: 80695528-80695528 on Build GRCh38
rs779474573 Chr.15: 80695486-80695486 on Build GRCh38
rs144819652 Chr.15: 80695301-80695301 on Build GRCh38
rs1031216273 Chr.15: 80695280-80695280 on Build GRCh38
rs769758711 Chr.15: 80695476-80695476 on Build GRCh38
rs1056531961 Chr.15: 80695181-80695181 on Build GRCh38
rs948203777 Chr.15: 80695232-80695232 on Build GRCh38
rs904097879 Chr.15: 80695267-80695267 on Build GRCh38
rs865792040 Chr.15: 80695139-80695139 on Build GRCh38
rs779184127 Chr.15: 80695400-80695400 on Build GRCh38
rs868416762 Chr.15: 80695294-80695294 on Build GRCh38
rs986004787 Chr.15: 80695138-80695138 on Build GRCh38
rs866773020 Chr.15: 80695092-80695092 on Build GRCh38
rs963544297 Chr.15: 80695366-80695366 on Build GRCh38
rs896589640 Chr.15: 80695222-80695222 on Build GRCh38
rs919187088 Chr.15: 80695371-80695371 on Build GRCh38
rs974477651 Chr.15: 80695164-80695164 on Build GRCh38
rs757046851 Chr.15: 80695409-80695409 on Build GRCh38
rs781014658 Chr.15: 80695423-80695423 on Build GRCh38
rs544887646 Chr.15: 80695115-80695116 on Build GRCh38
rs755087500 Chr.15: 80695393-80695393 on Build GRCh38
rs1030661121 Chr.15: 80695073-80695073 on Build GRCh38
rs1001164180 Chr.15: 80695274-80695275 on Build GRCh38
rs971939284 Chr.15: 80695367-80695367 on Build GRCh38
rs953306511 Chr.15: 80695132-80695132 on Build GRCh38
rs929967019 Chr.15: 80695176-80695176 on Build GRCh38
rs768105705 Chr.15: 80695498-80695498 on Build GRCh38
rs1000928583 Chr.15: 80695069-80695069 on Build GRCh38
rs1052610036 Chr.15: 80695543-80695543 on Build GRCh38
rs929936066 Chr.15: 80695378-80695381 on Build GRCh38
rs953184119 Chr.15: 80695331-80695331 on Build GRCh38
rs992185043 Chr.15: 80695393-80695394 on Build GRCh38
rs117959127 Chr.15: 80695334-80695334 on Build GRCh38
rs918549468 Chr.15: 80695167-80695167 on Build GRCh38
rs1045267338 Chr.15: 80695236-80695236 on Build GRCh38
rs948198298 Chr.15: 80695441-80695441 on Build GRCh38
rs552660333 Chr.15: 80695332-80695332 on Build GRCh38
rs62007944 Chr.15: 80695471-80695471 on Build GRCh38
rs112654755 Chr.15: 80695145-80695145 on Build GRCh38

View Full Product Details