Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82157623-82158134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFL1
Entrez Gene ID
79631
Gene Name
elongation factor like GTPase 1
Gene Aliases
EFTUD1, FAM42A, HsT19294, RIA1
UniGene
Hs.459114
Gene Chromosome Location
Chr.15: 82130220-82262763 on Build GRCh38
Transcripts
NM_001040610.2 NM_001322844.1 NM_001322845.1 NM_024580.5 NR_136410.1 XM_011522000.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629709 c.1929T>G p.A643A Chr.15: 82157814-82157814 on Build GRCh38
6652128 c.2006G>A p.R669Q Chr.15: 82157737-82157737 on Build GRCh38
3420660 c.2026G>T p.E676* Chr.15: 82157717-82157717 on Build GRCh38
1478391 c.1997A>G p.H666R Chr.15: 82157746-82157746 on Build GRCh38
5849909 c.2030+2_2030+3delTT p.? Chr.15: 82157710-82157711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976445767 Chr.15: 82157856-82157856 on Build GRCh38
rs750088840 Chr.15: 82157724-82157724 on Build GRCh38
rs761159776 Chr.15: 82157710-82157710 on Build GRCh38
rs781414667 Chr.15: 82157801-82157801 on Build GRCh38
rs757430233 Chr.15: 82157800-82157800 on Build GRCh38
rs34673670 Chr.15: 82157810-82157811 on Build GRCh38
rs747197141 Chr.15: 82157799-82157799 on Build GRCh38
rs1009559393 Chr.15: 82157934-82157934 on Build GRCh38
rs9944185 Chr.15: 82157927-82157927 on Build GRCh38
rs767762288 Chr.15: 82157753-82157755 on Build GRCh38
rs760454181 Chr.15: 82157728-82157728 on Build GRCh38
rs909715444 Chr.15: 82157948-82157948 on Build GRCh38
rs751588680 Chr.15: 82157908-82157908 on Build GRCh38
rs759755447 Chr.15: 82157880-82157880 on Build GRCh38
rs1009201883 Chr.15: 82158061-82158061 on Build GRCh38
rs369250493 Chr.15: 82157896-82157896 on Build GRCh38
rs371498555 Chr.15: 82157771-82157771 on Build GRCh38
rs926446917 Chr.15: 82157722-82157722 on Build GRCh38
rs772890027 Chr.15: 82157851-82157851 on Build GRCh38
rs768501559 Chr.15: 82157808-82157808 on Build GRCh38
rs913791442 Chr.15: 82157683-82157683 on Build GRCh38
rs10152266 Chr.15: 82158062-82158062 on Build GRCh38
rs748176247 Chr.15: 82157818-82157818 on Build GRCh38
rs779232326 Chr.15: 82157772-82157772 on Build GRCh38
rs375810682 Chr.15: 82157875-82157875 on Build GRCh38
rs760296936 Chr.15: 82157852-82157852 on Build GRCh38
rs747287007 Chr.15: 82157675-82157675 on Build GRCh38
rs778568710 Chr.15: 82157952-82157952 on Build GRCh38
rs202018584 Chr.15: 82157693-82157693 on Build GRCh38
rs779899955 Chr.15: 82157694-82157694 on Build GRCh38
rs753120805 Chr.15: 82157895-82157895 on Build GRCh38
rs772210380 Chr.15: 82157667-82157667 on Build GRCh38
rs573370586 Chr.15: 82157890-82157890 on Build GRCh38
rs766820458 Chr.15: 82157730-82157730 on Build GRCh38
rs752907647 Chr.15: 82157852-82157853 on Build GRCh38
rs553387088 Chr.15: 82157695-82157695 on Build GRCh38
rs774992945 Chr.15: 82158097-82158097 on Build GRCh38
rs369620042 Chr.15: 82157781-82157781 on Build GRCh38
rs1000390380 Chr.15: 82158003-82158003 on Build GRCh38
rs767204729 Chr.15: 82157713-82157713 on Build GRCh38
rs1003490040 Chr.15: 82158052-82158052 on Build GRCh38
rs79299829 Chr.15: 82157692-82157692 on Build GRCh38
rs192083503 Chr.15: 82158105-82158105 on Build GRCh38
rs200198460 Chr.15: 82157663-82157663 on Build GRCh38
rs1032070756 Chr.15: 82158024-82158024 on Build GRCh38
rs777875073 Chr.15: 82157785-82157785 on Build GRCh38
rs552647976 Chr.15: 82157876-82157876 on Build GRCh38
rs754115545 Chr.15: 82157757-82157757 on Build GRCh38
rs753256277 Chr.15: 82157774-82157774 on Build GRCh38
rs773687582 Chr.15: 82157989-82157989 on Build GRCh38
rs957786145 Chr.15: 82158034-82158034 on Build GRCh38
rs188727377 Chr.15: 82157996-82157996 on Build GRCh38
rs905145878 Chr.15: 82158043-82158043 on Build GRCh38
rs1047735497 Chr.15: 82157915-82157915 on Build GRCh38
rs764613364 Chr.15: 82157897-82157897 on Build GRCh38
rs756525782 Chr.15: 82157888-82157892 on Build GRCh38
rs540791285 Chr.15: 82157665-82157665 on Build GRCh38
rs1010072376 Chr.15: 82158090-82158090 on Build GRCh38
rs920704965 Chr.15: 82157844-82157844 on Build GRCh38
rs771685905 Chr.15: 82157843-82157843 on Build GRCh38
rs748976302 Chr.15: 82157802-82157802 on Build GRCh38
rs777963477 Chr.15: 82157668-82157668 on Build GRCh38
rs774152799 Chr.15: 82157810-82157810 on Build GRCh38
rs1044942507 Chr.15: 82158041-82158041 on Build GRCh38

View Full Product Details