Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82546751-82547268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB1
Entrez Gene ID
64506
Gene Name
cytoplasmic polyadenylation element binding protein 1
Gene Aliases
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1
UniGene
Hs.459132
Gene Chromosome Location
Chr.15: 82543201-82648016 on Build GRCh38
Transcripts
NM_001079533.1 NM_001079534.1 NM_001079535.1 NM_001288819.1 NM_001288820.1 NM_030594.4

SNP Detail

SNP ID Chromosome Location
rs375483967 Chr.15: 82546908-82546908 on Build GRCh38
rs200803573 Chr.15: 82547207-82547207 on Build GRCh38
rs1001276757 Chr.15: 82546823-82546823 on Build GRCh38
rs761159671 Chr.15: 82546809-82546809 on Build GRCh38
rs768160786 Chr.15: 82547161-82547161 on Build GRCh38
rs373749169 Chr.15: 82547184-82547184 on Build GRCh38
rs946722934 Chr.15: 82547041-82547041 on Build GRCh38
rs942760506 Chr.15: 82547069-82547069 on Build GRCh38
rs768078681 Chr.15: 82546838-82546838 on Build GRCh38
rs987025848 Chr.15: 82547003-82547003 on Build GRCh38
rs925242126 Chr.15: 82547106-82547106 on Build GRCh38
rs910628816 Chr.15: 82547091-82547091 on Build GRCh38
rs759667850 Chr.15: 82547109-82547109 on Build GRCh38
rs757095839 Chr.15: 82547125-82547125 on Build GRCh38
rs1049298382 Chr.15: 82547216-82547216 on Build GRCh38
rs1025125302 Chr.15: 82546798-82546798 on Build GRCh38
rs569487320 Chr.15: 82546981-82546981 on Build GRCh38
rs375389219 Chr.15: 82546827-82546827 on Build GRCh38
rs531307030 Chr.15: 82546894-82546894 on Build GRCh38
rs986210523 Chr.15: 82547055-82547055 on Build GRCh38
rs779375273 Chr.15: 82547151-82547151 on Build GRCh38
rs369687360 Chr.15: 82546919-82546919 on Build GRCh38
rs747825334 Chr.15: 82547165-82547165 on Build GRCh38
rs772522093 Chr.15: 82547173-82547173 on Build GRCh38
rs751328558 Chr.15: 82547122-82547122 on Build GRCh38
rs750879154 Chr.15: 82546966-82546966 on Build GRCh38
rs776571408 Chr.15: 82547224-82547224 on Build GRCh38
rs1014181918 Chr.15: 82546782-82546782 on Build GRCh38
rs761026665 Chr.15: 82547190-82547190 on Build GRCh38
rs374011661 Chr.15: 82547185-82547185 on Build GRCh38
rs748893773 Chr.15: 82547158-82547158 on Build GRCh38
rs756563787 Chr.15: 82546984-82546984 on Build GRCh38
rs773879980 Chr.15: 82546802-82546802 on Build GRCh38
rs143704907 Chr.15: 82546940-82546940 on Build GRCh38
rs200967570 Chr.15: 82547120-82547120 on Build GRCh38
rs1025699508 Chr.15: 82546795-82546796 on Build GRCh38
rs571323411 Chr.15: 82547081-82547081 on Build GRCh38
rs1026000556 Chr.15: 82546989-82546989 on Build GRCh38
rs951218181 Chr.15: 82546999-82546999 on Build GRCh38
rs915205073 Chr.15: 82547034-82547034 on Build GRCh38
rs906942653 Chr.15: 82546806-82546806 on Build GRCh38
rs750321237 Chr.15: 82547131-82547131 on Build GRCh38
rs778756628 Chr.15: 82547162-82547162 on Build GRCh38
rs973978193 Chr.15: 82546986-82546986 on Build GRCh38
rs562320657 Chr.15: 82546787-82546787 on Build GRCh38
rs1034944786 Chr.15: 82546971-82546971 on Build GRCh38
rs763687022 Chr.15: 82546794-82546794 on Build GRCh38
rs759706108 Chr.15: 82547244-82547244 on Build GRCh38
rs981937297 Chr.15: 82546945-82546945 on Build GRCh38
rs756083970 Chr.15: 82547140-82547140 on Build GRCh38
rs780988002 Chr.15: 82547126-82547126 on Build GRCh38
rs912424357 Chr.15: 82547011-82547011 on Build GRCh38

View Full Product Details