Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 84840323-84840828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK3
Entrez Gene ID
57538
Gene Name
alpha kinase 3
Gene Aliases
MAK, MIDORI
UniGene
Hs.459183
Gene Chromosome Location
Chr.15: 84816631-84873482 on Build GRCh38
Transcripts
NM_020778.4 XM_017022444.1 XM_017022445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383989 c.1657G>A p.E553K Chr.15: 84840330-84840330 on Build GRCh38
701546 c.2008C>G p.H670D Chr.15: 84840681-84840681 on Build GRCh38
4402736 c.2059C>T p.P687S Chr.15: 84840732-84840732 on Build GRCh38
1182614 c.2093G>A p.S698N Chr.15: 84840766-84840766 on Build GRCh38
3504756 c.1947G>A p.K649K Chr.15: 84840620-84840620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546889222 Chr.15: 84840437-84840437 on Build GRCh38
rs267604352 Chr.15: 84840666-84840666 on Build GRCh38
rs1004733773 Chr.15: 84840686-84840686 on Build GRCh38
rs768342367 Chr.15: 84840351-84840351 on Build GRCh38
rs541903958 Chr.15: 84840722-84840722 on Build GRCh38
rs142698933 Chr.15: 84840803-84840803 on Build GRCh38
rs768234276 Chr.15: 84840451-84840451 on Build GRCh38
rs781488810 Chr.15: 84840522-84840522 on Build GRCh38
rs899839701 Chr.15: 84840725-84840725 on Build GRCh38
rs776620295 Chr.15: 84840493-84840493 on Build GRCh38
rs755210023 Chr.15: 84840406-84840406 on Build GRCh38
rs752241199 Chr.15: 84840724-84840724 on Build GRCh38
rs180863308 Chr.15: 84840798-84840798 on Build GRCh38
rs762134213 Chr.15: 84840366-84840366 on Build GRCh38
rs747749026 Chr.15: 84840468-84840468 on Build GRCh38
rs3803405 Chr.15: 84840409-84840409 on Build GRCh38
rs3803406 Chr.15: 84840608-84840608 on Build GRCh38
rs773497563 Chr.15: 84840485-84840485 on Build GRCh38
rs781665070 Chr.15: 84840744-84840744 on Build GRCh38
rs748468548 Chr.15: 84840800-84840800 on Build GRCh38
rs1032440625 Chr.15: 84840431-84840431 on Build GRCh38
rs774736837 Chr.15: 84840696-84840696 on Build GRCh38
rs780739176 Chr.15: 84840784-84840784 on Build GRCh38
rs34409363 Chr.15: 84840661-84840661 on Build GRCh38
rs528843595 Chr.15: 84840545-84840545 on Build GRCh38
rs751347673 Chr.15: 84840410-84840410 on Build GRCh38
rs751947135 Chr.15: 84840411-84840420 on Build GRCh38
rs759626376 Chr.15: 84840497-84840497 on Build GRCh38
rs754238381 Chr.15: 84840583-84840583 on Build GRCh38
rs774505914 Chr.15: 84840799-84840799 on Build GRCh38
rs756727368 Chr.15: 84840783-84840783 on Build GRCh38
rs756056773 Chr.15: 84840527-84840527 on Build GRCh38
rs755561788 Chr.15: 84840488-84840489 on Build GRCh38
rs757145005 Chr.15: 84840413-84840413 on Build GRCh38
rs945161495 Chr.15: 84840521-84840521 on Build GRCh38
rs778460334 Chr.15: 84840653-84840653 on Build GRCh38
rs745807626 Chr.15: 84840427-84840427 on Build GRCh38
rs748981859 Chr.15: 84840808-84840808 on Build GRCh38
rs570681326 Chr.15: 84840620-84840620 on Build GRCh38
rs753944954 Chr.15: 84840404-84840404 on Build GRCh38
rs780011424 Chr.15: 84840528-84840528 on Build GRCh38
rs568908757 Chr.15: 84840582-84840582 on Build GRCh38
rs148053797 Chr.15: 84840690-84840690 on Build GRCh38
rs143928374 Chr.15: 84840371-84840371 on Build GRCh38
rs781080280 Chr.15: 84840425-84840425 on Build GRCh38
rs761883111 Chr.15: 84840700-84840700 on Build GRCh38
rs765187060 Chr.15: 84840505-84840505 on Build GRCh38
rs758630539 Chr.15: 84840520-84840520 on Build GRCh38
rs750686743 Chr.15: 84840346-84840346 on Build GRCh38
rs749765619 Chr.15: 84840792-84840792 on Build GRCh38
rs759573630 Chr.15: 84840715-84840715 on Build GRCh38
rs750284989 Chr.15: 84840621-84840621 on Build GRCh38
rs771343949 Chr.15: 84840672-84840672 on Build GRCh38
rs777704274 Chr.15: 84840575-84840575 on Build GRCh38
rs751233026 Chr.15: 84840733-84840733 on Build GRCh38
rs1033371309 Chr.15: 84840599-84840599 on Build GRCh38
rs375166303 Chr.15: 84840652-84840652 on Build GRCh38
rs375574739 Chr.15: 84840647-84840647 on Build GRCh38
rs749140872 Chr.15: 84840529-84840529 on Build GRCh38
rs150985765 Chr.15: 84840736-84840736 on Build GRCh38
rs962765876 Chr.15: 84840689-84840689 on Build GRCh38
rs753827719 Chr.15: 84840632-84840632 on Build GRCh38
rs962690116 Chr.15: 84840405-84840405 on Build GRCh38
rs558339788 Chr.15: 84840597-84840597 on Build GRCh38
rs766372253 Chr.15: 84840391-84840391 on Build GRCh38
rs759907114 Chr.15: 84840602-84840602 on Build GRCh38
rs752985553 Chr.15: 84840518-84840518 on Build GRCh38
rs202163284 Chr.15: 84840682-84840682 on Build GRCh38
rs778227903 Chr.15: 84840539-84840539 on Build GRCh38
rs368346727 Chr.15: 84840574-84840574 on Build GRCh38
rs770190564 Chr.15: 84840687-84840687 on Build GRCh38
rs746457084 Chr.15: 84840685-84840685 on Build GRCh38
rs143709902 Chr.15: 84840691-84840691 on Build GRCh38
rs769139957 Chr.15: 84840691-84840691 on Build GRCh38
rs747711369 Chr.15: 84840681-84840684 on Build GRCh38
rs932897432 Chr.15: 84840740-84840740 on Build GRCh38
rs867857578 Chr.15: 84840635-84840635 on Build GRCh38
rs966914903 Chr.15: 84840785-84840785 on Build GRCh38
rs147228539 Chr.15: 84840701-84840701 on Build GRCh38
rs761234447 Chr.15: 84840486-84840486 on Build GRCh38
rs114207635 Chr.15: 84840801-84840801 on Build GRCh38
rs199986476 Chr.15: 84840615-84840615 on Build GRCh38
rs754539409 Chr.15: 84840533-84840533 on Build GRCh38
rs115200412 Chr.15: 84840565-84840565 on Build GRCh38
rs748809965 Chr.15: 84840447-84840447 on Build GRCh38
rs758092874 Chr.15: 84840662-84840662 on Build GRCh38
rs757931507 Chr.15: 84840728-84840728 on Build GRCh38
rs774084724 Chr.15: 84840454-84840454 on Build GRCh38
rs748962163 Chr.15: 84840350-84840350 on Build GRCh38
rs746198662 Chr.15: 84840767-84840767 on Build GRCh38
rs55702300 Chr.15: 84840478-84840478 on Build GRCh38
rs982846198 Chr.15: 84840675-84840675 on Build GRCh38
rs750231929 Chr.15: 84840524-84840524 on Build GRCh38
rs746598957 Chr.15: 84840671-84840671 on Build GRCh38
rs756119370 Chr.15: 84840435-84840435 on Build GRCh38
rs146781915 Chr.15: 84840383-84840383 on Build GRCh38
rs771706498 Chr.15: 84840558-84840558 on Build GRCh38
rs777211313 Chr.15: 84840730-84840730 on Build GRCh38
rs768817367 Chr.15: 84840695-84840695 on Build GRCh38
rs371411006 Chr.15: 84840668-84840668 on Build GRCh38
rs773730492 Chr.15: 84840372-84840372 on Build GRCh38
rs766039293 Chr.15: 84840630-84840630 on Build GRCh38
rs774628152 Chr.15: 84840360-84840360 on Build GRCh38
rs777370400 Chr.15: 84840663-84840663 on Build GRCh38
rs760550091 Chr.15: 84840629-84840629 on Build GRCh38
rs777436102 Chr.15: 84840726-84840726 on Build GRCh38

View Full Product Details