Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13926564-13927076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516728 c.457C>T p.R153C Chr.16: 13926629-13926629 on Build GRCh38
94242 c.553C>T p.L185F Chr.16: 13926725-13926725 on Build GRCh38
6917294 c.458G>C p.R153P Chr.16: 13926630-13926630 on Build GRCh38
6956981 c.539G>A p.R180K Chr.16: 13926711-13926711 on Build GRCh38
6985212 c.467A>G p.N156S Chr.16: 13926639-13926639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184763511 Chr.16: 13926978-13926978 on Build GRCh38
rs938654300 Chr.16: 13926918-13926918 on Build GRCh38
rs139989614 Chr.16: 13926619-13926619 on Build GRCh38
rs771645219 Chr.16: 13926804-13926804 on Build GRCh38
rs775540682 Chr.16: 13926692-13926692 on Build GRCh38
rs3136093 Chr.16: 13926946-13926949 on Build GRCh38
rs67158443 Chr.16: 13926943-13926946 on Build GRCh38
rs377547617 Chr.16: 13926746-13926746 on Build GRCh38
rs748779012 Chr.16: 13926637-13926637 on Build GRCh38
rs149927607 Chr.16: 13926704-13926704 on Build GRCh38
rs1002918079 Chr.16: 13926595-13926595 on Build GRCh38
rs377014538 Chr.16: 13926621-13926621 on Build GRCh38
rs3136092 Chr.16: 13926643-13926643 on Build GRCh38
rs2020961 Chr.16: 13926675-13926675 on Build GRCh38
rs113129434 Chr.16: 13926727-13926727 on Build GRCh38
rs1029139919 Chr.16: 13926810-13926810 on Build GRCh38
rs374092262 Chr.16: 13926955-13926955 on Build GRCh38
rs373408411 Chr.16: 13926709-13926709 on Build GRCh38
rs369698147 Chr.16: 13926856-13926856 on Build GRCh38
rs894176210 Chr.16: 13926973-13926973 on Build GRCh38
rs761092120 Chr.16: 13926693-13926693 on Build GRCh38
rs372888350 Chr.16: 13926962-13926962 on Build GRCh38
rs121913050 Chr.16: 13926630-13926630 on Build GRCh38
rs1012646362 Chr.16: 13926645-13926645 on Build GRCh38
rs764739513 Chr.16: 13926802-13926803 on Build GRCh38
rs768003187 Chr.16: 13926859-13926859 on Build GRCh38
rs879880504 Chr.16: 13927007-13927007 on Build GRCh38
rs906343483 Chr.16: 13926981-13926981 on Build GRCh38
rs750673145 Chr.16: 13926713-13926713 on Build GRCh38
rs776329282 Chr.16: 13926750-13926755 on Build GRCh38
rs909315833 Chr.16: 13926847-13926847 on Build GRCh38
rs147455220 Chr.16: 13926598-13926598 on Build GRCh38
rs761297555 Chr.16: 13926781-13926783 on Build GRCh38
rs879228106 Chr.16: 13926914-13926914 on Build GRCh38
rs138724289 Chr.16: 13926592-13926592 on Build GRCh38
rs760865964 Chr.16: 13926593-13926593 on Build GRCh38
rs957009045 Chr.16: 13926706-13926706 on Build GRCh38
rs764093404 Chr.16: 13926597-13926597 on Build GRCh38
rs752672749 Chr.16: 13926626-13926626 on Build GRCh38
rs986184239 Chr.16: 13926829-13926829 on Build GRCh38
rs887033897 Chr.16: 13927011-13927011 on Build GRCh38
rs545724483 Chr.16: 13927015-13927015 on Build GRCh38
rs1057185300 Chr.16: 13926964-13926964 on Build GRCh38
rs756785482 Chr.16: 13926766-13926766 on Build GRCh38
rs189141453 Chr.16: 13927040-13927040 on Build GRCh38
rs745335783 Chr.16: 13926876-13926876 on Build GRCh38
rs979626150 Chr.16: 13926687-13926687 on Build GRCh38
rs145402255 Chr.16: 13926620-13926620 on Build GRCh38
rs765509563 Chr.16: 13926600-13926600 on Build GRCh38
rs573473317 Chr.16: 13927043-13927043 on Build GRCh38
rs770535929 Chr.16: 13926642-13926642 on Build GRCh38
rs867821652 Chr.16: 13926935-13926935 on Build GRCh38
rs753325454 Chr.16: 13926751-13926751 on Build GRCh38
rs756024042 Chr.16: 13926741-13926741 on Build GRCh38
rs778574449 Chr.16: 13926659-13926659 on Build GRCh38
rs1036207235 Chr.16: 13926877-13926877 on Build GRCh38
rs897728892 Chr.16: 13926908-13926908 on Build GRCh38
rs1023747150 Chr.16: 13926668-13926668 on Build GRCh38
rs750292787 Chr.16: 13926802-13926805 on Build GRCh38
rs989199304 Chr.16: 13926954-13926954 on Build GRCh38
rs879831172 Chr.16: 13926771-13926780 on Build GRCh38
rs1044904211 Chr.16: 13926977-13926977 on Build GRCh38
rs762535375 Chr.16: 13927031-13927031 on Build GRCh38
rs181993439 Chr.16: 13926647-13926647 on Build GRCh38
rs368419719 Chr.16: 13926778-13926778 on Build GRCh38
rs746296279 Chr.16: 13926644-13926644 on Build GRCh38
rs370474460 Chr.16: 13926774-13926774 on Build GRCh38
rs1021289269 Chr.16: 13927004-13927004 on Build GRCh38
rs923291168 Chr.16: 13926744-13926744 on Build GRCh38
rs767437700 Chr.16: 13926725-13926725 on Build GRCh38
rs765599689 Chr.16: 13926712-13926712 on Build GRCh38
rs772205098 Chr.16: 13926633-13926633 on Build GRCh38
rs924224208 Chr.16: 13926936-13926936 on Build GRCh38
rs776377390 Chr.16: 13926975-13926975 on Build GRCh38
rs751450497 Chr.16: 13926602-13926602 on Build GRCh38

View Full Product Details