Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19307454-19307960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC19A
Entrez Gene ID
728276
Gene Name
C-type lectin domain family 19 member A
Gene Aliases
-
UniGene
Hs.632180
Gene Chromosome Location
Chr.16: 19285783-19310947 on Build GRCh38
Transcripts
NM_001256720.1

SNP Detail

SNP ID Chromosome Location
rs543877645 Chr.16: 19307548-19307548 on Build GRCh38
rs748327746 Chr.16: 19307518-19307518 on Build GRCh38
rs987483012 Chr.16: 19307864-19307864 on Build GRCh38
rs559910402 Chr.16: 19307741-19307741 on Build GRCh38
rs1025159975 Chr.16: 19307690-19307690 on Build GRCh38
rs775396331 Chr.16: 19307600-19307600 on Build GRCh38
rs551792736 Chr.16: 19307858-19307858 on Build GRCh38
rs941151607 Chr.16: 19307497-19307499 on Build GRCh38
rs773105952 Chr.16: 19307535-19307535 on Build GRCh38
rs753744626 Chr.16: 19307634-19307636 on Build GRCh38
rs1056303218 Chr.16: 19307835-19307835 on Build GRCh38
rs756904724 Chr.16: 19307628-19307628 on Build GRCh38
rs760013625 Chr.16: 19307817-19307817 on Build GRCh38
rs974258320 Chr.16: 19307795-19307795 on Build GRCh38
rs535612663 Chr.16: 19307834-19307834 on Build GRCh38
rs903941188 Chr.16: 19307610-19307610 on Build GRCh38
rs776857639 Chr.16: 19307517-19307517 on Build GRCh38
rs903909755 Chr.16: 19307927-19307927 on Build GRCh38
rs530418149 Chr.16: 19307854-19307854 on Build GRCh38
rs896488026 Chr.16: 19307597-19307597 on Build GRCh38
rs541529275 Chr.16: 19307771-19307771 on Build GRCh38
rs763933490 Chr.16: 19307625-19307625 on Build GRCh38
rs759575378 Chr.16: 19307596-19307596 on Build GRCh38
rs529713645 Chr.16: 19307768-19307768 on Build GRCh38
rs1024106920 Chr.16: 19307609-19307609 on Build GRCh38
rs200394651 Chr.16: 19307505-19307505 on Build GRCh38
rs985126198 Chr.16: 19307724-19307724 on Build GRCh38
rs867265990 Chr.16: 19307577-19307577 on Build GRCh38
rs541236113 Chr.16: 19307660-19307660 on Build GRCh38
rs747300389 Chr.16: 19307501-19307501 on Build GRCh38
rs753577991 Chr.16: 19307627-19307627 on Build GRCh38
rs759014906 Chr.16: 19307651-19307651 on Build GRCh38
rs989456358 Chr.16: 19307500-19307500 on Build GRCh38
rs202158142 Chr.16: 19307650-19307650 on Build GRCh38
rs906805270 Chr.16: 19307707-19307707 on Build GRCh38
rs575173744 Chr.16: 19307759-19307759 on Build GRCh38
rs769866015 Chr.16: 19307530-19307530 on Build GRCh38
rs990003880 Chr.16: 19307830-19307830 on Build GRCh38
rs767351080 Chr.16: 19307598-19307598 on Build GRCh38
rs577490903 Chr.16: 19307644-19307644 on Build GRCh38
rs764855371 Chr.16: 19307649-19307649 on Build GRCh38
rs958144679 Chr.16: 19307765-19307765 on Build GRCh38
rs1028981315 Chr.16: 19307839-19307839 on Build GRCh38
rs563091523 Chr.16: 19307813-19307813 on Build GRCh38
rs1056333047 Chr.16: 19307549-19307549 on Build GRCh38
rs1045092320 Chr.16: 19307902-19307902 on Build GRCh38
rs1031588680 Chr.16: 19307733-19307733 on Build GRCh38
rs1012437361 Chr.16: 19307685-19307685 on Build GRCh38
rs187163813 Chr.16: 19307480-19307480 on Build GRCh38
rs150196116 Chr.16: 19307620-19307620 on Build GRCh38
rs999613104 Chr.16: 19307708-19307708 on Build GRCh38

View Full Product Details