Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19535763-19536287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCP110
Entrez Gene ID
9738
Gene Name
centriolar coiled-coil protein 110
Gene Aliases
CP110, Cep110
UniGene
Hs.279912
Gene Chromosome Location
Chr.16: 19523857-19553408 on Build GRCh38
Transcripts
NM_001199022.2 NM_001323569.1 NM_001323570.1 NM_001323571.1 NM_001323572.1 NM_001323576.1 NM_001323577.1 NM_014711.5 XM_011545991.2 XM_011545992.2 XM_017023908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177639 c.286A>G p.N96D Chr.16: 19535955-19535955 on Build GRCh38
7218564 c.366A>G p.T122T Chr.16: 19536035-19536035 on Build GRCh38
7218565 c.512C>T p.P171L Chr.16: 19536181-19536181 on Build GRCh38
5833016 c.342C>T p.V114V Chr.16: 19536011-19536011 on Build GRCh38
1189197 c.274A>T p.R92* Chr.16: 19535943-19535943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775640437 Chr.16: 19536036-19536036 on Build GRCh38
rs756384441 Chr.16: 19535878-19535878 on Build GRCh38
rs919849177 Chr.16: 19536228-19536228 on Build GRCh38
rs756733996 Chr.16: 19536080-19536080 on Build GRCh38
rs780478690 Chr.16: 19536134-19536134 on Build GRCh38
rs865892894 Chr.16: 19536232-19536232 on Build GRCh38
rs372440972 Chr.16: 19536061-19536061 on Build GRCh38
rs751432098 Chr.16: 19535961-19535961 on Build GRCh38
rs766354418 Chr.16: 19535955-19535955 on Build GRCh38
rs768700653 Chr.16: 19536046-19536046 on Build GRCh38
rs750384183 Chr.16: 19535917-19535917 on Build GRCh38
rs138700121 Chr.16: 19536199-19536199 on Build GRCh38
rs767342328 Chr.16: 19536064-19536064 on Build GRCh38
rs10492785 Chr.16: 19536035-19536035 on Build GRCh38
rs775384062 Chr.16: 19536243-19536243 on Build GRCh38
rs779339856 Chr.16: 19536012-19536012 on Build GRCh38
rs752367436 Chr.16: 19535976-19535976 on Build GRCh38
rs779435583 Chr.16: 19536091-19536091 on Build GRCh38
rs777629097 Chr.16: 19536075-19536075 on Build GRCh38
rs537803910 Chr.16: 19535962-19535962 on Build GRCh38
rs977398588 Chr.16: 19535845-19535845 on Build GRCh38
rs367940434 Chr.16: 19535950-19535950 on Build GRCh38
rs3751821 Chr.16: 19536181-19536181 on Build GRCh38
rs763610593 Chr.16: 19536073-19536073 on Build GRCh38
rs76903460 Chr.16: 19536052-19536052 on Build GRCh38
rs867196928 Chr.16: 19535795-19535795 on Build GRCh38
rs750970706 Chr.16: 19535794-19535794 on Build GRCh38
rs760935454 Chr.16: 19536041-19536041 on Build GRCh38
rs753364001 Chr.16: 19536077-19536077 on Build GRCh38
rs780760120 Chr.16: 19535963-19535963 on Build GRCh38
rs769967582 Chr.16: 19536188-19536188 on Build GRCh38
rs771457131 Chr.16: 19535997-19535997 on Build GRCh38
rs759448367 Chr.16: 19536062-19536062 on Build GRCh38
rs957323293 Chr.16: 19535897-19535897 on Build GRCh38
rs545746288 Chr.16: 19536100-19536100 on Build GRCh38
rs768955574 Chr.16: 19536164-19536164 on Build GRCh38
rs758746089 Chr.16: 19536130-19536130 on Build GRCh38
rs368558634 Chr.16: 19536066-19536066 on Build GRCh38
rs561301333 Chr.16: 19535983-19535983 on Build GRCh38
rs748288173 Chr.16: 19536182-19536182 on Build GRCh38
rs868404897 Chr.16: 19536231-19536231 on Build GRCh38
rs763557377 Chr.16: 19535894-19535895 on Build GRCh38
rs779817862 Chr.16: 19536022-19536022 on Build GRCh38
rs543691187 Chr.16: 19535862-19535863 on Build GRCh38
rs774501610 Chr.16: 19536195-19536195 on Build GRCh38
rs1014784490 Chr.16: 19535788-19535788 on Build GRCh38
rs760336340 Chr.16: 19536256-19536256 on Build GRCh38
rs776778626 Chr.16: 19536047-19536047 on Build GRCh38
rs372309095 Chr.16: 19536247-19536247 on Build GRCh38
rs898162949 Chr.16: 19536192-19536192 on Build GRCh38
rs747271722 Chr.16: 19536154-19536154 on Build GRCh38
rs759506196 Chr.16: 19536201-19536201 on Build GRCh38
rs546964651 Chr.16: 19536054-19536054 on Build GRCh38
rs748726525 Chr.16: 19535985-19535985 on Build GRCh38
rs746277493 Chr.16: 19536014-19536014 on Build GRCh38
rs767302038 Chr.16: 19536239-19536239 on Build GRCh38
rs556529548 Chr.16: 19535996-19535996 on Build GRCh38
rs202061336 Chr.16: 19536185-19536185 on Build GRCh38
rs755649388 Chr.16: 19535982-19535982 on Build GRCh38

View Full Product Details