Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19536932-19537478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCP110
Entrez Gene ID
9738
Gene Name
centriolar coiled-coil protein 110
Gene Aliases
CP110, Cep110
UniGene
Hs.279912
Gene Chromosome Location
Chr.16: 19523857-19553408 on Build GRCh38
Transcripts
NM_001199022.2 NM_001323569.1 NM_001323570.1 NM_001323571.1 NM_001323572.1 NM_001323576.1 NM_001323577.1 NM_014711.5 XM_011545991.2 XM_011545992.2 XM_017023908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3999755 c.1679C>T p.T560M Chr.16: 19537348-19537348 on Build GRCh38
6143852 c.1762C>G p.L588V Chr.16: 19537431-19537431 on Build GRCh38
7218737 c.1299G>A p.A433A Chr.16: 19536968-19536968 on Build GRCh38
3999756 c.1749A>G p.K583K Chr.16: 19537418-19537418 on Build GRCh38
6078645 c.1659A>T p.L553F Chr.16: 19537328-19537328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141945021 Chr.16: 19537374-19537374 on Build GRCh38
rs763990973 Chr.16: 19537090-19537090 on Build GRCh38
rs540075055 Chr.16: 19537050-19537050 on Build GRCh38
rs770143429 Chr.16: 19537141-19537141 on Build GRCh38
rs759598287 Chr.16: 19537162-19537162 on Build GRCh38
rs982066152 Chr.16: 19537070-19537070 on Build GRCh38
rs746475205 Chr.16: 19536998-19536998 on Build GRCh38
rs752659133 Chr.16: 19537173-19537173 on Build GRCh38
rs368056132 Chr.16: 19537231-19537231 on Build GRCh38
rs1049791892 Chr.16: 19537167-19537167 on Build GRCh38
rs778206968 Chr.16: 19537223-19537223 on Build GRCh38
rs372951911 Chr.16: 19537425-19537425 on Build GRCh38
rs762126255 Chr.16: 19537025-19537025 on Build GRCh38
rs370713605 Chr.16: 19536967-19536967 on Build GRCh38
rs959147949 Chr.16: 19537316-19537316 on Build GRCh38
rs760622290 Chr.16: 19537300-19537300 on Build GRCh38
rs778493534 Chr.16: 19536978-19536978 on Build GRCh38
rs982722527 Chr.16: 19536970-19536970 on Build GRCh38
rs763775545 Chr.16: 19536992-19536992 on Build GRCh38
rs202085873 Chr.16: 19537128-19537128 on Build GRCh38
rs112924275 Chr.16: 19537380-19537380 on Build GRCh38
rs762909525 Chr.16: 19537319-19537319 on Build GRCh38
rs766525448 Chr.16: 19537073-19537073 on Build GRCh38
rs754505654 Chr.16: 19537209-19537209 on Build GRCh38
rs749014531 Chr.16: 19537295-19537295 on Build GRCh38
rs762998961 Chr.16: 19537426-19537426 on Build GRCh38
rs757042918 Chr.16: 19536977-19536977 on Build GRCh38
rs751672362 Chr.16: 19537078-19537078 on Build GRCh38
rs376813668 Chr.16: 19537323-19537323 on Build GRCh38
rs771168953 Chr.16: 19537157-19537157 on Build GRCh38
rs746176518 Chr.16: 19537382-19537382 on Build GRCh38
rs374937720 Chr.16: 19537454-19537454 on Build GRCh38
rs779977765 Chr.16: 19537221-19537221 on Build GRCh38
rs143488344 Chr.16: 19537294-19537294 on Build GRCh38
rs747547368 Chr.16: 19537219-19537219 on Build GRCh38
rs369267187 Chr.16: 19537378-19537378 on Build GRCh38
rs973873957 Chr.16: 19536957-19536957 on Build GRCh38
rs750174647 Chr.16: 19537110-19537110 on Build GRCh38
rs943712177 Chr.16: 19537285-19537285 on Build GRCh38
rs757700534 Chr.16: 19537371-19537371 on Build GRCh38
rs759205882 Chr.16: 19537449-19537449 on Build GRCh38
rs148151798 Chr.16: 19537329-19537329 on Build GRCh38
rs757056118 Chr.16: 19537097-19537097 on Build GRCh38
rs878978154 Chr.16: 19537311-19537311 on Build GRCh38
rs769066188 Chr.16: 19537003-19537003 on Build GRCh38
rs564520797 Chr.16: 19537169-19537169 on Build GRCh38
rs374077305 Chr.16: 19537015-19537015 on Build GRCh38
rs7191012 Chr.16: 19536968-19536968 on Build GRCh38
rs779714453 Chr.16: 19536996-19536996 on Build GRCh38
rs530011102 Chr.16: 19537298-19537298 on Build GRCh38
rs754653297 Chr.16: 19537309-19537310 on Build GRCh38
rs749705871 Chr.16: 19537229-19537229 on Build GRCh38
rs151214000 Chr.16: 19537204-19537204 on Build GRCh38
rs373594620 Chr.16: 19537193-19537193 on Build GRCh38
rs777173621 Chr.16: 19537014-19537014 on Build GRCh38
rs990509431 Chr.16: 19536976-19536976 on Build GRCh38
rs758153024 Chr.16: 19537127-19537127 on Build GRCh38
rs368395482 Chr.16: 19537308-19537308 on Build GRCh38
rs374650721 Chr.16: 19537349-19537349 on Build GRCh38
rs375241886 Chr.16: 19537303-19537303 on Build GRCh38
rs141967827 Chr.16: 19537058-19537058 on Build GRCh38
rs759616349 Chr.16: 19537086-19537086 on Build GRCh38
rs368563531 Chr.16: 19537346-19537346 on Build GRCh38
rs370519570 Chr.16: 19537154-19537154 on Build GRCh38
rs559174969 Chr.16: 19537210-19537210 on Build GRCh38
rs531944701 Chr.16: 19537189-19537189 on Build GRCh38
rs924868218 Chr.16: 19537394-19537394 on Build GRCh38
rs780934450 Chr.16: 19537321-19537323 on Build GRCh38
rs781615627 Chr.16: 19537140-19537140 on Build GRCh38
rs774490930 Chr.16: 19537253-19537253 on Build GRCh38
rs745500763 Chr.16: 19536987-19536987 on Build GRCh38
rs752277360 Chr.16: 19537336-19537336 on Build GRCh38
rs751220765 Chr.16: 19537205-19537205 on Build GRCh38
rs778860781 Chr.16: 19537105-19537105 on Build GRCh38
rs1015535512 Chr.16: 19537244-19537244 on Build GRCh38
rs772359627 Chr.16: 19537392-19537392 on Build GRCh38
rs746111903 Chr.16: 19537273-19537273 on Build GRCh38
rs766354286 Chr.16: 19537443-19537443 on Build GRCh38
rs754666121 Chr.16: 19537324-19537324 on Build GRCh38
rs200290881 Chr.16: 19536959-19536959 on Build GRCh38
rs950351653 Chr.16: 19537028-19537028 on Build GRCh38
rs912139030 Chr.16: 19537237-19537237 on Build GRCh38
rs753805350 Chr.16: 19537095-19537095 on Build GRCh38
rs78180055 Chr.16: 19537348-19537348 on Build GRCh38
rs945496261 Chr.16: 19537181-19537181 on Build GRCh38
rs775552163 Chr.16: 19537393-19537393 on Build GRCh38
rs749777679 Chr.16: 19537356-19537356 on Build GRCh38
rs371930526 Chr.16: 19537322-19537322 on Build GRCh38
rs770190968 Chr.16: 19537036-19537036 on Build GRCh38
rs17227190 Chr.16: 19537418-19537418 on Build GRCh38
rs34535184 Chr.16: 19536994-19536995 on Build GRCh38
rs184551421 Chr.16: 19537159-19537159 on Build GRCh38
rs774235433 Chr.16: 19537444-19537444 on Build GRCh38
rs780717752 Chr.16: 19537160-19537160 on Build GRCh38
rs202054670 Chr.16: 19537317-19537317 on Build GRCh38
rs62000395 Chr.16: 19537139-19537139 on Build GRCh38
rs138742987 Chr.16: 19537200-19537200 on Build GRCh38

View Full Product Details