Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19555333-19555832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf62
Entrez Gene ID
57020
Gene Name
chromosome 16 open reading frame 62
Gene Aliases
-
UniGene
Hs.654964
Gene Chromosome Location
Chr.16: 19555415-19701163 on Build GRCh38
Transcripts
NM_001300743.1 NM_020314.5 XM_005255435.3 XM_006721065.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6522613 c.237C>A p.G79G Chr.16: 19555699-19555699 on Build GRCh38
5765121 c.270G>A p.M90I Chr.16: 19555732-19555732 on Build GRCh38
3420811 c.274G>A p.V92I Chr.16: 19555736-19555736 on Build GRCh38
5136239 c.236G>T p.G79V Chr.16: 19555698-19555698 on Build GRCh38
3396225 c.166T>A p.C56S Chr.16: 19555628-19555628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039040245 Chr.16: 19555432-19555432 on Build GRCh38
rs34938118 Chr.16: 19555678-19555678 on Build GRCh38
rs780387786 Chr.16: 19555609-19555609 on Build GRCh38
rs774943538 Chr.16: 19555615-19555616 on Build GRCh38
rs769724922 Chr.16: 19555703-19555703 on Build GRCh38
rs752067498 Chr.16: 19555510-19555510 on Build GRCh38
rs374088599 Chr.16: 19555708-19555708 on Build GRCh38
rs910480412 Chr.16: 19555691-19555691 on Build GRCh38
rs1047828570 Chr.16: 19555712-19555712 on Build GRCh38
rs929333581 Chr.16: 19555557-19555557 on Build GRCh38
rs372470053 Chr.16: 19555760-19555760 on Build GRCh38
rs759110253 Chr.16: 19555717-19555717 on Build GRCh38
rs867245959 Chr.16: 19555635-19555635 on Build GRCh38
rs761735355 Chr.16: 19555652-19555652 on Build GRCh38
rs747073011 Chr.16: 19555742-19555742 on Build GRCh38
rs750039345 Chr.16: 19555608-19555608 on Build GRCh38
rs541824567 Chr.16: 19555599-19555599 on Build GRCh38
rs575934357 Chr.16: 19555629-19555629 on Build GRCh38
rs764999552 Chr.16: 19555606-19555606 on Build GRCh38
rs775781129 Chr.16: 19555678-19555680 on Build GRCh38
rs1020644876 Chr.16: 19555771-19555771 on Build GRCh38
rs943348756 Chr.16: 19555454-19555454 on Build GRCh38
rs781557759 Chr.16: 19555548-19555548 on Build GRCh38
rs902576645 Chr.16: 19555784-19555784 on Build GRCh38
rs1001310559 Chr.16: 19555806-19555806 on Build GRCh38
rs376976906 Chr.16: 19555783-19555783 on Build GRCh38
rs550845692 Chr.16: 19555414-19555414 on Build GRCh38
rs768028926 Chr.16: 19555801-19555801 on Build GRCh38
rs757834559 Chr.16: 19555476-19555476 on Build GRCh38
rs1055293349 Chr.16: 19555457-19555457 on Build GRCh38
rs755421962 Chr.16: 19555531-19555531 on Build GRCh38
rs771162544 Chr.16: 19555415-19555416 on Build GRCh38
rs746007729 Chr.16: 19555694-19555694 on Build GRCh38
rs536387080 Chr.16: 19555622-19555622 on Build GRCh38
rs913698675 Chr.16: 19555397-19555397 on Build GRCh38
rs781640702 Chr.16: 19555614-19555614 on Build GRCh38
rs568024484 Chr.16: 19555384-19555384 on Build GRCh38
rs920683192 Chr.16: 19555487-19555487 on Build GRCh38
rs141815613 Chr.16: 19555735-19555735 on Build GRCh38
rs954666671 Chr.16: 19555417-19555417 on Build GRCh38
rs745530052 Chr.16: 19555770-19555770 on Build GRCh38
rs528712095 Chr.16: 19555700-19555700 on Build GRCh38
rs1053424902 Chr.16: 19555752-19555752 on Build GRCh38
rs557981394 Chr.16: 19555474-19555474 on Build GRCh38
rs907549895 Chr.16: 19555416-19555416 on Build GRCh38
rs546057974 Chr.16: 19555644-19555644 on Build GRCh38
rs1009302021 Chr.16: 19555767-19555767 on Build GRCh38
rs564734737 Chr.16: 19555659-19555659 on Build GRCh38
rs528916617 Chr.16: 19555392-19555392 on Build GRCh38
rs768766413 Chr.16: 19555649-19555649 on Build GRCh38
rs557331847 Chr.16: 19555601-19555601 on Build GRCh38
rs534826883 Chr.16: 19555505-19555505 on Build GRCh38
rs28384469 Chr.16: 19555729-19555729 on Build GRCh38
rs758547936 Chr.16: 19555699-19555699 on Build GRCh38
rs564913396 Chr.16: 19555390-19555392 on Build GRCh38
rs762537217 Chr.16: 19555654-19555654 on Build GRCh38
rs1000858515 Chr.16: 19555577-19555577 on Build GRCh38
rs570839348 Chr.16: 19555497-19555497 on Build GRCh38
rs773979943 Chr.16: 19555716-19555716 on Build GRCh38
rs974582629 Chr.16: 19555362-19555362 on Build GRCh38
rs896622664 Chr.16: 19555464-19555464 on Build GRCh38
rs779259183 Chr.16: 19555680-19555680 on Build GRCh38
rs776700627 Chr.16: 19555651-19555651 on Build GRCh38
rs761106328 Chr.16: 19555732-19555732 on Build GRCh38
rs933062176 Chr.16: 19555404-19555404 on Build GRCh38
rs759024408 Chr.16: 19555662-19555662 on Build GRCh38
rs539245354 Chr.16: 19555448-19555448 on Build GRCh38
rs753109676 Chr.16: 19555615-19555615 on Build GRCh38
rs777194425 Chr.16: 19555788-19555788 on Build GRCh38
rs760714356 Chr.16: 19555594-19555594 on Build GRCh38
rs907600683 Chr.16: 19555688-19555688 on Build GRCh38
rs748050474 Chr.16: 19555702-19555702 on Build GRCh38
rs749094001 Chr.16: 19555764-19555764 on Build GRCh38
rs766013550 Chr.16: 19555656-19555656 on Build GRCh38
rs371895254 Chr.16: 19555674-19555674 on Build GRCh38
rs898730747 Chr.16: 19555437-19555437 on Build GRCh38
rs769105076 Chr.16: 19555787-19555787 on Build GRCh38
rs936227605 Chr.16: 19555745-19555745 on Build GRCh38
rs758636685 Chr.16: 19555738-19555738 on Build GRCh38
rs540301415 Chr.16: 19555704-19555704 on Build GRCh38
rs774419494 Chr.16: 19555587-19555587 on Build GRCh38
rs777834754 Chr.16: 19555576-19555576 on Build GRCh38
rs1013681445 Chr.16: 19555471-19555471 on Build GRCh38
rs767923304 Chr.16: 19555669-19555670 on Build GRCh38
rs771124906 Chr.16: 19555623-19555623 on Build GRCh38
rs771661595 Chr.16: 19555781-19555781 on Build GRCh38
rs567348736 Chr.16: 19555427-19555427 on Build GRCh38
rs770094959 Chr.16: 19555570-19555570 on Build GRCh38
rs748876424 Chr.16: 19555659-19555660 on Build GRCh38
rs774061490 Chr.16: 19555769-19555769 on Build GRCh38
rs756417011 Chr.16: 19555617-19555617 on Build GRCh38
rs751019017 Chr.16: 19555613-19555613 on Build GRCh38
rs757464044 Chr.16: 19555679-19555679 on Build GRCh38
rs987788229 Chr.16: 19555426-19555426 on Build GRCh38
rs574465944 Chr.16: 19555580-19555580 on Build GRCh38
rs769481307 Chr.16: 19555653-19555653 on Build GRCh38
rs764633066 Chr.16: 19555734-19555734 on Build GRCh38
rs1014536322 Chr.16: 19555600-19555600 on Build GRCh38
rs1047962802 Chr.16: 19555559-19555559 on Build GRCh38
rs749473804 Chr.16: 19555619-19555619 on Build GRCh38
rs770908089 Chr.16: 19555586-19555586 on Build GRCh38
rs569139906 Chr.16: 19555445-19555445 on Build GRCh38
rs553068199 Chr.16: 19555564-19555564 on Build GRCh38
rs745914141 Chr.16: 19555632-19555632 on Build GRCh38
rs753193693 Chr.16: 19555675-19555675 on Build GRCh38
rs764377791 Chr.16: 19555676-19555676 on Build GRCh38
rs767016030 Chr.16: 19555722-19555722 on Build GRCh38
rs367897965 Chr.16: 19555737-19555737 on Build GRCh38
rs890190279 Chr.16: 19555772-19555772 on Build GRCh38
rs770708900 Chr.16: 19555766-19555766 on Build GRCh38
rs756044419 Chr.16: 19555747-19555747 on Build GRCh38

View Full Product Details