Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20780663-20781162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Gene Symbol
ERI2
Entrez Gene ID
112479
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
EXOD1, ZGRF5
UniGene
Hs.248437
Gene Chromosome Location
Chr.16: 20780193-20806473 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464256 c.881A>C p.Q294P Chr.16: 20781072-20781072 on Build GRCh38
1129036 c.907C>G p.L303V Chr.16: 20780722-20780722 on Build GRCh38
7161155 c.924G>C p.R308S Chr.16: 20780705-20780705 on Build GRCh38
6464258 c.896T>A p.F299Y Chr.16: 20781087-20781087 on Build GRCh38
5584353 c.719G>A p.G240E Chr.16: 20780794-20780794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761988200 Chr.16: 20781064-20781064 on Build GRCh38
rs754136957 Chr.16: 20780829-20780829 on Build GRCh38
rs771689378 Chr.16: 20780688-20780688 on Build GRCh38
rs932377665 Chr.16: 20780714-20780714 on Build GRCh38
rs1037236770 Chr.16: 20780986-20780986 on Build GRCh38
rs138743502 Chr.16: 20780721-20780721 on Build GRCh38
rs761804163 Chr.16: 20780717-20780717 on Build GRCh38
rs774415529 Chr.16: 20781043-20781043 on Build GRCh38
rs770203321 Chr.16: 20780882-20780883 on Build GRCh38
rs769940054 Chr.16: 20781136-20781136 on Build GRCh38
rs539126073 Chr.16: 20781130-20781130 on Build GRCh38
rs957325185 Chr.16: 20780907-20780907 on Build GRCh38
rs998676429 Chr.16: 20780881-20780881 on Build GRCh38
rs746186908 Chr.16: 20780825-20780825 on Build GRCh38
rs564108348 Chr.16: 20781012-20781012 on Build GRCh38
rs747948727 Chr.16: 20780879-20780879 on Build GRCh38
rs763377838 Chr.16: 20781079-20781079 on Build GRCh38
rs769765337 Chr.16: 20780883-20780883 on Build GRCh38
rs757706508 Chr.16: 20780745-20780745 on Build GRCh38
rs892965749 Chr.16: 20781093-20781093 on Build GRCh38
rs912337898 Chr.16: 20780951-20780951 on Build GRCh38
rs752920558 Chr.16: 20780728-20780728 on Build GRCh38
rs553596829 Chr.16: 20780788-20780788 on Build GRCh38
rs758785123 Chr.16: 20780984-20780984 on Build GRCh38
rs1013644207 Chr.16: 20780789-20780789 on Build GRCh38
rs770713919 Chr.16: 20781037-20781037 on Build GRCh38
rs995904841 Chr.16: 20780756-20780756 on Build GRCh38
rs772031793 Chr.16: 20781122-20781122 on Build GRCh38
rs759689506 Chr.16: 20780719-20780719 on Build GRCh38
rs760505175 Chr.16: 20780698-20780698 on Build GRCh38
rs151064311 Chr.16: 20781120-20781120 on Build GRCh38
rs775380862 Chr.16: 20780908-20780908 on Build GRCh38
rs752931707 Chr.16: 20780809-20780809 on Build GRCh38
rs776521152 Chr.16: 20780791-20780791 on Build GRCh38
rs778037946 Chr.16: 20780739-20780739 on Build GRCh38
rs776896545 Chr.16: 20780964-20780964 on Build GRCh38
rs760766792 Chr.16: 20780815-20780815 on Build GRCh38
rs767542975 Chr.16: 20780806-20780806 on Build GRCh38
rs748461710 Chr.16: 20780771-20780772 on Build GRCh38
rs770930975 Chr.16: 20780903-20780903 on Build GRCh38
rs750834013 Chr.16: 20780974-20780974 on Build GRCh38
rs113477819 Chr.16: 20780694-20780694 on Build GRCh38
rs550153768 Chr.16: 20781114-20781114 on Build GRCh38
rs140383492 Chr.16: 20780734-20780734 on Build GRCh38
rs150171056 Chr.16: 20780690-20780690 on Build GRCh38
rs747048651 Chr.16: 20781059-20781059 on Build GRCh38
rs35235681 Chr.16: 20780827-20780828 on Build GRCh38
rs764296808 Chr.16: 20780936-20780936 on Build GRCh38
rs773261373 Chr.16: 20780887-20780887 on Build GRCh38
rs143915869 Chr.16: 20781025-20781025 on Build GRCh38
rs762144103 Chr.16: 20780970-20780970 on Build GRCh38
rs779380694 Chr.16: 20780854-20780854 on Build GRCh38
rs5715 Chr.16: 20780866-20780866 on Build GRCh38
rs780992713 Chr.16: 20780876-20780876 on Build GRCh38
rs779399195 Chr.16: 20780749-20780749 on Build GRCh38
rs755776881 Chr.16: 20781104-20781104 on Build GRCh38
rs372646398 Chr.16: 20781094-20781094 on Build GRCh38
rs905107168 Chr.16: 20780750-20780750 on Build GRCh38
rs145282601 Chr.16: 20780774-20780774 on Build GRCh38
rs7196188 Chr.16: 20781113-20781113 on Build GRCh38
rs773394496 Chr.16: 20781069-20781069 on Build GRCh38
rs998645565 Chr.16: 20781072-20781072 on Build GRCh38
rs768133999 Chr.16: 20780869-20780869 on Build GRCh38
rs757686503 Chr.16: 20780845-20780845 on Build GRCh38
rs763848035 Chr.16: 20780703-20780703 on Build GRCh38
rs1037267717 Chr.16: 20780733-20780733 on Build GRCh38
rs747848031 Chr.16: 20781016-20781016 on Build GRCh38
rs745397760 Chr.16: 20780686-20780686 on Build GRCh38
rs575803623 Chr.16: 20780915-20780915 on Build GRCh38
rs746553392 Chr.16: 20780867-20780867 on Build GRCh38
rs986541687 Chr.16: 20780949-20780949 on Build GRCh38
rs765286234 Chr.16: 20780718-20780718 on Build GRCh38
rs13306603 Chr.16: 20780999-20780999 on Build GRCh38
rs749129574 Chr.16: 20781026-20781026 on Build GRCh38
rs760814889 Chr.16: 20780934-20780934 on Build GRCh38
rs953855930 Chr.16: 20780943-20780943 on Build GRCh38
rs780975938 Chr.16: 20781014-20781014 on Build GRCh38
rs762837717 Chr.16: 20780899-20780899 on Build GRCh38
rs145742097 Chr.16: 20781103-20781103 on Build GRCh38
rs755942547 Chr.16: 20781024-20781024 on Build GRCh38
rs750977868 Chr.16: 20780858-20780858 on Build GRCh38
rs754230915 Chr.16: 20780744-20780744 on Build GRCh38
rs117563656 Chr.16: 20781095-20781095 on Build GRCh38
rs756477969 Chr.16: 20780731-20780731 on Build GRCh38
rs973646056 Chr.16: 20780954-20780954 on Build GRCh38
rs52817836 Chr.16: 20781066-20781066 on Build GRCh38
rs148354017 Chr.16: 20781063-20781063 on Build GRCh38
rs765634803 Chr.16: 20780973-20780973 on Build GRCh38
rs759644780 Chr.16: 20780801-20780801 on Build GRCh38
rs768389216 Chr.16: 20780790-20780790 on Build GRCh38
rs761711838 Chr.16: 20780795-20780795 on Build GRCh38
rs757774573 Chr.16: 20780764-20780764 on Build GRCh38
rs772738496 Chr.16: 20781053-20781054 on Build GRCh38
rs137879235 Chr.16: 20780800-20780800 on Build GRCh38
rs776646668 Chr.16: 20781125-20781125 on Build GRCh38
rs776682176 Chr.16: 20780710-20780710 on Build GRCh38
rs542203778 Chr.16: 20780863-20780863 on Build GRCh38
rs752162811 Chr.16: 20781080-20781080 on Build GRCh38
rs370138574 Chr.16: 20780994-20780994 on Build GRCh38
rs757135564 Chr.16: 20781115-20781115 on Build GRCh38
rs768773188 Chr.16: 20781062-20781062 on Build GRCh38
rs1042186936 Chr.16: 20781105-20781105 on Build GRCh38
rs745379309 Chr.16: 20780751-20780751 on Build GRCh38

View Full Product Details